Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109167809-109168312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACACB
Entrez Gene ID
32
Gene Name
acetyl-CoA carboxylase beta
Gene Aliases
ACC2, ACCB, HACC275
UniGene
Hs.234898
Gene Chromosome Location
Chr.12: 109112666-109268226 on Build GRCh38
Transcripts
NM_001093.3 XM_005253876.4 XM_006719367.3 XM_011538259.2 XM_011538263.2 XM_011538264.2 XM_011538265.2 XM_017019252.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5882178 c.812C>T p.A271V Chr.12: 109167921-109167921 on Build GRCh38
278567 c.830G>A p.R277H Chr.12: 109167939-109167939 on Build GRCh38
1946163 c.922G>A p.A308T Chr.12: 109168031-109168031 on Build GRCh38
5010214 c.816C>T p.A272A Chr.12: 109167925-109167925 on Build GRCh38
1946160 c.829C>T p.R277C Chr.12: 109167938-109167938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147825347 Chr.12: 109168015-109168015 on Build GRCh38
rs748125910 Chr.12: 109167991-109167991 on Build GRCh38
rs890825033 Chr.12: 109168065-109168065 on Build GRCh38
rs144382279 Chr.12: 109167989-109167989 on Build GRCh38
rs990365203 Chr.12: 109168003-109168003 on Build GRCh38
rs77034837 Chr.12: 109168290-109168290 on Build GRCh38
rs34286 Chr.12: 109168191-109168191 on Build GRCh38
rs369281773 Chr.12: 109167828-109167828 on Build GRCh38
rs766077286 Chr.12: 109167881-109167881 on Build GRCh38
rs758231402 Chr.12: 109168066-109168066 on Build GRCh38
rs80074286 Chr.12: 109168130-109168130 on Build GRCh38
rs764163663 Chr.12: 109167864-109167864 on Build GRCh38
rs775526769 Chr.12: 109167847-109167847 on Build GRCh38
rs376649368 Chr.12: 109168088-109168088 on Build GRCh38
rs990202139 Chr.12: 109168238-109168238 on Build GRCh38
rs956049041 Chr.12: 109168223-109168223 on Build GRCh38
rs960763115 Chr.12: 109168148-109168148 on Build GRCh38
rs763571331 Chr.12: 109168013-109168013 on Build GRCh38
rs754737585 Chr.12: 109167948-109167948 on Build GRCh38
rs759233676 Chr.12: 109168024-109168024 on Build GRCh38
rs374025754 Chr.12: 109168052-109168052 on Build GRCh38
rs757251180 Chr.12: 109168041-109168041 on Build GRCh38
rs781228613 Chr.12: 109168067-109168067 on Build GRCh38
rs758097574 Chr.12: 109167879-109167879 on Build GRCh38
rs778646669 Chr.12: 109168048-109168048 on Build GRCh38
rs78715638 Chr.12: 109168031-109168031 on Build GRCh38
rs751203396 Chr.12: 109167943-109167943 on Build GRCh38
rs777136804 Chr.12: 109167865-109167865 on Build GRCh38
rs780584088 Chr.12: 109167897-109167897 on Build GRCh38
rs751291356 Chr.12: 109167893-109167893 on Build GRCh38
rs768707560 Chr.12: 109167926-109167926 on Build GRCh38
rs34285 Chr.12: 109168093-109168093 on Build GRCh38
rs554442545 Chr.12: 109167837-109167837 on Build GRCh38
rs148414563 Chr.12: 109167922-109167922 on Build GRCh38
rs752191018 Chr.12: 109167949-109167949 on Build GRCh38
rs374378849 Chr.12: 109167850-109167850 on Build GRCh38
rs747227529 Chr.12: 109167983-109167983 on Build GRCh38
rs553898952 Chr.12: 109167913-109167913 on Build GRCh38
rs201698181 Chr.12: 109168078-109168078 on Build GRCh38
rs368005004 Chr.12: 109167936-109167936 on Build GRCh38
rs777523810 Chr.12: 109167972-109167972 on Build GRCh38
rs781305786 Chr.12: 109167990-109167990 on Build GRCh38
rs765627578 Chr.12: 109167869-109167869 on Build GRCh38
rs774926883 Chr.12: 109168017-109168017 on Build GRCh38
rs573637382 Chr.12: 109168004-109168004 on Build GRCh38
rs763672231 Chr.12: 109168032-109168032 on Build GRCh38
rs775622765 Chr.12: 109167923-109167923 on Build GRCh38
rs761581589 Chr.12: 109167968-109167968 on Build GRCh38
rs750791232 Chr.12: 109167871-109167871 on Build GRCh38
rs772321705 Chr.12: 109167846-109167846 on Build GRCh38
rs1002932158 Chr.12: 109168192-109168192 on Build GRCh38
rs370590640 Chr.12: 109167938-109167938 on Build GRCh38
rs548728398 Chr.12: 109168133-109168133 on Build GRCh38
rs4766516 Chr.12: 109167925-109167925 on Build GRCh38
rs748106611 Chr.12: 109167898-109167898 on Build GRCh38
rs773170756 Chr.12: 109168129-109168129 on Build GRCh38
rs756987109 Chr.12: 109167977-109167977 on Build GRCh38
rs954342022 Chr.12: 109168262-109168262 on Build GRCh38
rs1022290728 Chr.12: 109168261-109168261 on Build GRCh38
rs776355369 Chr.12: 109167871-109167873 on Build GRCh38
rs767124185 Chr.12: 109168029-109168029 on Build GRCh38
rs753759964 Chr.12: 109168038-109168038 on Build GRCh38
rs187596384 Chr.12: 109167838-109167838 on Build GRCh38
rs577886748 Chr.12: 109168087-109168087 on Build GRCh38
rs764952631 Chr.12: 109168039-109168040 on Build GRCh38
rs897061645 Chr.12: 109167836-109167836 on Build GRCh38
rs533919628 Chr.12: 109167914-109167914 on Build GRCh38
rs559963888 Chr.12: 109168098-109168098 on Build GRCh38
rs778738335 Chr.12: 109167981-109167981 on Build GRCh38
rs752418463 Chr.12: 109168030-109168030 on Build GRCh38
rs996341898 Chr.12: 109167875-109167875 on Build GRCh38
rs749130358 Chr.12: 109167921-109167921 on Build GRCh38
rs201630101 Chr.12: 109168023-109168023 on Build GRCh38
rs756056275 Chr.12: 109167970-109167970 on Build GRCh38
rs773787420 Chr.12: 109167939-109167939 on Build GRCh38
rs531807144 Chr.12: 109168197-109168197 on Build GRCh38
rs754475585 Chr.12: 109167896-109167896 on Build GRCh38
rs781030070 Chr.12: 109168210-109168210 on Build GRCh38
rs1048796144 Chr.12: 109168119-109168119 on Build GRCh38
rs763366636 Chr.12: 109167942-109167942 on Build GRCh38
rs1014002025 Chr.12: 109168166-109168169 on Build GRCh38
rs762283633 Chr.12: 109167868-109167868 on Build GRCh38
rs150967404 Chr.12: 109167976-109167976 on Build GRCh38
rs776640811 Chr.12: 109167933-109167933 on Build GRCh38

View Full Product Details