Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110485056-110485594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM216A
Entrez Gene ID
29902
Gene Name
family with sequence similarity 216 member A
Gene Aliases
C12orf24, HSU79274
UniGene
Hs.436618
Gene Chromosome Location
Chr.12: 110468424-110490387 on Build GRCh38
Transcripts
NM_013300.2 XM_005253875.4 XM_011538246.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
467781 c.190A>G p.I64V Chr.12: 110485083-110485083 on Build GRCh38
1947121 c.293C>T p.A98V Chr.12: 110485186-110485186 on Build GRCh38
5716632 c.242G>A p.W81* Chr.12: 110485135-110485135 on Build GRCh38
7167283 c.235G>A p.E79K Chr.12: 110485128-110485128 on Build GRCh38
1746745 c.283A>G p.M95V Chr.12: 110485176-110485176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924279616 Chr.12: 110485103-110485103 on Build GRCh38
rs377189568 Chr.12: 110485511-110485511 on Build GRCh38
rs535361681 Chr.12: 110485557-110485557 on Build GRCh38
rs898488135 Chr.12: 110485542-110485542 on Build GRCh38
rs748720351 Chr.12: 110485109-110485109 on Build GRCh38
rs759101296 Chr.12: 110485134-110485134 on Build GRCh38
rs750512068 Chr.12: 110485523-110485523 on Build GRCh38
rs775104878 Chr.12: 110485151-110485151 on Build GRCh38
rs771136874 Chr.12: 110485117-110485117 on Build GRCh38
rs1043262494 Chr.12: 110485163-110485163 on Build GRCh38
rs1051345045 Chr.12: 110485474-110485474 on Build GRCh38
rs753726329 Chr.12: 110485430-110485430 on Build GRCh38
rs778350925 Chr.12: 110485187-110485187 on Build GRCh38
rs974737216 Chr.12: 110485389-110485389 on Build GRCh38
rs761641882 Chr.12: 110485226-110485226 on Build GRCh38
rs561724594 Chr.12: 110485263-110485263 on Build GRCh38
rs369801733 Chr.12: 110485224-110485224 on Build GRCh38
rs76211437 Chr.12: 110485290-110485290 on Build GRCh38
rs781235800 Chr.12: 110485097-110485097 on Build GRCh38
rs932547786 Chr.12: 110485459-110485459 on Build GRCh38
rs750153312 Chr.12: 110485231-110485231 on Build GRCh38
rs142652628 Chr.12: 110485380-110485380 on Build GRCh38
rs924269570 Chr.12: 110485371-110485371 on Build GRCh38
rs1014147293 Chr.12: 110485316-110485316 on Build GRCh38
rs765310739 Chr.12: 110485465-110485465 on Build GRCh38
rs995471702 Chr.12: 110485567-110485568 on Build GRCh38
rs895563141 Chr.12: 110485514-110485514 on Build GRCh38
rs771388561 Chr.12: 110485200-110485200 on Build GRCh38
rs749702912 Chr.12: 110485196-110485196 on Build GRCh38
rs151005221 Chr.12: 110485484-110485484 on Build GRCh38
rs757078138 Chr.12: 110485161-110485161 on Build GRCh38
rs921950419 Chr.12: 110485397-110485397 on Build GRCh38
rs147340116 Chr.12: 110485341-110485341 on Build GRCh38
rs755062314 Chr.12: 110485079-110485079 on Build GRCh38
rs921172394 Chr.12: 110485142-110485142 on Build GRCh38
rs755188957 Chr.12: 110485179-110485179 on Build GRCh38
rs764146051 Chr.12: 110485158-110485158 on Build GRCh38
rs142328790 Chr.12: 110485159-110485159 on Build GRCh38
rs764882117 Chr.12: 110485164-110485164 on Build GRCh38
rs142702785 Chr.12: 110485207-110485207 on Build GRCh38
rs760740481 Chr.12: 110485155-110485155 on Build GRCh38
rs376976483 Chr.12: 110485186-110485186 on Build GRCh38
rs200632805 Chr.12: 110485203-110485203 on Build GRCh38
rs995125630 Chr.12: 110485303-110485303 on Build GRCh38
rs939893839 Chr.12: 110485504-110485504 on Build GRCh38
rs569324365 Chr.12: 110485320-110485320 on Build GRCh38
rs868580882 Chr.12: 110485204-110485204 on Build GRCh38
rs373871530 Chr.12: 110485227-110485227 on Build GRCh38
rs751865123 Chr.12: 110485167-110485167 on Build GRCh38
rs188622256 Chr.12: 110485214-110485214 on Build GRCh38
rs76614119 Chr.12: 110485357-110485357 on Build GRCh38
rs773615954 Chr.12: 110485164-110485165 on Build GRCh38
rs781298202 Chr.12: 110485180-110485180 on Build GRCh38
rs369985489 Chr.12: 110485216-110485216 on Build GRCh38
rs995490415 Chr.12: 110485555-110485555 on Build GRCh38
rs771822871 Chr.12: 110485206-110485206 on Build GRCh38
rs534292044 Chr.12: 110485443-110485443 on Build GRCh38
rs551270420 Chr.12: 110485304-110485304 on Build GRCh38
rs370898712 Chr.12: 110485250-110485250 on Build GRCh38
rs753939820 Chr.12: 110485210-110485211 on Build GRCh38
rs748585966 Chr.12: 110485113-110485113 on Build GRCh38
rs770265412 Chr.12: 110485112-110485112 on Build GRCh38
rs762687139 Chr.12: 110485237-110485237 on Build GRCh38
rs1013885135 Chr.12: 110485255-110485255 on Build GRCh38
rs7956133 Chr.12: 110485124-110485124 on Build GRCh38
rs748250217 Chr.12: 110485185-110485185 on Build GRCh38
rs983586082 Chr.12: 110485092-110485092 on Build GRCh38
rs1022621069 Chr.12: 110485271-110485271 on Build GRCh38
rs767207006 Chr.12: 110485148-110485148 on Build GRCh38
rs759094330 Chr.12: 110485202-110485202 on Build GRCh38
rs770349155 Chr.12: 110485114-110485114 on Build GRCh38
rs1051004987 Chr.12: 110485188-110485188 on Build GRCh38
rs539886809 Chr.12: 110485251-110485251 on Build GRCh38
rs989888744 Chr.12: 110485427-110485427 on Build GRCh38

View Full Product Details