Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 21539367-21539889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICU2
Entrez Gene ID
221154
Gene Name
mitochondrial calcium uptake 2
Gene Aliases
1110008L20Rik, EFHA1
UniGene
Hs.412103
Gene Chromosome Location
Chr.13: 21492689-21604216 on Build GRCh38
Transcripts
NM_152726.2 XM_017020433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3467669 c.361A>T p.K121* Chr.13: 21539686-21539686 on Build GRCh38
280731 c.396C>T p.I132I Chr.13: 21539372-21539372 on Build GRCh38
7297450 c.387A>G p.K129K Chr.13: 21539660-21539660 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767642955 Chr.13: 21539426-21539426 on Build GRCh38
rs188416133 Chr.13: 21539841-21539841 on Build GRCh38
rs892815696 Chr.13: 21539849-21539849 on Build GRCh38
rs941660599 Chr.13: 21539455-21539455 on Build GRCh38
rs1000841539 Chr.13: 21539500-21539500 on Build GRCh38
rs990485936 Chr.13: 21539473-21539473 on Build GRCh38
rs756430115 Chr.13: 21539701-21539701 on Build GRCh38
rs750655143 Chr.13: 21539607-21539607 on Build GRCh38
rs780710061 Chr.13: 21539738-21539738 on Build GRCh38
rs963245962 Chr.13: 21539638-21539638 on Build GRCh38
rs373449163 Chr.13: 21539726-21539726 on Build GRCh38
rs776314628 Chr.13: 21539637-21539637 on Build GRCh38
rs761852383 Chr.13: 21539658-21539658 on Build GRCh38
rs201089587 Chr.13: 21539640-21539640 on Build GRCh38
rs758209126 Chr.13: 21539718-21539718 on Build GRCh38
rs542910372 Chr.13: 21539519-21539520 on Build GRCh38
rs751651127 Chr.13: 21539423-21539423 on Build GRCh38
rs1014010021 Chr.13: 21539593-21539593 on Build GRCh38
rs537781806 Chr.13: 21539589-21539589 on Build GRCh38
rs566704548 Chr.13: 21539491-21539491 on Build GRCh38
rs746943380 Chr.13: 21539732-21539732 on Build GRCh38
rs564523568 Chr.13: 21539519-21539519 on Build GRCh38
rs532882116 Chr.13: 21539556-21539556 on Build GRCh38
rs910362172 Chr.13: 21539476-21539476 on Build GRCh38
rs566453981 Chr.13: 21539582-21539582 on Build GRCh38
rs554434141 Chr.13: 21539564-21539564 on Build GRCh38
rs186603558 Chr.13: 21539584-21539584 on Build GRCh38
rs925482982 Chr.13: 21539775-21539775 on Build GRCh38
rs752546254 Chr.13: 21539716-21539716 on Build GRCh38
rs1046583284 Chr.13: 21539768-21539768 on Build GRCh38
rs550611729 Chr.13: 21539449-21539449 on Build GRCh38
rs1025017546 Chr.13: 21539595-21539596 on Build GRCh38
rs909140319 Chr.13: 21539434-21539434 on Build GRCh38
rs551407448 Chr.13: 21539577-21539577 on Build GRCh38
rs540482215 Chr.13: 21539534-21539534 on Build GRCh38
rs1046844161 Chr.13: 21539857-21539857 on Build GRCh38
rs368474294 Chr.13: 21539405-21539405 on Build GRCh38
rs987204552 Chr.13: 21539553-21539553 on Build GRCh38
rs558194360 Chr.13: 21539773-21539773 on Build GRCh38
rs754864018 Chr.13: 21539614-21539614 on Build GRCh38
rs866773097 Chr.13: 21539684-21539684 on Build GRCh38
rs370440530 Chr.13: 21539737-21539737 on Build GRCh38
rs1039910033 Chr.13: 21539580-21539580 on Build GRCh38
rs942000179 Chr.13: 21539713-21539713 on Build GRCh38
rs377404805 Chr.13: 21539692-21539692 on Build GRCh38
rs373098990 Chr.13: 21539508-21539508 on Build GRCh38
rs191794298 Chr.13: 21539588-21539588 on Build GRCh38
rs910275252 Chr.13: 21539743-21539743 on Build GRCh38
rs933058164 Chr.13: 21539545-21539545 on Build GRCh38
rs778817593 Chr.13: 21539495-21539495 on Build GRCh38
rs182335660 Chr.13: 21539546-21539546 on Build GRCh38
rs184070439 Chr.13: 21539824-21539824 on Build GRCh38
rs546322071 Chr.13: 21539509-21539509 on Build GRCh38
rs1053257524 Chr.13: 21539848-21539848 on Build GRCh38
rs529635777 Chr.13: 21539541-21539541 on Build GRCh38
rs775244851 Chr.13: 21539572-21539572 on Build GRCh38
rs555161727 Chr.13: 21539587-21539587 on Build GRCh38
rs202246015 Chr.13: 21539629-21539629 on Build GRCh38
rs533750734 Chr.13: 21539492-21539492 on Build GRCh38
rs575642685 Chr.13: 21539639-21539639 on Build GRCh38
rs757358143 Chr.13: 21539425-21539425 on Build GRCh38
rs2798281 Chr.13: 21539496-21539496 on Build GRCh38
rs550220127 Chr.13: 21539502-21539502 on Build GRCh38
rs538994318 Chr.13: 21539536-21539536 on Build GRCh38
rs909214887 Chr.13: 21539504-21539504 on Build GRCh38
rs936896506 Chr.13: 21539803-21539803 on Build GRCh38
rs767677069 Chr.13: 21539662-21539662 on Build GRCh38
rs770460970 Chr.13: 21539631-21539631 on Build GRCh38
rs774913545 Chr.13: 21539656-21539656 on Build GRCh38
rs778885214 Chr.13: 21539617-21539617 on Build GRCh38
rs567769366 Chr.13: 21539489-21539489 on Build GRCh38
rs535705208 Chr.13: 21539619-21539619 on Build GRCh38
rs568906199 Chr.13: 21539549-21539549 on Build GRCh38
rs975872950 Chr.13: 21539605-21539605 on Build GRCh38
rs568944449 Chr.13: 21539459-21539459 on Build GRCh38
rs923384727 Chr.13: 21539494-21539494 on Build GRCh38
rs758283948 Chr.13: 21539624-21539624 on Build GRCh38
rs539550377 Chr.13: 21539482-21539482 on Build GRCh38
rs535402756 Chr.13: 21539444-21539444 on Build GRCh38
rs766653605 Chr.13: 21539715-21539715 on Build GRCh38
rs188776728 Chr.13: 21539505-21539505 on Build GRCh38
rs200426098 Chr.13: 21539409-21539409 on Build GRCh38
rs949586781 Chr.13: 21539528-21539528 on Build GRCh38
rs949636990 Chr.13: 21539746-21539746 on Build GRCh38
rs933112882 Chr.13: 21539477-21539477 on Build GRCh38
rs991097031 Chr.13: 21539719-21539719 on Build GRCh38
rs34005684 Chr.13: 21539530-21539530 on Build GRCh38
rs777700192 Chr.13: 21539626-21539626 on Build GRCh38
rs193249746 Chr.13: 21539540-21539540 on Build GRCh38
rs541789924 Chr.13: 21539503-21539503 on Build GRCh38
rs373155264 Chr.13: 21539594-21539594 on Build GRCh38
rs745330585 Chr.13: 21539740-21539740 on Build GRCh38

View Full Product Details