Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11747335-11747843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGTRAP
Entrez Gene ID
57085
Gene Name
angiotensin II receptor associated protein
Gene Aliases
ATRAP
UniGene
Hs.464438
Gene Chromosome Location
Chr.1: 11736085-11750771 on Build GRCh38
Transcripts
NM_001040194.1 NM_001040195.1 NM_001040196.1 NM_001040197.1 NM_020350.4 XM_011541799.2 XM_011541800.2 XM_011541802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3471989 c.63G>A p.W21* Chr.1: 11747440-11747440 on Build GRCh38
894526 c.154G>T p.D52Y Chr.1: 11747531-11747531 on Build GRCh38
6313874 c.212G>T p.G71V Chr.1: 11747492-11747492 on Build GRCh38
7354847 c.166A>G p.M56V Chr.1: 11747543-11747543 on Build GRCh38
369711 c.63G>T p.W21C Chr.1: 11747440-11747440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17875945 Chr.1: 11747370-11747370 on Build GRCh38
rs771850377 Chr.1: 11747526-11747526 on Build GRCh38
rs952627812 Chr.1: 11747634-11747634 on Build GRCh38
rs753490228 Chr.1: 11747745-11747745 on Build GRCh38
rs761607183 Chr.1: 11747447-11747447 on Build GRCh38
rs202125743 Chr.1: 11747458-11747458 on Build GRCh38
rs759145116 Chr.1: 11747541-11747541 on Build GRCh38
rs932250102 Chr.1: 11747767-11747767 on Build GRCh38
rs200088833 Chr.1: 11747519-11747519 on Build GRCh38
rs528955240 Chr.1: 11747585-11747585 on Build GRCh38
rs184958903 Chr.1: 11747416-11747416 on Build GRCh38
rs753003693 Chr.1: 11747400-11747400 on Build GRCh38
rs781089026 Chr.1: 11747415-11747415 on Build GRCh38
rs770017595 Chr.1: 11747528-11747528 on Build GRCh38
rs944901638 Chr.1: 11747622-11747622 on Build GRCh38
rs993993233 Chr.1: 11747616-11747616 on Build GRCh38
rs535211564 Chr.1: 11747373-11747373 on Build GRCh38
rs779891933 Chr.1: 11747424-11747424 on Build GRCh38
rs1023338584 Chr.1: 11747716-11747716 on Build GRCh38
rs758327465 Chr.1: 11747407-11747407 on Build GRCh38
rs756327887 Chr.1: 11747506-11747506 on Build GRCh38
rs1051074035 Chr.1: 11747602-11747602 on Build GRCh38
rs935560179 Chr.1: 11747362-11747362 on Build GRCh38
rs981297563 Chr.1: 11747736-11747736 on Build GRCh38
rs552639085 Chr.1: 11747818-11747818 on Build GRCh38
rs764388746 Chr.1: 11747500-11747500 on Build GRCh38
rs778219177 Chr.1: 11747412-11747412 on Build GRCh38
rs774325451 Chr.1: 11747445-11747445 on Build GRCh38
rs760195806 Chr.1: 11747468-11747468 on Build GRCh38
rs17875946 Chr.1: 11747639-11747639 on Build GRCh38
rs762295271 Chr.1: 11747551-11747551 on Build GRCh38
rs970829349 Chr.1: 11747642-11747642 on Build GRCh38
rs749572710 Chr.1: 11747513-11747513 on Build GRCh38
rs562373594 Chr.1: 11747674-11747674 on Build GRCh38
rs375235317 Chr.1: 11747398-11747398 on Build GRCh38
rs771971566 Chr.1: 11747584-11747584 on Build GRCh38
rs766005493 Chr.1: 11747484-11747484 on Build GRCh38
rs768052027 Chr.1: 11747623-11747623 on Build GRCh38
rs141555536 Chr.1: 11747442-11747442 on Build GRCh38
rs768613161 Chr.1: 11747443-11747443 on Build GRCh38
rs17875947 Chr.1: 11747702-11747702 on Build GRCh38
rs746794742 Chr.1: 11747501-11747501 on Build GRCh38
rs561506896 Chr.1: 11747490-11747490 on Build GRCh38
rs956792081 Chr.1: 11747819-11747819 on Build GRCh38
rs889371602 Chr.1: 11747603-11747603 on Build GRCh38
rs375907716 Chr.1: 11747510-11747510 on Build GRCh38
rs961311251 Chr.1: 11747409-11747409 on Build GRCh38
rs1027289210 Chr.1: 11747698-11747698 on Build GRCh38
rs76425954 Chr.1: 11747402-11747402 on Build GRCh38
rs747983763 Chr.1: 11747522-11747522 on Build GRCh38
rs545806751 Chr.1: 11747548-11747548 on Build GRCh38
rs761053816 Chr.1: 11747694-11747694 on Build GRCh38
rs770083300 Chr.1: 11747428-11747428 on Build GRCh38
rs766985189 Chr.1: 11747396-11747396 on Build GRCh38
rs183704861 Chr.1: 11747581-11747581 on Build GRCh38
rs1049419629 Chr.1: 11747803-11747803 on Build GRCh38
rs760025737 Chr.1: 11747577-11747577 on Build GRCh38
rs144060195 Chr.1: 11747543-11747543 on Build GRCh38
rs139457681 Chr.1: 11747569-11747569 on Build GRCh38
rs772533948 Chr.1: 11747466-11747466 on Build GRCh38
rs1008647746 Chr.1: 11747448-11747448 on Build GRCh38
rs753525447 Chr.1: 11747489-11747489 on Build GRCh38
rs146377549 Chr.1: 11747520-11747520 on Build GRCh38
rs775910675 Chr.1: 11747434-11747434 on Build GRCh38
rs1012698545 Chr.1: 11747639-11747639 on Build GRCh38
rs200291217 Chr.1: 11747552-11747552 on Build GRCh38
rs755185908 Chr.1: 11747576-11747576 on Build GRCh38
rs115781535 Chr.1: 11747397-11747397 on Build GRCh38
rs11578987 Chr.1: 11747558-11747558 on Build GRCh38
rs993184105 Chr.1: 11747693-11747693 on Build GRCh38
rs1011555037 Chr.1: 11747422-11747422 on Build GRCh38
rs371460221 Chr.1: 11747414-11747414 on Build GRCh38
rs746896821 Chr.1: 11747587-11747587 on Build GRCh38
rs12752700 Chr.1: 11747369-11747369 on Build GRCh38
rs758738252 Chr.1: 11747497-11747497 on Build GRCh38
rs993152131 Chr.1: 11747482-11747482 on Build GRCh38
rs372056053 Chr.1: 11747579-11747579 on Build GRCh38
rs745920945 Chr.1: 11747503-11747503 on Build GRCh38
rs971326838 Chr.1: 11747733-11747733 on Build GRCh38
rs570994766 Chr.1: 11747372-11747372 on Build GRCh38
rs201093684 Chr.1: 11747534-11747534 on Build GRCh38
rs760177353 Chr.1: 11747531-11747531 on Build GRCh38
rs1012582106 Chr.1: 11747705-11747705 on Build GRCh38
rs1011216253 Chr.1: 11747371-11747371 on Build GRCh38

View Full Product Details