Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11750399-11750906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGTRAP
Entrez Gene ID
57085
Gene Name
angiotensin II receptor associated protein
Gene Aliases
ATRAP
UniGene
Hs.464438
Gene Chromosome Location
Chr.1: 11736085-11750771 on Build GRCh38
Transcripts
NM_001040194.1 NM_001040195.1 NM_001040196.1 NM_001040197.1 NM_020350.4 XM_011541799.2 XM_011541800.2 XM_011541802.1

SNP Detail

SNP ID Chromosome Location
rs1014328971 Chr.1: 11750493-11750493 on Build GRCh38
rs893199015 Chr.1: 11750849-11750849 on Build GRCh38
rs773431143 Chr.1: 11750671-11750671 on Build GRCh38
rs1014647294 Chr.1: 11750878-11750878 on Build GRCh38
rs192750191 Chr.1: 11750532-11750532 on Build GRCh38
rs940546071 Chr.1: 11750716-11750716 on Build GRCh38
rs12064912 Chr.1: 11750715-11750715 on Build GRCh38
rs12955 Chr.1: 11750435-11750435 on Build GRCh38
rs979299098 Chr.1: 11750561-11750561 on Build GRCh38
rs1033232222 Chr.1: 11750602-11750602 on Build GRCh38
rs555047072 Chr.1: 11750559-11750559 on Build GRCh38
rs919660455 Chr.1: 11750713-11750713 on Build GRCh38
rs998918964 Chr.1: 11750598-11750598 on Build GRCh38
rs937224535 Chr.1: 11750795-11750795 on Build GRCh38
rs1128065 Chr.1: 11750634-11750634 on Build GRCh38
rs547940822 Chr.1: 11750723-11750723 on Build GRCh38
rs949252751 Chr.1: 11750731-11750731 on Build GRCh38
rs375481278 Chr.1: 11750815-11750815 on Build GRCh38
rs1033432173 Chr.1: 11750603-11750603 on Build GRCh38
rs11121821 Chr.1: 11750787-11750787 on Build GRCh38
rs1033133727 Chr.1: 11750848-11750848 on Build GRCh38
rs367931219 Chr.1: 11750610-11750610 on Build GRCh38
rs1028256461 Chr.1: 11750875-11750875 on Build GRCh38
rs773861482 Chr.1: 11750797-11750797 on Build GRCh38
rs1128084 Chr.1: 11750712-11750712 on Build GRCh38
rs967877339 Chr.1: 11750592-11750592 on Build GRCh38
rs1041808499 Chr.1: 11750879-11750879 on Build GRCh38
rs530496330 Chr.1: 11750864-11750864 on Build GRCh38
rs6540993 Chr.1: 11750708-11750708 on Build GRCh38
rs780352993 Chr.1: 11750736-11750736 on Build GRCh38
rs1128055 Chr.1: 11750614-11750614 on Build GRCh38
rs530765354 Chr.1: 11750437-11750438 on Build GRCh38
rs14962 Chr.1: 11750574-11750574 on Build GRCh38
rs1037533842 Chr.1: 11750775-11750775 on Build GRCh38
rs1128059 Chr.1: 11750617-11750617 on Build GRCh38
rs8617 Chr.1: 11750448-11750448 on Build GRCh38
rs529604721 Chr.1: 11750423-11750423 on Build GRCh38
rs1020774299 Chr.1: 11750462-11750462 on Build GRCh38
rs939768086 Chr.1: 11750695-11750695 on Build GRCh38
rs781675344 Chr.1: 11750607-11750607 on Build GRCh38
rs183691853 Chr.1: 11750619-11750619 on Build GRCh38
rs1028562129 Chr.1: 11750609-11750609 on Build GRCh38
rs576416904 Chr.1: 11750604-11750604 on Build GRCh38
rs1054776173 Chr.1: 11750719-11750719 on Build GRCh38

View Full Product Details