Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12661549-12661948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AADACL4
Entrez Gene ID
343066
Gene Name
arylacetamide deacetylase like 4
Gene Aliases
-
UniGene
Hs.209954
Gene Chromosome Location
Chr.1: 12644547-12667086 on Build GRCh38
Transcripts
NM_001013630.1 XM_017001152.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6506930 c.399C>A p.G133G Chr.1: 12661804-12661804 on Build GRCh38
2180896 c.405C>A p.C135* Chr.1: 12661810-12661810 on Build GRCh38
3472494 c.424A>G p.T142A Chr.1: 12661829-12661829 on Build GRCh38
5673004 c.421G>T p.E141* Chr.1: 12661826-12661826 on Build GRCh38
6506931 c.400C>A p.L134M Chr.1: 12661805-12661805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534073202 Chr.1: 12661618-12661618 on Build GRCh38
rs780985429 Chr.1: 12661793-12661793 on Build GRCh38
rs528423855 Chr.1: 12661848-12661848 on Build GRCh38
rs748784919 Chr.1: 12661817-12661817 on Build GRCh38
rs764108264 Chr.1: 12661887-12661887 on Build GRCh38
rs751740979 Chr.1: 12661891-12661891 on Build GRCh38
rs778741834 Chr.1: 12661820-12661820 on Build GRCh38
rs773761962 Chr.1: 12661751-12661751 on Build GRCh38
rs750222347 Chr.1: 12661898-12661898 on Build GRCh38
rs1028051231 Chr.1: 12661648-12661648 on Build GRCh38
rs959488322 Chr.1: 12661596-12661596 on Build GRCh38
rs185024112 Chr.1: 12661823-12661823 on Build GRCh38
rs4846132 Chr.1: 12661704-12661704 on Build GRCh38
rs1019331946 Chr.1: 12661677-12661677 on Build GRCh38
rs751760899 Chr.1: 12661777-12661777 on Build GRCh38
rs973040483 Chr.1: 12661683-12661683 on Build GRCh38
rs923886396 Chr.1: 12661676-12661676 on Build GRCh38
rs1053064143 Chr.1: 12661863-12661863 on Build GRCh38
rs113519885 Chr.1: 12661639-12661639 on Build GRCh38
rs568228458 Chr.1: 12661579-12661579 on Build GRCh38
rs961924553 Chr.1: 12661681-12661681 on Build GRCh38
rs760917734 Chr.1: 12661760-12661760 on Build GRCh38
rs766728790 Chr.1: 12661761-12661761 on Build GRCh38
rs777027803 Chr.1: 12661762-12661762 on Build GRCh38
rs1046388280 Chr.1: 12661598-12661598 on Build GRCh38
rs949434321 Chr.1: 12661626-12661626 on Build GRCh38
rs368222648 Chr.1: 12661893-12661893 on Build GRCh38
rs776776235 Chr.1: 12661601-12661601 on Build GRCh38
rs760020522 Chr.1: 12661764-12661764 on Build GRCh38
rs768413361 Chr.1: 12661742-12661742 on Build GRCh38
rs770263571 Chr.1: 12661847-12661847 on Build GRCh38
rs1053585592 Chr.1: 12661737-12661737 on Build GRCh38
rs779773583 Chr.1: 12661814-12661814 on Build GRCh38
rs764266102 Chr.1: 12661776-12661776 on Build GRCh38
rs1027206068 Chr.1: 12661895-12661895 on Build GRCh38
rs559483525 Chr.1: 12661824-12661824 on Build GRCh38
rs890187095 Chr.1: 12661850-12661850 on Build GRCh38
rs769475698 Chr.1: 12661646-12661646 on Build GRCh38
rs765366526 Chr.1: 12661767-12661767 on Build GRCh38
rs554156479 Chr.1: 12661687-12661687 on Build GRCh38
rs747789410 Chr.1: 12661821-12661821 on Build GRCh38
rs936596759 Chr.1: 12661693-12661693 on Build GRCh38
rs889552000 Chr.1: 12661668-12661668 on Build GRCh38
rs118115612 Chr.1: 12661728-12661728 on Build GRCh38
rs554194165 Chr.1: 12661635-12661635 on Build GRCh38
rs923998639 Chr.1: 12661838-12661838 on Build GRCh38
rs760019354 Chr.1: 12661845-12661845 on Build GRCh38
rs768464403 Chr.1: 12661819-12661819 on Build GRCh38
rs747814222 Chr.1: 12661692-12661692 on Build GRCh38
rs1014578876 Chr.1: 12661872-12661872 on Build GRCh38
rs201004446 Chr.1: 12661774-12661774 on Build GRCh38
rs576414593 Chr.1: 12661656-12661656 on Build GRCh38
rs375470521 Chr.1: 12661810-12661810 on Build GRCh38
rs763049052 Chr.1: 12661871-12661871 on Build GRCh38
rs892997009 Chr.1: 12661623-12661623 on Build GRCh38
rs757023395 Chr.1: 12661788-12661788 on Build GRCh38
rs202203165 Chr.1: 12661859-12661859 on Build GRCh38
rs146724349 Chr.1: 12661608-12661608 on Build GRCh38
rs749019836 Chr.1: 12661741-12661741 on Build GRCh38
rs745800653 Chr.1: 12661807-12661807 on Build GRCh38
rs995762358 Chr.1: 12661894-12661894 on Build GRCh38
rs752760333 Chr.1: 12661770-12661770 on Build GRCh38

View Full Product Details