Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15114848-15115345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAZN
Entrez Gene ID
23254
Gene Name
kazrin, periplakin interacting protein
Gene Aliases
KAZ
UniGene
Hs.368823
Gene Chromosome Location
Chr.1: 13893387-15118048 on Build GRCh38
Transcripts
NM_201628.2 XM_005245795.4 XM_011541074.2 XM_011541075.2 XM_011541076.2 XM_011541077.2 XM_011541081.2 NR_027136.1

Gene Symbol
TMEM51-AS1
Entrez Gene ID
200197
Gene Name
TMEM51 antisense RNA 1
Gene Aliases
C1orf126
UniGene
Hs.531424
Gene Chromosome Location
Chr.1: 15111815-15152464 on Build GRCh38
Transcripts
NM_201628.2 XM_005245795.4 XM_011541074.2 XM_011541075.2 XM_011541076.2 XM_011541077.2 XM_011541081.2 NR_027136.1

SNP Detail

SNP ID Chromosome Location
rs540535635 Chr.1: 15114897-15114897 on Build GRCh38
rs943653787 Chr.1: 15115218-15115218 on Build GRCh38
rs535093819 Chr.1: 15115234-15115234 on Build GRCh38
rs933711151 Chr.1: 15115032-15115032 on Build GRCh38
rs902021361 Chr.1: 15115224-15115224 on Build GRCh38
rs889243497 Chr.1: 15114979-15114979 on Build GRCh38
rs533094140 Chr.1: 15115015-15115015 on Build GRCh38
rs1052339180 Chr.1: 15115285-15115285 on Build GRCh38
rs962414557 Chr.1: 15114987-15114987 on Build GRCh38
rs549662548 Chr.1: 15115265-15115265 on Build GRCh38
rs376648854 Chr.1: 15115112-15115112 on Build GRCh38
rs889359068 Chr.1: 15115209-15115209 on Build GRCh38
rs939429294 Chr.1: 15115117-15115117 on Build GRCh38
rs186129323 Chr.1: 15115321-15115321 on Build GRCh38
rs1023779540 Chr.1: 15114887-15114888 on Build GRCh38
rs951809693 Chr.1: 15115029-15115029 on Build GRCh38
rs764671867 Chr.1: 15115148-15115148 on Build GRCh38
rs1043966643 Chr.1: 15114968-15114968 on Build GRCh38
rs913871391 Chr.1: 15115147-15115147 on Build GRCh38
rs1006767354 Chr.1: 15115130-15115130 on Build GRCh38
rs984666760 Chr.1: 15115044-15115044 on Build GRCh38
rs1025949677 Chr.1: 15115028-15115028 on Build GRCh38
rs905393911 Chr.1: 15115127-15115127 on Build GRCh38
rs985099210 Chr.1: 15114946-15114946 on Build GRCh38
rs910965693 Chr.1: 15115221-15115221 on Build GRCh38
rs993227401 Chr.1: 15115264-15115264 on Build GRCh38
rs1011086665 Chr.1: 15114940-15114940 on Build GRCh38
rs530726105 Chr.1: 15114932-15114932 on Build GRCh38
rs942527848 Chr.1: 15115065-15115065 on Build GRCh38
rs181890568 Chr.1: 15115305-15115305 on Build GRCh38
rs1022664229 Chr.1: 15115314-15115314 on Build GRCh38
rs988646306 Chr.1: 15115131-15115131 on Build GRCh38
rs979638315 Chr.1: 15115253-15115253 on Build GRCh38
rs1019565249 Chr.1: 15114994-15114994 on Build GRCh38
rs550764521 Chr.1: 15114980-15114980 on Build GRCh38
rs546356397 Chr.1: 15115053-15115053 on Build GRCh38
rs705581 Chr.1: 15114998-15114998 on Build GRCh38
rs955021461 Chr.1: 15115187-15115187 on Build GRCh38
rs1005880421 Chr.1: 15115306-15115306 on Build GRCh38
rs189297762 Chr.1: 15115273-15115273 on Build GRCh38
rs150840609 Chr.1: 15115032-15115033 on Build GRCh38
rs986554393 Chr.1: 15115213-15115213 on Build GRCh38
rs749995198 Chr.1: 15115153-15115153 on Build GRCh38
rs1043683497 Chr.1: 15115066-15115066 on Build GRCh38
rs936217362 Chr.1: 15115031-15115031 on Build GRCh38
rs538072008 Chr.1: 15115293-15115293 on Build GRCh38
rs561791516 Chr.1: 15114922-15114922 on Build GRCh38
rs1016517768 Chr.1: 15115134-15115134 on Build GRCh38
rs757868340 Chr.1: 15115182-15115182 on Build GRCh38
rs1052412637 Chr.1: 15115092-15115092 on Build GRCh38
rs866619223 Chr.1: 15115129-15115129 on Build GRCh38
rs1040522595 Chr.1: 15115223-15115223 on Build GRCh38
rs981258003 Chr.1: 15115094-15115094 on Build GRCh38
rs935350308 Chr.1: 15115085-15115085 on Build GRCh38
rs1025291437 Chr.1: 15115181-15115181 on Build GRCh38
rs969995049 Chr.1: 15115076-15115076 on Build GRCh38

View Full Product Details