Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15117505-15118019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAZN
Entrez Gene ID
23254
Gene Name
kazrin, periplakin interacting protein
Gene Aliases
KAZ
UniGene
Hs.368823
Gene Chromosome Location
Chr.1: 13893387-15118048 on Build GRCh38
Transcripts
NM_201628.2 XM_005245795.4 XM_011541074.2 XM_011541075.2 XM_011541076.2 XM_011541077.2 XM_011541081.2 NR_027136.1

Gene Symbol
TMEM51-AS1
Entrez Gene ID
200197
Gene Name
TMEM51 antisense RNA 1
Gene Aliases
C1orf126
UniGene
Hs.531424
Gene Chromosome Location
Chr.1: 15111815-15152464 on Build GRCh38
Transcripts
NM_201628.2 XM_005245795.4 XM_011541074.2 XM_011541075.2 XM_011541076.2 XM_011541077.2 XM_011541081.2 NR_027136.1

SNP Detail

SNP ID Chromosome Location
rs142383624 Chr.1: 15117827-15117827 on Build GRCh38
rs1029303816 Chr.1: 15117551-15117551 on Build GRCh38
rs997018924 Chr.1: 15117544-15117544 on Build GRCh38
rs550368893 Chr.1: 15117535-15117535 on Build GRCh38
rs112092999 Chr.1: 15117601-15117601 on Build GRCh38
rs1054529919 Chr.1: 15117765-15117765 on Build GRCh38
rs890919554 Chr.1: 15117793-15117793 on Build GRCh38
rs936229519 Chr.1: 15117938-15117938 on Build GRCh38
rs528702812 Chr.1: 15117548-15117548 on Build GRCh38
rs372145543 Chr.1: 15117658-15117659 on Build GRCh38
rs565467188 Chr.1: 15117785-15117785 on Build GRCh38
rs962646919 Chr.1: 15117586-15117586 on Build GRCh38
rs538390228 Chr.1: 15117838-15117838 on Build GRCh38
rs138789670 Chr.1: 15117948-15117948 on Build GRCh38
rs547386675 Chr.1: 15117716-15117716 on Build GRCh38
rs907248295 Chr.1: 15117809-15117809 on Build GRCh38
rs893131073 Chr.1: 15117560-15117560 on Build GRCh38
rs1027300467 Chr.1: 15117679-15117679 on Build GRCh38
rs951711675 Chr.1: 15117684-15117684 on Build GRCh38
rs990491944 Chr.1: 15117602-15117602 on Build GRCh38
rs921405575 Chr.1: 15117977-15117977 on Build GRCh38
rs950192081 Chr.1: 15117898-15117898 on Build GRCh38
rs575152081 Chr.1: 15117987-15117987 on Build GRCh38
rs937412907 Chr.1: 15117635-15117635 on Build GRCh38
rs1003867486 Chr.1: 15117808-15117808 on Build GRCh38
rs532688629 Chr.1: 15117565-15117565 on Build GRCh38
rs2235784 Chr.1: 15117792-15117792 on Build GRCh38
rs34041832 Chr.1: 15117664-15117665 on Build GRCh38
rs758642717 Chr.1: 15117558-15117558 on Build GRCh38
rs992043435 Chr.1: 15117879-15117879 on Build GRCh38
rs569237397 Chr.1: 15117885-15117885 on Build GRCh38
rs192693676 Chr.1: 15117566-15117566 on Build GRCh38
rs1054362597 Chr.1: 15117772-15117772 on Build GRCh38
rs755003691 Chr.1: 15117724-15117724 on Build GRCh38
rs540974031 Chr.1: 15117945-15117945 on Build GRCh38
rs977375551 Chr.1: 15117913-15117913 on Build GRCh38
rs111508029 Chr.1: 15117752-15117752 on Build GRCh38
rs982681987 Chr.1: 15117559-15117559 on Build GRCh38
rs2235783 Chr.1: 15117991-15117991 on Build GRCh38
rs900014418 Chr.1: 15117779-15117779 on Build GRCh38
rs72873846 Chr.1: 15117735-15117735 on Build GRCh38
rs1033478683 Chr.1: 15117863-15117863 on Build GRCh38
rs959189081 Chr.1: 15117639-15117639 on Build GRCh38
rs558610735 Chr.1: 15117849-15117849 on Build GRCh38
rs907405264 Chr.1: 15117620-15117620 on Build GRCh38
rs137939353 Chr.1: 15117655-15117656 on Build GRCh38
rs747122673 Chr.1: 15117711-15117711 on Build GRCh38
rs893183331 Chr.1: 15117777-15117777 on Build GRCh38
rs146596693 Chr.1: 15117866-15117866 on Build GRCh38
rs894623526 Chr.1: 15117854-15117854 on Build GRCh38
rs563789414 Chr.1: 15117543-15117543 on Build GRCh38
rs759153824 Chr.1: 15117941-15117941 on Build GRCh38
rs766878851 Chr.1: 15117990-15117990 on Build GRCh38
rs183972203 Chr.1: 15117652-15117652 on Build GRCh38
rs903434625 Chr.1: 15117751-15117751 on Build GRCh38
rs924633660 Chr.1: 15117935-15117935 on Build GRCh38

View Full Product Details