Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30462381-30462884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGB1
Entrez Gene ID
3146
Gene Name
high mobility group box 1
Gene Aliases
HMG-1, HMG1, HMG3, SBP-1
UniGene
Hs.434102
Gene Chromosome Location
Chr.13: 30457916-30617597 on Build GRCh38
Transcripts
NM_001313892.1 NM_001313893.1 NM_002128.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7185437 c.470A>T p.K157M Chr.13: 30462539-30462539 on Build GRCh38
281728 c.443C>G p.A148G Chr.13: 30462566-30462566 on Build GRCh38
7158375 c.332C>T p.P111L Chr.13: 30462677-30462677 on Build GRCh38
261304 c.378G>A p.A126A Chr.13: 30462631-30462631 on Build GRCh38
3688671 c.447G>A p.A149A Chr.13: 30462562-30462562 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760333795 Chr.13: 30462709-30462709 on Build GRCh38
rs568608708 Chr.13: 30462843-30462843 on Build GRCh38
rs768258079 Chr.13: 30462715-30462715 on Build GRCh38
rs754370309 Chr.13: 30462538-30462538 on Build GRCh38
rs529452831 Chr.13: 30462725-30462725 on Build GRCh38
rs529404831 Chr.13: 30462765-30462765 on Build GRCh38
rs143984318 Chr.13: 30462661-30462661 on Build GRCh38
rs539199302 Chr.13: 30462855-30462855 on Build GRCh38
rs768430647 Chr.13: 30462598-30462598 on Build GRCh38
rs756193888 Chr.13: 30462748-30462748 on Build GRCh38
rs775262922 Chr.13: 30462433-30462433 on Build GRCh38
rs767316971 Chr.13: 30462516-30462516 on Build GRCh38
rs3742305 Chr.13: 30462505-30462505 on Build GRCh38
rs749911025 Chr.13: 30462463-30462463 on Build GRCh38
rs4483702 Chr.13: 30462413-30462413 on Build GRCh38
rs899505127 Chr.13: 30462651-30462651 on Build GRCh38
rs754286912 Chr.13: 30462467-30462467 on Build GRCh38
rs202081642 Chr.13: 30462691-30462691 on Build GRCh38
rs200955676 Chr.13: 30462577-30462577 on Build GRCh38
rs1022518793 Chr.13: 30462622-30462622 on Build GRCh38
rs751292341 Chr.13: 30462454-30462454 on Build GRCh38
rs780035673 Chr.13: 30462452-30462452 on Build GRCh38
rs540146818 Chr.13: 30462752-30462752 on Build GRCh38
rs755433870 Chr.13: 30462471-30462471 on Build GRCh38
rs1050976635 Chr.13: 30462828-30462828 on Build GRCh38
rs979398038 Chr.13: 30462848-30462848 on Build GRCh38
rs1017938457 Chr.13: 30462414-30462414 on Build GRCh38
rs570121784 Chr.13: 30462823-30462823 on Build GRCh38
rs779076230 Chr.13: 30462483-30462483 on Build GRCh38
rs749178584 Chr.13: 30462500-30462500 on Build GRCh38
rs1929606 Chr.13: 30462544-30462544 on Build GRCh38
rs1026867229 Chr.13: 30462438-30462438 on Build GRCh38
rs745973116 Chr.13: 30462747-30462747 on Build GRCh38
rs767149749 Chr.13: 30462462-30462462 on Build GRCh38
rs990900975 Chr.13: 30462720-30462720 on Build GRCh38
rs959837793 Chr.13: 30462757-30462757 on Build GRCh38
rs761493027 Chr.13: 30462455-30462455 on Build GRCh38
rs747688524 Chr.13: 30462725-30462725 on Build GRCh38
rs916577339 Chr.13: 30462419-30462419 on Build GRCh38
rs768503924 Chr.13: 30462503-30462503 on Build GRCh38
rs769681654 Chr.13: 30462431-30462431 on Build GRCh38
rs780660128 Chr.13: 30462465-30462465 on Build GRCh38
rs771979933 Chr.13: 30462697-30462697 on Build GRCh38
rs772976540 Chr.13: 30462519-30462519 on Build GRCh38
rs149451974 Chr.13: 30462597-30462597 on Build GRCh38
rs918470233 Chr.13: 30462770-30462770 on Build GRCh38
rs4540927 Chr.13: 30462407-30462407 on Build GRCh38
rs4384465 Chr.13: 30462734-30462734 on Build GRCh38
rs377149884 Chr.13: 30462727-30462727 on Build GRCh38
rs141813238 Chr.13: 30462489-30462489 on Build GRCh38
rs1039414517 Chr.13: 30462845-30462845 on Build GRCh38
rs772229919 Chr.13: 30462487-30462487 on Build GRCh38
rs368122446 Chr.13: 30462547-30462547 on Build GRCh38
rs17074619 Chr.13: 30462420-30462420 on Build GRCh38
rs764441758 Chr.13: 30462726-30462726 on Build GRCh38
rs781066692 Chr.13: 30462589-30462589 on Build GRCh38
rs765706189 Chr.13: 30462717-30462717 on Build GRCh38
rs770980857 Chr.13: 30462497-30462497 on Build GRCh38
rs762859464 Chr.13: 30462442-30462442 on Build GRCh38
rs1052595901 Chr.13: 30462541-30462541 on Build GRCh38
rs200788197 Chr.13: 30462492-30462492 on Build GRCh38
rs775952971 Chr.13: 30462710-30462710 on Build GRCh38
rs61733675 Chr.13: 30462701-30462701 on Build GRCh38
rs756653646 Chr.13: 30462503-30462505 on Build GRCh38
rs760319277 Chr.13: 30462522-30462522 on Build GRCh38
rs761587427 Chr.13: 30462513-30462513 on Build GRCh38
rs763784125 Chr.13: 30462449-30462449 on Build GRCh38
rs1060348 Chr.13: 30462700-30462700 on Build GRCh38
rs757425784 Chr.13: 30462580-30462580 on Build GRCh38
rs4597168 Chr.13: 30462581-30462581 on Build GRCh38
rs890949959 Chr.13: 30462838-30462838 on Build GRCh38
rs550763562 Chr.13: 30462804-30462804 on Build GRCh38
rs369858592 Chr.13: 30462737-30462737 on Build GRCh38
rs748413203 Chr.13: 30462484-30462484 on Build GRCh38
rs137943992 Chr.13: 30462571-30462571 on Build GRCh38
rs112327776 Chr.13: 30462405-30462405 on Build GRCh38
rs753022161 Chr.13: 30462562-30462562 on Build GRCh38
rs747133847 Chr.13: 30462496-30462496 on Build GRCh38
rs765798136 Chr.13: 30462531-30462531 on Build GRCh38
rs758734227 Chr.13: 30462563-30462563 on Build GRCh38
rs753183343 Chr.13: 30462722-30462722 on Build GRCh38
rs769500373 Chr.13: 30462739-30462743 on Build GRCh38
rs777649504 Chr.13: 30462762-30462762 on Build GRCh38
rs892814911 Chr.13: 30462858-30462858 on Build GRCh38
rs774313138 Chr.13: 30462688-30462688 on Build GRCh38
rs551442918 Chr.13: 30462714-30462715 on Build GRCh38
rs780796190 Chr.13: 30462723-30462724 on Build GRCh38
rs925547616 Chr.13: 30462852-30462852 on Build GRCh38
rs760116781 Chr.13: 30462714-30462714 on Build GRCh38
rs771994119 Chr.13: 30462761-30462761 on Build GRCh38
rs757365859 Chr.13: 30462729-30462729 on Build GRCh38
rs766834979 Chr.13: 30462461-30462461 on Build GRCh38
rs775556193 Chr.13: 30462499-30462499 on Build GRCh38
rs951300005 Chr.13: 30462440-30462440 on Build GRCh38
rs527620039 Chr.13: 30462451-30462451 on Build GRCh38
rs192331953 Chr.13: 30462786-30462786 on Build GRCh38

View Full Product Details