Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 31140042-31140542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPH1
Entrez Gene ID
10808
Gene Name
heat shock protein family H (Hsp110) member 1
Gene Aliases
HSP105, HSP105A, HSP105B, NY-CO-25
UniGene
Hs.743267
Gene Chromosome Location
Chr.13: 31134974-31163109 on Build GRCh38
Transcripts
NM_001286503.1 NM_001286504.1 NM_001286505.1 NM_006644.3 XM_005266236.1 XM_011534887.2 XM_011534888.1 XM_017020361.1 XM_017020362.1 XM_017020363.1 XM_017020364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7301817 c.1882G>A p.E628K Chr.13: 31140282-31140282 on Build GRCh38
1728407 c.1855-1G>T p.? Chr.13: 31140310-31140310 on Build GRCh38
7224855 c.1885T>C p.L629L Chr.13: 31140285-31140285 on Build GRCh38
6325454 c.1947G>A p.L649L Chr.13: 31140217-31140217 on Build GRCh38
7206128 c.1979G>A p.C660Y Chr.13: 31140191-31140191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144947618 Chr.13: 31140232-31140232 on Build GRCh38
rs909530520 Chr.13: 31140500-31140500 on Build GRCh38
rs955995091 Chr.13: 31140091-31140091 on Build GRCh38
rs778474328 Chr.13: 31140123-31140123 on Build GRCh38
rs140483871 Chr.13: 31140285-31140285 on Build GRCh38
rs907570943 Chr.13: 31140472-31140472 on Build GRCh38
rs186473539 Chr.13: 31140191-31140191 on Build GRCh38
rs768452037 Chr.13: 31140147-31140148 on Build GRCh38
rs920297125 Chr.13: 31140173-31140173 on Build GRCh38
rs761289195 Chr.13: 31140271-31140271 on Build GRCh38
rs763525052 Chr.13: 31140147-31140147 on Build GRCh38
rs773783686 Chr.13: 31140134-31140134 on Build GRCh38
rs757280330 Chr.13: 31140197-31140197 on Build GRCh38
rs777066821 Chr.13: 31140166-31140167 on Build GRCh38
rs752822342 Chr.13: 31140195-31140195 on Build GRCh38
rs772677801 Chr.13: 31140272-31140272 on Build GRCh38
rs4450270 Chr.13: 31140100-31140100 on Build GRCh38
rs1029467589 Chr.13: 31140296-31140296 on Build GRCh38
rs750850284 Chr.13: 31140155-31140155 on Build GRCh38
rs754136189 Chr.13: 31140175-31140175 on Build GRCh38
rs768292196 Chr.13: 31140253-31140253 on Build GRCh38
rs773908278 Chr.13: 31140263-31140263 on Build GRCh38
rs779835850 Chr.13: 31140238-31140238 on Build GRCh38
rs756013064 Chr.13: 31140358-31140358 on Build GRCh38
rs750337368 Chr.13: 31140345-31140345 on Build GRCh38
rs752877054 Chr.13: 31140335-31140335 on Build GRCh38
rs755198084 Chr.13: 31140186-31140186 on Build GRCh38
rs776954995 Chr.13: 31140339-31140339 on Build GRCh38
rs543489338 Chr.13: 31140126-31140126 on Build GRCh38
rs566365040 Chr.13: 31140328-31140328 on Build GRCh38
rs1047157381 Chr.13: 31140145-31140145 on Build GRCh38
rs982590400 Chr.13: 31140498-31140498 on Build GRCh38
rs780072479 Chr.13: 31140143-31140144 on Build GRCh38
rs531724533 Chr.13: 31140094-31140094 on Build GRCh38
rs974945651 Chr.13: 31140431-31140432 on Build GRCh38
rs762093862 Chr.13: 31140316-31140316 on Build GRCh38
rs562191685 Chr.13: 31140188-31140188 on Build GRCh38
rs888142389 Chr.13: 31140206-31140206 on Build GRCh38
rs780958272 Chr.13: 31140203-31140203 on Build GRCh38
rs951187431 Chr.13: 31140453-31140453 on Build GRCh38
rs9563762 Chr.13: 31140452-31140452 on Build GRCh38
rs767895169 Chr.13: 31140151-31140151 on Build GRCh38
rs754193474 Chr.13: 31140321-31140321 on Build GRCh38
rs570448614 Chr.13: 31140425-31140425 on Build GRCh38
rs192106781 Chr.13: 31140367-31140367 on Build GRCh38
rs762373098 Chr.13: 31140137-31140137 on Build GRCh38
rs139144467 Chr.13: 31140242-31140242 on Build GRCh38
rs34849590 Chr.13: 31140074-31140074 on Build GRCh38
rs765756707 Chr.13: 31140362-31140366 on Build GRCh38
rs745774631 Chr.13: 31140207-31140207 on Build GRCh38
rs200665817 Chr.13: 31140159-31140159 on Build GRCh38
rs373644441 Chr.13: 31140304-31140304 on Build GRCh38
rs987361349 Chr.13: 31140389-31140389 on Build GRCh38
rs1047497313 Chr.13: 31140466-31140466 on Build GRCh38
rs200118537 Chr.13: 31140337-31140337 on Build GRCh38
rs919455609 Chr.13: 31140442-31140442 on Build GRCh38
rs765420089 Chr.13: 31140370-31140370 on Build GRCh38
rs766726879 Chr.13: 31140162-31140162 on Build GRCh38
rs7491727 Chr.13: 31140401-31140401 on Build GRCh38
rs766667127 Chr.13: 31140317-31140317 on Build GRCh38
rs911772441 Chr.13: 31140399-31140399 on Build GRCh38
rs750515174 Chr.13: 31140433-31140433 on Build GRCh38
rs930331585 Chr.13: 31140180-31140180 on Build GRCh38
rs971659600 Chr.13: 31140404-31140404 on Build GRCh38

View Full Product Details