Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 32318765-32319289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRCA2
Entrez Gene ID
675
Gene Name
BRCA2, DNA repair associated
Gene Aliases
BRCC2, BROVCA2, FACD, FAD, FAD1, FANCD, FANCD1, GLM3, PNCA2, XRCC11
UniGene
Hs.34012
Gene Chromosome Location
Chr.13: 32315480-32399672 on Build GRCh38
Transcripts
NM_000059.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6986816 c.266C>T p.P89L Chr.13: 32319275-32319275 on Build GRCh38
327779 c.205C>T p.P69S Chr.13: 32319214-32319214 on Build GRCh38
6890974 c.94T>C p.F32L Chr.13: 32319103-32319103 on Build GRCh38
23939 c.75_76insCCAATGA p.I27fs*6 Chr.13: 32319084-32319085 on Build GRCh38
327778 c.205C>T p.P69S Chr.13: 32319214-32319214 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs397507909 Chr.13: 32319081-32319081 on Build GRCh38
rs80358961 Chr.13: 32319082-32319082 on Build GRCh38
rs397507603 Chr.13: 32319181-32319181 on Build GRCh38
rs397507376 Chr.13: 32319070-32319071 on Build GRCh38
rs397507573 Chr.13: 32319124-32319124 on Build GRCh38
rs80358450 Chr.13: 32319173-32319173 on Build GRCh38
rs276174818 Chr.13: 32319224-32319224 on Build GRCh38
rs80359722 Chr.13: 32319095-32319096 on Build GRCh38
rs876658614 Chr.13: 32319214-32319214 on Build GRCh38
rs864622464 Chr.13: 32319091-32319091 on Build GRCh38
rs431825283 Chr.13: 32319155-32319155 on Build GRCh38
rs80359320 Chr.13: 32319213-32319213 on Build GRCh38
rs886040414 Chr.13: 32319233-32319234 on Build GRCh38
rs536936252 Chr.13: 32318914-32318914 on Build GRCh38
rs114446594 Chr.13: 32319240-32319240 on Build GRCh38
rs876660504 Chr.13: 32319185-32319185 on Build GRCh38
rs779367922 Chr.13: 32319174-32319174 on Build GRCh38
rs80359182 Chr.13: 32319100-32319100 on Build GRCh38
rs80359214 Chr.13: 32319102-32319102 on Build GRCh38
rs886040432 Chr.13: 32319259-32319259 on Build GRCh38
rs4987046 Chr.13: 32319134-32319134 on Build GRCh38
rs80358507 Chr.13: 32319250-32319250 on Build GRCh38
rs80359287 Chr.13: 32319160-32319160 on Build GRCh38
rs786201783 Chr.13: 32319132-32319132 on Build GRCh38
rs951413972 Chr.13: 32318900-32318900 on Build GRCh38
rs397507903 Chr.13: 32319080-32319080 on Build GRCh38
rs886040429 Chr.13: 32319257-32319258 on Build GRCh38
rs397507595 Chr.13: 32319167-32319167 on Build GRCh38
rs80359262 Chr.13: 32319115-32319116 on Build GRCh38
rs886040340 Chr.13: 32319112-32319113 on Build GRCh38
rs80358505 Chr.13: 32319249-32319249 on Build GRCh38
rs886040406 Chr.13: 32319221-32319222 on Build GRCh38
rs548945825 Chr.13: 32318924-32318924 on Build GRCh38
rs760511134 Chr.13: 32318885-32318885 on Build GRCh38
rs749827015 Chr.13: 32319164-32319164 on Build GRCh38
rs397507615 Chr.13: 32319200-32319200 on Build GRCh38
rs1012688095 Chr.13: 32319210-32319210 on Build GRCh38
rs886040341 Chr.13: 32319113-32319119 on Build GRCh38
rs886038048 Chr.13: 32319213-32319214 on Build GRCh38
rs397507628 Chr.13: 32319253-32319253 on Build GRCh38
rs770475551 Chr.13: 32319057-32319057 on Build GRCh38
rs786201716 Chr.13: 32319131-32319131 on Build GRCh38
rs776638534 Chr.13: 32319192-32319192 on Build GRCh38
rs80358500 Chr.13: 32319238-32319238 on Build GRCh38
rs566846890 Chr.13: 32318955-32318955 on Build GRCh38
rs730881554 Chr.13: 32319116-32319116 on Build GRCh38
rs397508045 Chr.13: 32319101-32319101 on Build GRCh38
rs756971498 Chr.13: 32319128-32319128 on Build GRCh38
rs483353112 Chr.13: 32319141-32319142 on Build GRCh38
rs779648876 Chr.13: 32319156-32319156 on Build GRCh38
rs28897701 Chr.13: 32319232-32319232 on Build GRCh38
rs398122745 Chr.13: 32319241-32319241 on Build GRCh38
rs80358425 Chr.13: 32319145-32319145 on Build GRCh38
rs971747455 Chr.13: 32318837-32318837 on Build GRCh38
rs769152395 Chr.13: 32319075-32319075 on Build GRCh38
rs1022837001 Chr.13: 32318797-32318797 on Build GRCh38
rs397507902 Chr.13: 32319080-32319080 on Build GRCh38
rs1012738609 Chr.13: 32318791-32318791 on Build GRCh38
rs905964686 Chr.13: 32318852-32318852 on Build GRCh38
rs397507617 Chr.13: 32319205-32319205 on Build GRCh38
rs80358502 Chr.13: 32319244-32319244 on Build GRCh38
rs876660626 Chr.13: 32319247-32319247 on Build GRCh38
rs80358498 Chr.13: 32319236-32319236 on Build GRCh38
rs777887676 Chr.13: 32318840-32318840 on Build GRCh38
rs746683798 Chr.13: 32319028-32319028 on Build GRCh38
rs80358435 Chr.13: 32319154-32319154 on Build GRCh38
rs866071378 Chr.13: 32319157-32319157 on Build GRCh38
rs533970381 Chr.13: 32318968-32318968 on Build GRCh38
rs80359275 Chr.13: 32319137-32319137 on Build GRCh38
rs81002830 Chr.13: 32319070-32319070 on Build GRCh38
rs397507581 Chr.13: 32319142-32319142 on Build GRCh38
rs572782576 Chr.13: 32319266-32319266 on Build GRCh38
rs751417001 Chr.13: 32319219-32319219 on Build GRCh38
rs924234502 Chr.13: 32319183-32319183 on Build GRCh38
rs1032184694 Chr.13: 32318915-32318915 on Build GRCh38
rs876661275 Chr.13: 32319118-32319118 on Build GRCh38
rs1035406554 Chr.13: 32318879-32318879 on Build GRCh38
rs11571587 Chr.13: 32319174-32319176 on Build GRCh38
rs921513445 Chr.13: 32319168-32319168 on Build GRCh38
rs80359299 Chr.13: 32319179-32319180 on Build GRCh38
rs276174878 Chr.13: 32319070-32319070 on Build GRCh38
rs80358480 Chr.13: 32319198-32319198 on Build GRCh38
rs772146565 Chr.13: 32319087-32319087 on Build GRCh38
rs80358515 Chr.13: 32319259-32319259 on Build GRCh38
rs1021312661 Chr.13: 32318931-32318931 on Build GRCh38
rs397507630 Chr.13: 32319255-32319256 on Build GRCh38
rs886040346 Chr.13: 32319119-32319119 on Build GRCh38
rs745336881 Chr.13: 32319055-32319055 on Build GRCh38
rs397508057 Chr.13: 32319103-32319103 on Build GRCh38
rs756807012 Chr.13: 32319035-32319035 on Build GRCh38
rs80358463 Chr.13: 32319188-32319188 on Build GRCh38
rs896067660 Chr.13: 32318834-32318834 on Build GRCh38
rs760655471 Chr.13: 32319099-32319099 on Build GRCh38
rs886040428 Chr.13: 32319256-32319256 on Build GRCh38
rs762819719 Chr.13: 32319111-32319111 on Build GRCh38
rs80358391 Chr.13: 32319109-32319109 on Build GRCh38
rs397507619 Chr.13: 32319224-32319224 on Build GRCh38
rs80359034 Chr.13: 32319088-32319088 on Build GRCh38
rs886038047 Chr.13: 32319129-32319132 on Build GRCh38
rs1037725365 Chr.13: 32318814-32318814 on Build GRCh38
rs878853297 Chr.13: 32319169-32319170 on Build GRCh38
rs886040353 Chr.13: 32319135-32319135 on Build GRCh38
rs28897700 Chr.13: 32319207-32319207 on Build GRCh38
rs863224303 Chr.13: 32319231-32319231 on Build GRCh38
rs886037796 Chr.13: 32319170-32319170 on Build GRCh38
rs771011731 Chr.13: 32319190-32319190 on Build GRCh38
rs587778128 Chr.13: 32319202-32319202 on Build GRCh38
rs56091799 Chr.13: 32319184-32319184 on Build GRCh38
rs752172011 Chr.13: 32319060-32319060 on Build GRCh38
rs201523522 Chr.13: 32319180-32319180 on Build GRCh38
rs1009399268 Chr.13: 32318916-32318916 on Build GRCh38
rs886040354 Chr.13: 32319136-32319137 on Build GRCh38
rs864622434 Chr.13: 32319153-32319153 on Build GRCh38
rs80358445 Chr.13: 32319170-32319170 on Build GRCh38
rs786201770 Chr.13: 32319074-32319074 on Build GRCh38
rs80358415 Chr.13: 32319130-32319130 on Build GRCh38
rs376931156 Chr.13: 32319218-32319218 on Build GRCh38
rs565480894 Chr.13: 32319026-32319026 on Build GRCh38
rs1044419895 Chr.13: 32319222-32319222 on Build GRCh38
rs397507882 Chr.13: 32319061-32319061 on Build GRCh38
rs781354453 Chr.13: 32319060-32319061 on Build GRCh38
rs886040344 Chr.13: 32319118-32319121 on Build GRCh38
rs747261314 Chr.13: 32319058-32319058 on Build GRCh38
rs397507883 Chr.13: 32319068-32319068 on Build GRCh38
rs80358454 Chr.13: 32319176-32319176 on Build GRCh38
rs1003162870 Chr.13: 32319239-32319239 on Build GRCh38
rs398122724 Chr.13: 32319125-32319125 on Build GRCh38
rs397507622 Chr.13: 32319235-32319235 on Build GRCh38
rs780782927 Chr.13: 32319044-32319044 on Build GRCh38
rs879255300 Chr.13: 32319090-32319092 on Build GRCh38
rs774521832 Chr.13: 32319077-32319077 on Build GRCh38
rs1002802766 Chr.13: 32318846-32318846 on Build GRCh38
rs755556921 Chr.13: 32319061-32319061 on Build GRCh38
rs398122741 Chr.13: 32319226-32319226 on Build GRCh38
rs886040400 Chr.13: 32319211-32319212 on Build GRCh38
rs397508065 Chr.13: 32319106-32319106 on Build GRCh38
rs876658407 Chr.13: 32319101-32319115 on Build GRCh38
rs398122733 Chr.13: 32319195-32319195 on Build GRCh38
rs569481738 Chr.13: 32318907-32318907 on Build GRCh38

View Full Product Details