Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 36431712-36432246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNA1
Entrez Gene ID
8900
Gene Name
cyclin A1
Gene Aliases
CT146
UniGene
Hs.417050
Gene Chromosome Location
Chr.13: 36430488-36442882 on Build GRCh38
Transcripts
NM_001111046.1 NM_001111047.1 XM_011535294.2 XM_011535295.2

SNP Detail

SNP ID Chromosome Location
rs897163392 Chr.13: 36431858-36431858 on Build GRCh38
rs982390512 Chr.13: 36432104-36432104 on Build GRCh38
rs1055483382 Chr.13: 36431905-36431905 on Build GRCh38
rs190970812 Chr.13: 36431782-36431782 on Build GRCh38
rs916828677 Chr.13: 36432097-36432097 on Build GRCh38
rs998920517 Chr.13: 36431991-36431991 on Build GRCh38
rs965715102 Chr.13: 36431980-36431980 on Build GRCh38
rs553662173 Chr.13: 36432121-36432121 on Build GRCh38
rs901014756 Chr.13: 36432206-36432206 on Build GRCh38
rs910800060 Chr.13: 36431773-36431773 on Build GRCh38
rs1033057048 Chr.13: 36432059-36432059 on Build GRCh38
rs976712596 Chr.13: 36432116-36432116 on Build GRCh38
rs183114570 Chr.13: 36431817-36431817 on Build GRCh38
rs1039977603 Chr.13: 36432194-36432194 on Build GRCh38
rs967034851 Chr.13: 36432042-36432042 on Build GRCh38
rs1024325933 Chr.13: 36431960-36431960 on Build GRCh38
rs958179583 Chr.13: 36432073-36432073 on Build GRCh38
rs139049981 Chr.13: 36431930-36431931 on Build GRCh38
rs888254349 Chr.13: 36431907-36431907 on Build GRCh38
rs1048497964 Chr.13: 36432149-36432149 on Build GRCh38
rs1032750925 Chr.13: 36432063-36432063 on Build GRCh38
rs1018173386 Chr.13: 36431973-36431973 on Build GRCh38
rs1048529355 Chr.13: 36431880-36431880 on Build GRCh38
rs926154320 Chr.13: 36431755-36431755 on Build GRCh38
rs978451273 Chr.13: 36432061-36432061 on Build GRCh38
rs984578616 Chr.13: 36432155-36432155 on Build GRCh38
rs925431032 Chr.13: 36431788-36431788 on Build GRCh38
rs557573220 Chr.13: 36432178-36432178 on Build GRCh38
rs986387477 Chr.13: 36431767-36431767 on Build GRCh38
rs550380815 Chr.13: 36431732-36431732 on Build GRCh38
rs942507582 Chr.13: 36432186-36432186 on Build GRCh38
rs1046863704 Chr.13: 36431841-36431841 on Build GRCh38
rs957570976 Chr.13: 36432066-36432066 on Build GRCh38
rs990985528 Chr.13: 36432096-36432096 on Build GRCh38
rs978483856 Chr.13: 36431787-36431787 on Build GRCh38
rs1006757971 Chr.13: 36431921-36431921 on Build GRCh38
rs908951480 Chr.13: 36432164-36432164 on Build GRCh38
rs569058337 Chr.13: 36431762-36431762 on Build GRCh38
rs936853168 Chr.13: 36431791-36431791 on Build GRCh38
rs994216856 Chr.13: 36431874-36431874 on Build GRCh38
rs929052110 Chr.13: 36431793-36431793 on Build GRCh38
rs571993725 Chr.13: 36432130-36432130 on Build GRCh38
rs187701340 Chr.13: 36431818-36431818 on Build GRCh38
rs997854232 Chr.13: 36432015-36432015 on Build GRCh38
rs1020331342 Chr.13: 36432024-36432024 on Build GRCh38
rs912977801 Chr.13: 36431778-36431778 on Build GRCh38
rs548306334 Chr.13: 36431806-36431806 on Build GRCh38
rs937710119 Chr.13: 36432173-36432173 on Build GRCh38
rs142757694 Chr.13: 36432175-36432175 on Build GRCh38
rs949632116 Chr.13: 36432098-36432098 on Build GRCh38
rs113328220 Chr.13: 36432069-36432069 on Build GRCh38

View Full Product Details