Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 36949631-36950166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG5
Entrez Gene ID
29880
Gene Name
ALG5, dolichyl-phosphate beta-glucosyltransferase
Gene Aliases
bA421P11.2
UniGene
Hs.507769
Gene Chromosome Location
Chr.13: 36949770-36999367 on Build GRCh38
Transcripts
NM_001142364.1 NM_013338.4 XM_017020552.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3468672 c.913T>A p.F305I Chr.13: 36950004-36950004 on Build GRCh38
696648 c.949C>T p.L317F Chr.13: 36949968-36949968 on Build GRCh38
141410 c.890T>A p.L297Q Chr.13: 36950027-36950027 on Build GRCh38
5376716 c.875C>T p.P292L Chr.13: 36950042-36950042 on Build GRCh38
175957 c.925C>T p.R309* Chr.13: 36949992-36949992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979194984 Chr.13: 36949772-36949772 on Build GRCh38
rs757791751 Chr.13: 36950087-36950087 on Build GRCh38
rs752193105 Chr.13: 36949955-36949955 on Build GRCh38
rs200601599 Chr.13: 36949915-36949915 on Build GRCh38
rs1007669432 Chr.13: 36949964-36949964 on Build GRCh38
rs758051668 Chr.13: 36949956-36949956 on Build GRCh38
rs939674956 Chr.13: 36949880-36949880 on Build GRCh38
rs760891409 Chr.13: 36949927-36949927 on Build GRCh38
rs937213374 Chr.13: 36950127-36950127 on Build GRCh38
rs139405331 Chr.13: 36949660-36949660 on Build GRCh38
rs145141807 Chr.13: 36949910-36949910 on Build GRCh38
rs927136346 Chr.13: 36950119-36950119 on Build GRCh38
rs564683661 Chr.13: 36949693-36949693 on Build GRCh38
rs746675683 Chr.13: 36949969-36949969 on Build GRCh38
rs763684684 Chr.13: 36950097-36950097 on Build GRCh38
rs753985955 Chr.13: 36949925-36949925 on Build GRCh38
rs118068142 Chr.13: 36949674-36949674 on Build GRCh38
rs766807133 Chr.13: 36950053-36950053 on Build GRCh38
rs189426750 Chr.13: 36949937-36949937 on Build GRCh38
rs112936182 Chr.13: 36949668-36949668 on Build GRCh38
rs1033497946 Chr.13: 36950140-36950140 on Build GRCh38
rs761864874 Chr.13: 36950029-36950029 on Build GRCh38
rs766857143 Chr.13: 36949936-36949936 on Build GRCh38
rs569952453 Chr.13: 36949879-36949879 on Build GRCh38
rs959186164 Chr.13: 36950141-36950141 on Build GRCh38
rs967946119 Chr.13: 36949768-36949768 on Build GRCh38
rs747100998 Chr.13: 36949719-36949722 on Build GRCh38
rs773248756 Chr.13: 36950046-36950046 on Build GRCh38
rs551637415 Chr.13: 36949802-36949802 on Build GRCh38
rs937267818 Chr.13: 36949755-36949755 on Build GRCh38
rs112451384 Chr.13: 36949805-36949805 on Build GRCh38
rs141152029 Chr.13: 36949948-36949948 on Build GRCh38
rs1001000911 Chr.13: 36949868-36949868 on Build GRCh38
rs780965497 Chr.13: 36950124-36950124 on Build GRCh38
rs945744579 Chr.13: 36949847-36949849 on Build GRCh38
rs372788214 Chr.13: 36950063-36950063 on Build GRCh38
rs1056775165 Chr.13: 36949788-36949788 on Build GRCh38
rs1019024110 Chr.13: 36950001-36950001 on Build GRCh38
rs917612059 Chr.13: 36949855-36949855 on Build GRCh38
rs201593403 Chr.13: 36949997-36949997 on Build GRCh38
rs937740741 Chr.13: 36949797-36949797 on Build GRCh38
rs146939104 Chr.13: 36950042-36950042 on Build GRCh38
rs1018932172 Chr.13: 36949655-36949655 on Build GRCh38
rs370016246 Chr.13: 36949991-36949991 on Build GRCh38
rs754291767 Chr.13: 36950062-36950062 on Build GRCh38
rs991903992 Chr.13: 36950142-36950142 on Build GRCh38
rs753256475 Chr.13: 36950085-36950085 on Build GRCh38
rs751135072 Chr.13: 36950101-36950101 on Build GRCh38
rs769762711 Chr.13: 36950007-36950007 on Build GRCh38
rs371970364 Chr.13: 36949885-36949885 on Build GRCh38
rs777229864 Chr.13: 36949967-36949967 on Build GRCh38
rs984364265 Chr.13: 36949731-36949731 on Build GRCh38
rs750786899 Chr.13: 36949924-36949924 on Build GRCh38
rs370578181 Chr.13: 36950125-36950125 on Build GRCh38
rs187354787 Chr.13: 36949735-36949735 on Build GRCh38
rs1041438808 Chr.13: 36949849-36949849 on Build GRCh38
rs745781616 Chr.13: 36949998-36949998 on Build GRCh38
rs918709701 Chr.13: 36949685-36949685 on Build GRCh38
rs770954445 Chr.13: 36949902-36949903 on Build GRCh38
rs751569349 Chr.13: 36950038-36950038 on Build GRCh38
rs553668958 Chr.13: 36949908-36949908 on Build GRCh38
rs553374135 Chr.13: 36950112-36950112 on Build GRCh38
rs3818380 Chr.13: 36950118-36950118 on Build GRCh38
rs774461644 Chr.13: 36949898-36949898 on Build GRCh38
rs765898624 Chr.13: 36950078-36950078 on Build GRCh38
rs774410648 Chr.13: 36950023-36950023 on Build GRCh38
rs926381106 Chr.13: 36949778-36949778 on Build GRCh38
rs778547845 Chr.13: 36949954-36949954 on Build GRCh38

View Full Product Details