Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 48709159-48709703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYSLTR2
Entrez Gene ID
57105
Gene Name
cysteinyl leukotriene receptor 2
Gene Aliases
CYSLT2, CYSLT2R, GPCR21, HG57, HPN321, KPG_011, PSEC0146, hGPCR21
UniGene
Hs.253706
Gene Chromosome Location
Chr.13: 48653711-48709362 on Build GRCh38
Transcripts
NM_001308465.1 NM_001308467.1 NM_001308468.1 NM_001308469.1 NM_001308470.1 NM_001308471.1 NM_001308476.1 NM_020377.3

SNP Detail

SNP ID Chromosome Location
rs191388414 Chr.13: 48709365-48709365 on Build GRCh38
rs942177059 Chr.13: 48709416-48709416 on Build GRCh38
rs950421438 Chr.13: 48709654-48709654 on Build GRCh38
rs202241447 Chr.13: 48709183-48709183 on Build GRCh38
rs1040318734 Chr.13: 48709264-48709264 on Build GRCh38
rs557051146 Chr.13: 48709480-48709480 on Build GRCh38
rs201596957 Chr.13: 48709395-48709395 on Build GRCh38
rs944545156 Chr.13: 48709457-48709457 on Build GRCh38
rs201495751 Chr.13: 48709331-48709331 on Build GRCh38
rs568803889 Chr.13: 48709377-48709377 on Build GRCh38
rs961919887 Chr.13: 48709368-48709368 on Build GRCh38
rs1016183808 Chr.13: 48709629-48709629 on Build GRCh38
rs950417428 Chr.13: 48709378-48709378 on Build GRCh38
rs976317740 Chr.13: 48709463-48709463 on Build GRCh38
rs1057186507 Chr.13: 48709277-48709277 on Build GRCh38
rs563699529 Chr.13: 48709527-48709527 on Build GRCh38
rs912277 Chr.13: 48709515-48709515 on Build GRCh38
rs1030472105 Chr.13: 48709633-48709633 on Build GRCh38
rs927436396 Chr.13: 48709450-48709450 on Build GRCh38
rs537470183 Chr.13: 48709319-48709319 on Build GRCh38
rs1021269152 Chr.13: 48709339-48709339 on Build GRCh38
rs980304125 Chr.13: 48709423-48709423 on Build GRCh38
rs200395119 Chr.13: 48709328-48709328 on Build GRCh38
rs61183001 Chr.13: 48709668-48709668 on Build GRCh38
rs765054183 Chr.13: 48709187-48709187 on Build GRCh38
rs1043626579 Chr.13: 48709492-48709492 on Build GRCh38
rs200341304 Chr.13: 48709281-48709281 on Build GRCh38
rs185091766 Chr.13: 48709553-48709553 on Build GRCh38
rs141215411 Chr.13: 48709481-48709481 on Build GRCh38
rs746044642 Chr.13: 48709552-48709552 on Build GRCh38
rs181058850 Chr.13: 48709372-48709372 on Build GRCh38
rs145097279 Chr.13: 48709528-48709528 on Build GRCh38
rs201217026 Chr.13: 48709238-48709238 on Build GRCh38
rs768846993 Chr.13: 48709608-48709608 on Build GRCh38
rs201471962 Chr.13: 48709297-48709297 on Build GRCh38
rs201112163 Chr.13: 48709181-48709181 on Build GRCh38
rs780223417 Chr.13: 48709565-48709565 on Build GRCh38
rs912638463 Chr.13: 48709454-48709454 on Build GRCh38
rs537337477 Chr.13: 48709405-48709408 on Build GRCh38
rs535657121 Chr.13: 48709451-48709451 on Build GRCh38
rs74809557 Chr.13: 48709346-48709346 on Build GRCh38
rs149666125 Chr.13: 48709237-48709237 on Build GRCh38
rs571585193 Chr.13: 48709591-48709591 on Build GRCh38
rs900331836 Chr.13: 48709616-48709616 on Build GRCh38
rs1013690558 Chr.13: 48709355-48709355 on Build GRCh38
rs912278 Chr.13: 48709351-48709351 on Build GRCh38
rs747063023 Chr.13: 48709605-48709605 on Build GRCh38
rs983733137 Chr.13: 48709394-48709394 on Build GRCh38
rs1000622473 Chr.13: 48709347-48709347 on Build GRCh38
rs892358017 Chr.13: 48709269-48709269 on Build GRCh38
rs201969295 Chr.13: 48709324-48709324 on Build GRCh38
rs145528722 Chr.13: 48709261-48709261 on Build GRCh38
rs199804767 Chr.13: 48709338-48709338 on Build GRCh38
rs896239608 Chr.13: 48709547-48709547 on Build GRCh38
rs1015927444 Chr.13: 48709666-48709666 on Build GRCh38
rs1017312907 Chr.13: 48709349-48709349 on Build GRCh38
rs932728825 Chr.13: 48709186-48709186 on Build GRCh38
rs909420850 Chr.13: 48709405-48709405 on Build GRCh38
rs945097147 Chr.13: 48709307-48709307 on Build GRCh38
rs954054672 Chr.13: 48709669-48709669 on Build GRCh38
rs1323552 Chr.13: 48709632-48709632 on Build GRCh38
rs1017427038 Chr.13: 48709604-48709604 on Build GRCh38
rs570035356 Chr.13: 48709288-48709288 on Build GRCh38
rs910405002 Chr.13: 48709256-48709256 on Build GRCh38
rs2407249 Chr.13: 48709659-48709659 on Build GRCh38

View Full Product Details