Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 77000022-77000554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN5
Entrez Gene ID
1203
Gene Name
ceroid-lipofuscinosis, neuronal 5
Gene Aliases
NCL
UniGene
Hs.30213
Gene Chromosome Location
Chr.13: 76991924-77002517 on Build GRCh38
Transcripts
NM_006493.2 XM_011534917.2 XM_017020538.1

Gene Symbol
FBXL3
Entrez Gene ID
26224
Gene Name
F-box and leucine rich repeat protein 3
Gene Aliases
FBL3, FBL3A, FBXL3A
UniGene
Hs.508284
Gene Chromosome Location
Chr.13: 76992597-77027196 on Build GRCh38
Transcripts
NM_006493.2 XM_011534917.2 XM_017020538.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4833896 c.757G>C p.E253Q Chr.13: 77000502-77000502 on Build GRCh38
5991114 c.717C>A p.N239K Chr.13: 77000462-77000462 on Build GRCh38
7361568 c.726C>A p.N242K Chr.13: 77000471-77000471 on Build GRCh38
469562 c.741G>T p.W247C Chr.13: 77000486-77000486 on Build GRCh38
372739 c.760A>G p.T254A Chr.13: 77000505-77000505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs386833981 Chr.13: 77000517-77000517 on Build GRCh38
rs763409893 Chr.13: 77000262-77000262 on Build GRCh38
rs760343764 Chr.13: 77000443-77000444 on Build GRCh38
rs972774933 Chr.13: 77000214-77000216 on Build GRCh38
rs768569438 Chr.13: 77000073-77000073 on Build GRCh38
rs140913712 Chr.13: 77000060-77000065 on Build GRCh38
rs957751304 Chr.13: 77000107-77000107 on Build GRCh38
rs778310133 Chr.13: 77000460-77000460 on Build GRCh38
rs58575113 Chr.13: 77000384-77000384 on Build GRCh38
rs562901964 Chr.13: 77000117-77000117 on Build GRCh38
rs80226022 Chr.13: 77000239-77000239 on Build GRCh38
rs375050582 Chr.13: 77000064-77000069 on Build GRCh38
rs17071289 Chr.13: 77000069-77000069 on Build GRCh38
rs765648216 Chr.13: 77000189-77000189 on Build GRCh38
rs775100164 Chr.13: 77000514-77000514 on Build GRCh38
rs986397211 Chr.13: 77000122-77000122 on Build GRCh38
rs1007869176 Chr.13: 77000359-77000359 on Build GRCh38
rs940044893 Chr.13: 77000194-77000194 on Build GRCh38
rs769765595 Chr.13: 77000188-77000188 on Build GRCh38
rs772501269 Chr.13: 77000451-77000451 on Build GRCh38
rs150138832 Chr.13: 77000348-77000348 on Build GRCh38
rs763980789 Chr.13: 77000519-77000519 on Build GRCh38
rs1018161062 Chr.13: 77000123-77000123 on Build GRCh38
rs1055774310 Chr.13: 77000285-77000285 on Build GRCh38
rs928612392 Chr.13: 77000177-77000177 on Build GRCh38
rs930465173 Chr.13: 77000257-77000257 on Build GRCh38
rs1040327440 Chr.13: 77000360-77000360 on Build GRCh38
rs796514613 Chr.13: 77000403-77000403 on Build GRCh38
rs113639365 Chr.13: 77000137-77000137 on Build GRCh38
rs771601428 Chr.13: 77000435-77000439 on Build GRCh38
rs1037943250 Chr.13: 77000300-77000300 on Build GRCh38
rs898027499 Chr.13: 77000308-77000308 on Build GRCh38
rs544250899 Chr.13: 77000093-77000093 on Build GRCh38
rs11840125 Chr.13: 77000430-77000430 on Build GRCh38
rs762787810 Chr.13: 77000518-77000518 on Build GRCh38
rs767037205 Chr.13: 77000286-77000286 on Build GRCh38
rs190202279 Chr.13: 77000227-77000227 on Build GRCh38
rs990067914 Chr.13: 77000147-77000147 on Build GRCh38
rs138611001 Chr.13: 77000471-77000471 on Build GRCh38
rs942300012 Chr.13: 77000289-77000289 on Build GRCh38
rs369100769 Chr.13: 77000461-77000461 on Build GRCh38
rs998543477 Chr.13: 77000389-77000390 on Build GRCh38
rs879014639 Chr.13: 77000408-77000408 on Build GRCh38
rs775075753 Chr.13: 77000444-77000444 on Build GRCh38
rs761876915 Chr.13: 77000185-77000185 on Build GRCh38
rs548433888 Chr.13: 77000317-77000317 on Build GRCh38
rs534857590 Chr.13: 77000393-77000393 on Build GRCh38
rs377507341 Chr.13: 77000329-77000329 on Build GRCh38
rs541258261 Chr.13: 77000264-77000264 on Build GRCh38
rs914421609 Chr.13: 77000175-77000175 on Build GRCh38
rs945891726 Chr.13: 77000178-77000178 on Build GRCh38
rs67836242 Chr.13: 77000361-77000361 on Build GRCh38
rs771119692 Chr.13: 77000464-77000464 on Build GRCh38
rs879601738 Chr.13: 77000267-77000269 on Build GRCh38
rs530787053 Chr.13: 77000347-77000347 on Build GRCh38
rs140122775 Chr.13: 77000529-77000529 on Build GRCh38
rs548315727 Chr.13: 77000256-77000256 on Build GRCh38
rs58476086 Chr.13: 77000403-77000404 on Build GRCh38
rs1031436024 Chr.13: 77000095-77000095 on Build GRCh38
rs928662833 Chr.13: 77000213-77000213 on Build GRCh38
rs560043541 Chr.13: 77000274-77000274 on Build GRCh38
rs963735922 Chr.13: 77000443-77000443 on Build GRCh38
rs77376472 Chr.13: 77000402-77000402 on Build GRCh38
rs1025356896 Chr.13: 77000315-77000315 on Build GRCh38
rs772636070 Chr.13: 77000273-77000273 on Build GRCh38
rs879845206 Chr.13: 77000260-77000263 on Build GRCh38
rs79224677 Chr.13: 77000405-77000405 on Build GRCh38
rs1025345541 Chr.13: 77000085-77000085 on Build GRCh38
rs769662577 Chr.13: 77000507-77000507 on Build GRCh38
rs759765950 Chr.13: 77000475-77000475 on Build GRCh38
rs141010472 Chr.13: 77000311-77000311 on Build GRCh38

View Full Product Details