Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 77005901-77006414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL3
Entrez Gene ID
26224
Gene Name
F-box and leucine rich repeat protein 3
Gene Aliases
FBL3, FBL3A, FBXL3A
UniGene
Hs.508284
Gene Chromosome Location
Chr.13: 76992597-77027196 on Build GRCh38
Transcripts
NM_012158.2 XM_005266336.1 XM_005266337.2 XM_017020538.1

SNP Detail

SNP ID Chromosome Location
rs915914427 Chr.13: 77006032-77006032 on Build GRCh38
rs962346636 Chr.13: 77006072-77006072 on Build GRCh38
rs938868522 Chr.13: 77006196-77006196 on Build GRCh38
rs1022891216 Chr.13: 77006342-77006342 on Build GRCh38
rs1025477675 Chr.13: 77006097-77006097 on Build GRCh38
rs529817729 Chr.13: 77005971-77005971 on Build GRCh38
rs990148546 Chr.13: 77005998-77006000 on Build GRCh38
rs981537139 Chr.13: 77006372-77006372 on Build GRCh38
rs145523101 Chr.13: 77006100-77006100 on Build GRCh38
rs577060101 Chr.13: 77006378-77006378 on Build GRCh38
rs907365476 Chr.13: 77005973-77005973 on Build GRCh38
rs894623837 Chr.13: 77006298-77006298 on Build GRCh38
rs532576117 Chr.13: 77006180-77006180 on Build GRCh38
rs986626582 Chr.13: 77006105-77006105 on Build GRCh38
rs536354710 Chr.13: 77006301-77006301 on Build GRCh38
rs558855730 Chr.13: 77006338-77006338 on Build GRCh38
rs572163097 Chr.13: 77006064-77006064 on Build GRCh38
rs1036219801 Chr.13: 77006385-77006385 on Build GRCh38
rs565688679 Chr.13: 77006266-77006266 on Build GRCh38
rs199785804 Chr.13: 77006048-77006048 on Build GRCh38
rs1040549134 Chr.13: 77006188-77006191 on Build GRCh38
rs914613170 Chr.13: 77006162-77006162 on Build GRCh38
rs531912967 Chr.13: 77006080-77006080 on Build GRCh38
rs528240621 Chr.13: 77006150-77006150 on Build GRCh38
rs767380321 Chr.13: 77005961-77005961 on Build GRCh38
rs966627054 Chr.13: 77005934-77005942 on Build GRCh38
rs1003007007 Chr.13: 77005988-77005988 on Build GRCh38
rs554617432 Chr.13: 77006302-77006302 on Build GRCh38
rs994085427 Chr.13: 77006086-77006086 on Build GRCh38
rs1011935130 Chr.13: 77006053-77006053 on Build GRCh38
rs893350748 Chr.13: 77006318-77006321 on Build GRCh38
rs999097444 Chr.13: 77005972-77005972 on Build GRCh38
rs999461461 Chr.13: 77006251-77006251 on Build GRCh38
rs911028205 Chr.13: 77006113-77006113 on Build GRCh38
rs551306672 Chr.13: 77006308-77006308 on Build GRCh38
rs970481253 Chr.13: 77006359-77006359 on Build GRCh38
rs201500431 Chr.13: 77006047-77006047 on Build GRCh38
rs970132217 Chr.13: 77006050-77006050 on Build GRCh38
rs888798810 Chr.13: 77006155-77006155 on Build GRCh38
rs559446738 Chr.13: 77005948-77005948 on Build GRCh38
rs561916950 Chr.13: 77005990-77005990 on Build GRCh38
rs939211428 Chr.13: 77006087-77006089 on Build GRCh38
rs930551249 Chr.13: 77006115-77006115 on Build GRCh38
rs957357549 Chr.13: 77005995-77005997 on Build GRCh38
rs760174938 Chr.13: 77005971-77005975 on Build GRCh38
rs138113313 Chr.13: 77006139-77006139 on Build GRCh38
rs541907334 Chr.13: 77006039-77006039 on Build GRCh38
rs116619495 Chr.13: 77006320-77006320 on Build GRCh38
rs867990061 Chr.13: 77006132-77006132 on Build GRCh38
rs142691597 Chr.13: 77006194-77006194 on Build GRCh38
rs964189433 Chr.13: 77006127-77006127 on Build GRCh38
rs1041997671 Chr.13: 77006182-77006182 on Build GRCh38

View Full Product Details