Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 77006549-77007054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL3
Entrez Gene ID
26224
Gene Name
F-box and leucine rich repeat protein 3
Gene Aliases
FBL3, FBL3A, FBXL3A
UniGene
Hs.508284
Gene Chromosome Location
Chr.13: 76992597-77027196 on Build GRCh38
Transcripts
NM_012158.2 XM_005266336.1 XM_005266337.2 XM_017020538.1

SNP Detail

SNP ID Chromosome Location
rs548027467 Chr.13: 77006740-77006740 on Build GRCh38
rs558475664 Chr.13: 77006971-77006971 on Build GRCh38
rs7982885 Chr.13: 77006597-77006597 on Build GRCh38
rs967382551 Chr.13: 77006673-77006673 on Build GRCh38
rs826432 Chr.13: 77006697-77006697 on Build GRCh38
rs960311139 Chr.13: 77006977-77006977 on Build GRCh38
rs1003137044 Chr.13: 77006965-77006965 on Build GRCh38
rs922969957 Chr.13: 77006886-77006886 on Build GRCh38
rs565616462 Chr.13: 77006685-77006685 on Build GRCh38
rs929708892 Chr.13: 77006832-77006832 on Build GRCh38
rs942164960 Chr.13: 77006787-77006787 on Build GRCh38
rs906070088 Chr.13: 77006949-77006950 on Build GRCh38
rs570586287 Chr.13: 77007002-77007002 on Build GRCh38
rs144603004 Chr.13: 77006968-77006968 on Build GRCh38
rs890778264 Chr.13: 77006861-77006861 on Build GRCh38
rs898222883 Chr.13: 77006797-77006797 on Build GRCh38
rs748671291 Chr.13: 77006850-77006850 on Build GRCh38
rs991758091 Chr.13: 77007001-77007001 on Build GRCh38
rs943045470 Chr.13: 77006842-77006846 on Build GRCh38
rs967480573 Chr.13: 77006928-77006928 on Build GRCh38
rs929761365 Chr.13: 77007013-77007013 on Build GRCh38
rs781473688 Chr.13: 77006764-77006764 on Build GRCh38
rs1021683130 Chr.13: 77006652-77006652 on Build GRCh38
rs897174506 Chr.13: 77006980-77006980 on Build GRCh38
rs1038094011 Chr.13: 77006791-77006791 on Build GRCh38
rs1057821 Chr.13: 77006788-77006788 on Build GRCh38
rs530016538 Chr.13: 77006826-77006826 on Build GRCh38
rs766838516 Chr.13: 77006732-77006732 on Build GRCh38
rs973564068 Chr.13: 77007005-77007005 on Build GRCh38
rs543642909 Chr.13: 77006669-77006669 on Build GRCh38
rs1021738754 Chr.13: 77006917-77006917 on Build GRCh38
rs1018140608 Chr.13: 77006593-77006593 on Build GRCh38
rs1035698048 Chr.13: 77006741-77006741 on Build GRCh38
rs905856224 Chr.13: 77006718-77006718 on Build GRCh38
rs1026130007 Chr.13: 77006770-77006771 on Build GRCh38
rs867253318 Chr.13: 77006869-77006869 on Build GRCh38
rs899673721 Chr.13: 77006888-77006888 on Build GRCh38
rs951884941 Chr.13: 77006772-77006772 on Build GRCh38
rs755369045 Chr.13: 77006676-77006676 on Build GRCh38
rs117178661 Chr.13: 77007010-77007010 on Build GRCh38
rs548313462 Chr.13: 77006842-77006842 on Build GRCh38
rs984691343 Chr.13: 77006779-77006779 on Build GRCh38
rs1044626323 Chr.13: 77006947-77006947 on Build GRCh38
rs766673885 Chr.13: 77006612-77006612 on Build GRCh38
rs534629250 Chr.13: 77007004-77007004 on Build GRCh38
rs45620238 Chr.13: 77006887-77006887 on Build GRCh38
rs563349886 Chr.13: 77006630-77006630 on Build GRCh38
rs938977512 Chr.13: 77006702-77006702 on Build GRCh38
rs914723333 Chr.13: 77006918-77006918 on Build GRCh38
rs1011384281 Chr.13: 77006911-77006911 on Build GRCh38
rs183383833 Chr.13: 77006663-77006663 on Build GRCh38
rs893318037 Chr.13: 77006693-77006693 on Build GRCh38
rs367644997 Chr.13: 77006931-77006931 on Build GRCh38
rs187549546 Chr.13: 77006769-77006769 on Build GRCh38
rs1023203372 Chr.13: 77006710-77006710 on Build GRCh38
rs942217430 Chr.13: 77007003-77007003 on Build GRCh38

View Full Product Details