Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 95587456-95587960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DZIP1
Entrez Gene ID
22873
Gene Name
DAZ interacting zinc finger protein 1
Gene Aliases
DZIP, DZIPt1
UniGene
Hs.656580
Gene Chromosome Location
Chr.13: 95578202-95644706 on Build GRCh38
Transcripts
NM_014934.4 NM_198968.3 XM_005254032.1 XM_006719916.1 XM_006719917.2 XM_006719918.1 XM_006719920.1 XM_017020444.1 XM_017020445.1 XM_017020446.1 XM_017020447.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3470026 c.2104C>T p.P702S Chr.13: 95587653-95587653 on Build GRCh38
1951637 c.2212C>G p.P738A Chr.13: 95587545-95587545 on Build GRCh38
4216287 c.2029A>G p.T677A Chr.13: 95587728-95587728 on Build GRCh38
3470022 c.2176G>A p.G726R Chr.13: 95587581-95587581 on Build GRCh38
6650964 c.2173G>A p.E725K Chr.13: 95587584-95587584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977207741 Chr.13: 95587859-95587859 on Build GRCh38
rs1056480760 Chr.13: 95587920-95587920 on Build GRCh38
rs577965114 Chr.13: 95587753-95587753 on Build GRCh38
rs267603867 Chr.13: 95587695-95587695 on Build GRCh38
rs202197331 Chr.13: 95587494-95587494 on Build GRCh38
rs779503028 Chr.13: 95587653-95587653 on Build GRCh38
rs114917226 Chr.13: 95587481-95587481 on Build GRCh38
rs775730696 Chr.13: 95587618-95587618 on Build GRCh38
rs377335041 Chr.13: 95587621-95587621 on Build GRCh38
rs764136702 Chr.13: 95587746-95587746 on Build GRCh38
rs751499210 Chr.13: 95587632-95587632 on Build GRCh38
rs765997376 Chr.13: 95587771-95587771 on Build GRCh38
rs757041731 Chr.13: 95587563-95587563 on Build GRCh38
rs750249882 Chr.13: 95587758-95587758 on Build GRCh38
rs771261333 Chr.13: 95587598-95587598 on Build GRCh38
rs777381535 Chr.13: 95587549-95587549 on Build GRCh38
rs767222103 Chr.13: 95587636-95587636 on Build GRCh38
rs1056973963 Chr.13: 95587536-95587536 on Build GRCh38
rs771258025 Chr.13: 95587689-95587689 on Build GRCh38
rs201380520 Chr.13: 95587619-95587619 on Build GRCh38
rs536467966 Chr.13: 95587581-95587581 on Build GRCh38
rs770171062 Chr.13: 95587704-95587704 on Build GRCh38
rs947012774 Chr.13: 95587824-95587827 on Build GRCh38
rs780873105 Chr.13: 95587570-95587570 on Build GRCh38
rs148618215 Chr.13: 95587667-95587667 on Build GRCh38
rs144822400 Chr.13: 95587646-95587646 on Build GRCh38
rs760772618 Chr.13: 95587506-95587506 on Build GRCh38
rs938442309 Chr.13: 95587914-95587914 on Build GRCh38
rs866515221 Chr.13: 95587629-95587629 on Build GRCh38
rs372822102 Chr.13: 95587519-95587519 on Build GRCh38
rs368463836 Chr.13: 95587759-95587759 on Build GRCh38
rs1013116095 Chr.13: 95587602-95587602 on Build GRCh38
rs371016866 Chr.13: 95587576-95587576 on Build GRCh38
rs374870127 Chr.13: 95587611-95587611 on Build GRCh38
rs754497843 Chr.13: 95587672-95587672 on Build GRCh38
rs926522922 Chr.13: 95587812-95587812 on Build GRCh38
rs773635726 Chr.13: 95587597-95587597 on Build GRCh38
rs779554094 Chr.13: 95587585-95587585 on Build GRCh38
rs376315570 Chr.13: 95587542-95587542 on Build GRCh38
rs116643114 Chr.13: 95587492-95587492 on Build GRCh38
rs765476930 Chr.13: 95587533-95587533 on Build GRCh38
rs267603868 Chr.13: 95587696-95587696 on Build GRCh38
rs772020920 Chr.13: 95587591-95587591 on Build GRCh38
rs141127846 Chr.13: 95587674-95587674 on Build GRCh38
rs111764824 Chr.13: 95587645-95587645 on Build GRCh38
rs775894382 Chr.13: 95587723-95587723 on Build GRCh38
rs749585345 Chr.13: 95587676-95587676 on Build GRCh38
rs765347543 Chr.13: 95587616-95587616 on Build GRCh38
rs976428588 Chr.13: 95587624-95587624 on Build GRCh38
rs149427250 Chr.13: 95587635-95587635 on Build GRCh38
rs781577041 Chr.13: 95587599-95587600 on Build GRCh38
rs892427028 Chr.13: 95587928-95587928 on Build GRCh38
rs201600364 Chr.13: 95587592-95587592 on Build GRCh38
rs142424864 Chr.13: 95587595-95587595 on Build GRCh38
rs753549694 Chr.13: 95587778-95587778 on Build GRCh38
rs762313730 Chr.13: 95587491-95587491 on Build GRCh38
rs774280342 Chr.13: 95587751-95587751 on Build GRCh38
rs754058464 Chr.13: 95587526-95587526 on Build GRCh38
rs563846714 Chr.13: 95587797-95587797 on Build GRCh38
rs909691485 Chr.13: 95587477-95587477 on Build GRCh38
rs938816418 Chr.13: 95587524-95587524 on Build GRCh38
rs370788709 Chr.13: 95587650-95587650 on Build GRCh38
rs746034662 Chr.13: 95587700-95587700 on Build GRCh38
rs748362339 Chr.13: 95587686-95587687 on Build GRCh38
rs529410751 Chr.13: 95587877-95587877 on Build GRCh38
rs775499708 Chr.13: 95587620-95587620 on Build GRCh38
rs755095914 Chr.13: 95587503-95587504 on Build GRCh38
rs763101283 Chr.13: 95587733-95587733 on Build GRCh38
rs761672431 Chr.13: 95587752-95587752 on Build GRCh38
rs776868438 Chr.13: 95587692-95587692 on Build GRCh38
rs542843844 Chr.13: 95587883-95587883 on Build GRCh38
rs11070136 Chr.13: 95587551-95587551 on Build GRCh38
rs759456440 Chr.13: 95587584-95587584 on Build GRCh38
rs776905933 Chr.13: 95587604-95587604 on Build GRCh38

View Full Product Details