Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 95786898-95787445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC3
Entrez Gene ID
5611
Gene Name
DnaJ heat shock protein family (Hsp40) member C3
Gene Aliases
ACPHD, ERdj6, HP58, P58, P58IPK, PRKRI
UniGene
Hs.59214
Gene Chromosome Location
Chr.13: 95677139-95794989 on Build GRCh38
Transcripts
NM_006260.4 XM_011521104.2 XM_011521105.1 XM_017020674.1 XM_017020675.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
338501 c.1209-1G>A p.? Chr.13: 95787006-95787006 on Build GRCh38
4828173 c.1209-1G>C p.? Chr.13: 95787006-95787006 on Build GRCh38
4166615 c.1243C>T p.R415* Chr.13: 95787041-95787041 on Build GRCh38
949031 c.1292A>G p.E431G Chr.13: 95787090-95787090 on Build GRCh38
4524432 c.1270G>T p.D424Y Chr.13: 95787068-95787068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185832840 Chr.13: 95787121-95787121 on Build GRCh38
rs17882407 Chr.13: 95787381-95787381 on Build GRCh38
rs1012227577 Chr.13: 95787317-95787317 on Build GRCh38
rs944448620 Chr.13: 95787148-95787148 on Build GRCh38
rs887447274 Chr.13: 95787246-95787246 on Build GRCh38
rs953409004 Chr.13: 95787298-95787298 on Build GRCh38
rs958019765 Chr.13: 95787327-95787327 on Build GRCh38
rs988577704 Chr.13: 95787345-95787345 on Build GRCh38
rs777106962 Chr.13: 95786984-95786987 on Build GRCh38
rs17885953 Chr.13: 95787216-95787216 on Build GRCh38
rs202072148 Chr.13: 95787000-95787000 on Build GRCh38
rs767087128 Chr.13: 95786956-95786956 on Build GRCh38
rs545428697 Chr.13: 95787153-95787153 on Build GRCh38
rs561895883 Chr.13: 95787257-95787257 on Build GRCh38
rs542140095 Chr.13: 95787249-95787249 on Build GRCh38
rs773544778 Chr.13: 95787258-95787258 on Build GRCh38
rs750714045 Chr.13: 95787207-95787207 on Build GRCh38
rs907261951 Chr.13: 95787241-95787241 on Build GRCh38
rs768498039 Chr.13: 95787041-95787041 on Build GRCh38
rs965783573 Chr.13: 95787391-95787391 on Build GRCh38
rs533921429 Chr.13: 95786969-95786969 on Build GRCh38
rs910275280 Chr.13: 95787147-95787147 on Build GRCh38
rs771748365 Chr.13: 95787112-95787112 on Build GRCh38
rs17881125 Chr.13: 95786930-95786930 on Build GRCh38
rs867959661 Chr.13: 95787285-95787285 on Build GRCh38
rs986216979 Chr.13: 95787310-95787310 on Build GRCh38
rs948556509 Chr.13: 95787235-95787235 on Build GRCh38
rs748237816 Chr.13: 95786987-95786991 on Build GRCh38
rs781542314 Chr.13: 95787199-95787199 on Build GRCh38
rs929200869 Chr.13: 95787206-95787206 on Build GRCh38
rs201172876 Chr.13: 95787180-95787180 on Build GRCh38
rs574670452 Chr.13: 95786934-95786934 on Build GRCh38
rs1021899730 Chr.13: 95787329-95787329 on Build GRCh38
rs138435112 Chr.13: 95787040-95787040 on Build GRCh38
rs1044633409 Chr.13: 95787236-95787236 on Build GRCh38
rs774135777 Chr.13: 95787048-95787048 on Build GRCh38
rs997672122 Chr.13: 95787286-95787286 on Build GRCh38
rs778152919 Chr.13: 95786999-95786999 on Build GRCh38
rs1031627771 Chr.13: 95787269-95787269 on Build GRCh38
rs753246719 Chr.13: 95786973-95786973 on Build GRCh38
rs753371769 Chr.13: 95787169-95787169 on Build GRCh38
rs376804113 Chr.13: 95786960-95786960 on Build GRCh38
rs560356546 Chr.13: 95787387-95787387 on Build GRCh38
rs764514828 Chr.13: 95787177-95787177 on Build GRCh38
rs371390750 Chr.13: 95787102-95787102 on Build GRCh38
rs954456513 Chr.13: 95787368-95787368 on Build GRCh38
rs770043101 Chr.13: 95787021-95787021 on Build GRCh38
rs780107548 Chr.13: 95787030-95787030 on Build GRCh38
rs760150982 Chr.13: 95787142-95787142 on Build GRCh38
rs1022202097 Chr.13: 95787035-95787035 on Build GRCh38
rs745930127 Chr.13: 95787001-95787001 on Build GRCh38
rs758938629 Chr.13: 95786978-95786978 on Build GRCh38
rs769797527 Chr.13: 95787193-95787219 on Build GRCh38
rs1055351954 Chr.13: 95787220-95787222 on Build GRCh38
rs935866746 Chr.13: 95787163-95787163 on Build GRCh38
rs548701173 Chr.13: 95786951-95786951 on Build GRCh38
rs2289813 Chr.13: 95787278-95787278 on Build GRCh38
rs1002915793 Chr.13: 95787259-95787259 on Build GRCh38
rs1030930352 Chr.13: 95787295-95787295 on Build GRCh38
rs761515145 Chr.13: 95787053-95787053 on Build GRCh38
rs925281679 Chr.13: 95787162-95787162 on Build GRCh38
rs763461583 Chr.13: 95787175-95787175 on Build GRCh38
rs1005882198 Chr.13: 95787253-95787255 on Build GRCh38
rs956249780 Chr.13: 95786976-95786976 on Build GRCh38

View Full Product Details