Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 102731720-102732231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC168
Entrez Gene ID
643677
Gene Name
coiled-coil domain containing 168
Gene Aliases
C13orf40
UniGene
Hs.368382
Gene Chromosome Location
Chr.13: 102729367-102759072 on Build GRCh38
Transcripts
NM_001146197.1 XM_011521106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1365264 c.4905C>T p.I1635I Chr.13: 102731905-102731905 on Build GRCh38
1365265 c.4821delA p.K1607fs*3 Chr.13: 102731989-102731989 on Build GRCh38
7225005 c.5072C>T p.P1691L Chr.13: 102731738-102731738 on Build GRCh38
4650137 c.4838C>T p.T1613I Chr.13: 102731972-102731972 on Build GRCh38
6225373 c.5047G>A p.A1683T Chr.13: 102731763-102731763 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758656324 Chr.13: 102732146-102732146 on Build GRCh38
rs536877767 Chr.13: 102731879-102731879 on Build GRCh38
rs774117233 Chr.13: 102731980-102731980 on Build GRCh38
rs534716273 Chr.13: 102731784-102731784 on Build GRCh38
rs1041513496 Chr.13: 102731837-102731837 on Build GRCh38
rs1003516335 Chr.13: 102732013-102732013 on Build GRCh38
rs745818204 Chr.13: 102731928-102731928 on Build GRCh38
rs752940990 Chr.13: 102731866-102731866 on Build GRCh38
rs373455595 Chr.13: 102732072-102732072 on Build GRCh38
rs768405500 Chr.13: 102731972-102731972 on Build GRCh38
rs775384324 Chr.13: 102731963-102731963 on Build GRCh38
rs760097996 Chr.13: 102732114-102732114 on Build GRCh38
rs894273065 Chr.13: 102731889-102731889 on Build GRCh38
rs998709873 Chr.13: 102732196-102732196 on Build GRCh38
rs999137397 Chr.13: 102731787-102731787 on Build GRCh38
rs550683435 Chr.13: 102732126-102732126 on Build GRCh38
rs866478231 Chr.13: 102731788-102731788 on Build GRCh38
rs960283297 Chr.13: 102731869-102731869 on Build GRCh38
rs555275413 Chr.13: 102731961-102731961 on Build GRCh38
rs1035618539 Chr.13: 102731836-102731836 on Build GRCh38
rs888815149 Chr.13: 102731989-102731989 on Build GRCh38
rs1011325089 Chr.13: 102732028-102732028 on Build GRCh38
rs753131590 Chr.13: 102732132-102732132 on Build GRCh38
rs181400241 Chr.13: 102731824-102731824 on Build GRCh38
rs933629741 Chr.13: 102731798-102731798 on Build GRCh38
rs566264361 Chr.13: 102731868-102731868 on Build GRCh38
rs951551837 Chr.13: 102732068-102732068 on Build GRCh38
rs186542518 Chr.13: 102731964-102731964 on Build GRCh38
rs537011276 Chr.13: 102732018-102732018 on Build GRCh38
rs748979990 Chr.13: 102732036-102732036 on Build GRCh38
rs764362201 Chr.13: 102732207-102732207 on Build GRCh38
rs781160708 Chr.13: 102731908-102731908 on Build GRCh38
rs754393763 Chr.13: 102731778-102731778 on Build GRCh38
rs867956404 Chr.13: 102732161-102732161 on Build GRCh38
rs908712754 Chr.13: 102731906-102731906 on Build GRCh38
rs952785392 Chr.13: 102731904-102731904 on Build GRCh38
rs761481193 Chr.13: 102732071-102732071 on Build GRCh38
rs765652233 Chr.13: 102731794-102731794 on Build GRCh38
rs757320027 Chr.13: 102731905-102731905 on Build GRCh38
rs577208241 Chr.13: 102732075-102732075 on Build GRCh38
rs758682044 Chr.13: 102731876-102731876 on Build GRCh38
rs765205143 Chr.13: 102731912-102731912 on Build GRCh38
rs1035991984 Chr.13: 102732200-102732200 on Build GRCh38
rs761722559 Chr.13: 102731880-102731880 on Build GRCh38

View Full Product Details