Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 106489993-106490488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNB2
Entrez Gene ID
1948
Gene Name
ephrin B2
Gene Aliases
EPLG5, HTKL, Htk-L, LERK5
UniGene
Hs.149239
Gene Chromosome Location
Chr.13: 106489731-106535040 on Build GRCh38
Transcripts
NM_004093.3 XM_017020406.1

SNP Detail

SNP ID Chromosome Location
rs904503688 Chr.13: 106490303-106490303 on Build GRCh38
rs1007806121 Chr.13: 106490278-106490278 on Build GRCh38
rs543806858 Chr.13: 106490432-106490432 on Build GRCh38
rs1043732107 Chr.13: 106490045-106490045 on Build GRCh38
rs147699607 Chr.13: 106490339-106490339 on Build GRCh38
rs766699055 Chr.13: 106490302-106490302 on Build GRCh38
rs761042959 Chr.13: 106490436-106490436 on Build GRCh38
rs764644883 Chr.13: 106490450-106490450 on Build GRCh38
rs889457773 Chr.13: 106490364-106490364 on Build GRCh38
rs572040062 Chr.13: 106490326-106490326 on Build GRCh38
rs188440777 Chr.13: 106490377-106490377 on Build GRCh38
rs948125102 Chr.13: 106490414-106490414 on Build GRCh38
rs781204585 Chr.13: 106490347-106490347 on Build GRCh38
rs933513550 Chr.13: 106490462-106490462 on Build GRCh38
rs935337768 Chr.13: 106490456-106490456 on Build GRCh38
rs56077446 Chr.13: 106490151-106490151 on Build GRCh38
rs558257367 Chr.13: 106490248-106490248 on Build GRCh38
rs956584071 Chr.13: 106490338-106490338 on Build GRCh38
rs935202396 Chr.13: 106490098-106490098 on Build GRCh38
rs754227899 Chr.13: 106490024-106490024 on Build GRCh38
rs961694781 Chr.13: 106490335-106490335 on Build GRCh38
rs1006421324 Chr.13: 106490304-106490304 on Build GRCh38
rs751755894 Chr.13: 106490313-106490313 on Build GRCh38
rs545135627 Chr.13: 106490163-106490163 on Build GRCh38
rs750770677 Chr.13: 106490173-106490173 on Build GRCh38
rs747989071 Chr.13: 106490463-106490463 on Build GRCh38
rs1036057795 Chr.13: 106490072-106490072 on Build GRCh38
rs534822495 Chr.13: 106490073-106490073 on Build GRCh38
rs767710658 Chr.13: 106490172-106490172 on Build GRCh38
rs948172710 Chr.13: 106490033-106490034 on Build GRCh38
rs901653992 Chr.13: 106490229-106490229 on Build GRCh38
rs556251042 Chr.13: 106490076-106490076 on Build GRCh38
rs908881020 Chr.13: 106490048-106490048 on Build GRCh38
rs369201447 Chr.13: 106490152-106490152 on Build GRCh38
rs997331878 Chr.13: 106490250-106490250 on Build GRCh38
rs960976231 Chr.13: 106490351-106490351 on Build GRCh38
rs971808903 Chr.13: 106490445-106490445 on Build GRCh38
rs1043116887 Chr.13: 106490291-106490291 on Build GRCh38
rs980639120 Chr.13: 106490398-106490398 on Build GRCh38
rs908912159 Chr.13: 106490422-106490422 on Build GRCh38
rs918964638 Chr.13: 106490458-106490458 on Build GRCh38
rs941808470 Chr.13: 106490057-106490057 on Build GRCh38

View Full Product Details