Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21098706-21099253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM253
Entrez Gene ID
643382
Gene Name
transmembrane protein 253
Gene Aliases
C14orf176, C14orf95, NCRNA00220
UniGene
Hs.540107
Gene Chromosome Location
Chr.14: 21098937-21103724 on Build GRCh38
Transcripts
NM_001146683.1 XM_011537078.2 XM_011537082.2 NM_001101672.1 NM_001102454.1 XM_005267739.1 XM_006720164.3

Gene Symbol
ZNF219
Entrez Gene ID
51222
Gene Name
zinc finger protein 219
Gene Aliases
ZFP219
UniGene
Hs.250493
Gene Chromosome Location
Chr.14: 21090046-21104704 on Build GRCh38
Transcripts
NM_001146683.1 XM_011537078.2 XM_011537082.2 NM_001101672.1 NM_001102454.1 XM_005267739.1 XM_006720164.3

SNP Detail

SNP ID Chromosome Location
rs1012758851 Chr.14: 21098821-21098821 on Build GRCh38
rs895191454 Chr.14: 21098820-21098820 on Build GRCh38
rs565327875 Chr.14: 21099074-21099074 on Build GRCh38
rs761685518 Chr.14: 21098983-21098984 on Build GRCh38
rs1014658104 Chr.14: 21098755-21098755 on Build GRCh38
rs948118258 Chr.14: 21099187-21099187 on Build GRCh38
rs920742785 Chr.14: 21099005-21099005 on Build GRCh38
rs1003661457 Chr.14: 21098776-21098776 on Build GRCh38
rs1000157852 Chr.14: 21099224-21099224 on Build GRCh38
rs780975787 Chr.14: 21098815-21098815 on Build GRCh38
rs143814256 Chr.14: 21099177-21099177 on Build GRCh38
rs901517024 Chr.14: 21099191-21099191 on Build GRCh38
rs12889457 Chr.14: 21099174-21099174 on Build GRCh38
rs542857911 Chr.14: 21098916-21098916 on Build GRCh38
rs1029204351 Chr.14: 21098793-21098793 on Build GRCh38
rs775042924 Chr.14: 21099134-21099134 on Build GRCh38
rs904349297 Chr.14: 21099207-21099207 on Build GRCh38
rs1036471825 Chr.14: 21099185-21099185 on Build GRCh38
rs1056443982 Chr.14: 21099175-21099175 on Build GRCh38
rs983694127 Chr.14: 21099057-21099057 on Build GRCh38
rs953232307 Chr.14: 21098801-21098801 on Build GRCh38
rs771701955 Chr.14: 21098861-21098861 on Build GRCh38
rs1000510519 Chr.14: 21098894-21098894 on Build GRCh38
rs964788253 Chr.14: 21098981-21098981 on Build GRCh38
rs559356509 Chr.14: 21098952-21098952 on Build GRCh38
rs1056556788 Chr.14: 21098810-21098810 on Build GRCh38
rs960368273 Chr.14: 21098953-21098953 on Build GRCh38
rs745460237 Chr.14: 21098929-21098929 on Build GRCh38
rs769515528 Chr.14: 21099004-21099004 on Build GRCh38
rs141801608 Chr.14: 21099077-21099077 on Build GRCh38
rs930938123 Chr.14: 21099046-21099046 on Build GRCh38
rs756355848 Chr.14: 21098862-21098862 on Build GRCh38
rs563448693 Chr.14: 21098847-21098847 on Build GRCh38
rs751524840 Chr.14: 21098775-21098775 on Build GRCh38
rs1048662678 Chr.14: 21099161-21099161 on Build GRCh38
rs191453915 Chr.14: 21098913-21098913 on Build GRCh38
rs551555110 Chr.14: 21098973-21098973 on Build GRCh38
rs772620075 Chr.14: 21099212-21099212 on Build GRCh38
rs991588751 Chr.14: 21098988-21098988 on Build GRCh38
rs528390137 Chr.14: 21099006-21099006 on Build GRCh38
rs911651877 Chr.14: 21098976-21098976 on Build GRCh38
rs543586855 Chr.14: 21098767-21098767 on Build GRCh38
rs564627691 Chr.14: 21099123-21099123 on Build GRCh38
rs756689342 Chr.14: 21098896-21098896 on Build GRCh38
rs1023197904 Chr.14: 21098836-21098836 on Build GRCh38
rs754840402 Chr.14: 21098803-21098803 on Build GRCh38
rs1044268646 Chr.14: 21099198-21099198 on Build GRCh38
rs762384406 Chr.14: 21099030-21099035 on Build GRCh38
rs551445571 Chr.14: 21099036-21099036 on Build GRCh38
rs997350828 Chr.14: 21098781-21098781 on Build GRCh38
rs908816901 Chr.14: 21099166-21099166 on Build GRCh38

View Full Product Details