Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21101784-21102290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM253
Entrez Gene ID
643382
Gene Name
transmembrane protein 253
Gene Aliases
C14orf176, C14orf95, NCRNA00220
UniGene
Hs.540107
Gene Chromosome Location
Chr.14: 21098937-21103724 on Build GRCh38
Transcripts
NM_001146683.1 XM_006720233.3 XM_011537078.2 XM_011537079.1 XM_011537081.2 XM_011537082.2 XM_011537083.2 XM_011537084.2 XM_011537085.2 NM_001102454.1 XM_006720164.3

Gene Symbol
ZNF219
Entrez Gene ID
51222
Gene Name
zinc finger protein 219
Gene Aliases
ZFP219
UniGene
Hs.250493
Gene Chromosome Location
Chr.14: 21090046-21104704 on Build GRCh38
Transcripts
NM_001146683.1 XM_006720233.3 XM_011537078.2 XM_011537079.1 XM_011537081.2 XM_011537082.2 XM_011537083.2 XM_011537084.2 XM_011537085.2 NM_001102454.1 XM_006720164.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7294185 c.160G>A p.A54T Chr.14: 21101916-21101916 on Build GRCh38
7232681 c.270T>C p.L90L Chr.14: 21102114-21102114 on Build GRCh38
3983159 c.228C>T p.L76L Chr.14: 21102072-21102072 on Build GRCh38
954358 c.195G>A p.A65A Chr.14: 21101951-21101951 on Build GRCh38
1152705 c.196C>T p.L66L Chr.14: 21101952-21101952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780603176 Chr.14: 21102021-21102021 on Build GRCh38
rs778750437 Chr.14: 21101994-21101994 on Build GRCh38
rs905511403 Chr.14: 21102099-21102099 on Build GRCh38
rs754712981 Chr.14: 21101975-21101975 on Build GRCh38
rs745696 Chr.14: 21102114-21102114 on Build GRCh38
rs773894082 Chr.14: 21102093-21102093 on Build GRCh38
rs962559060 Chr.14: 21101888-21101888 on Build GRCh38
rs1016719252 Chr.14: 21101860-21101860 on Build GRCh38
rs79713019 Chr.14: 21101998-21101998 on Build GRCh38
rs183259006 Chr.14: 21102098-21102098 on Build GRCh38
rs752694251 Chr.14: 21102105-21102105 on Build GRCh38
rs897208794 Chr.14: 21102224-21102224 on Build GRCh38
rs901389905 Chr.14: 21101877-21101877 on Build GRCh38
rs61450523 Chr.14: 21102115-21102115 on Build GRCh38
rs535943725 Chr.14: 21102128-21102128 on Build GRCh38
rs757512802 Chr.14: 21101959-21101959 on Build GRCh38
rs996873083 Chr.14: 21101879-21101879 on Build GRCh38
rs754094609 Chr.14: 21101953-21101953 on Build GRCh38
rs368718917 Chr.14: 21101847-21101847 on Build GRCh38
rs755925330 Chr.14: 21101950-21101950 on Build GRCh38
rs769073990 Chr.14: 21102026-21102026 on Build GRCh38
rs993290147 Chr.14: 21101962-21101962 on Build GRCh38
rs767412875 Chr.14: 21101916-21101916 on Build GRCh38
rs189090055 Chr.14: 21102138-21102138 on Build GRCh38
rs199880599 Chr.14: 21102110-21102110 on Build GRCh38
rs770551710 Chr.14: 21102077-21102077 on Build GRCh38
rs751024919 Chr.14: 21101895-21101895 on Build GRCh38
rs776824899 Chr.14: 21102057-21102057 on Build GRCh38
rs1038125779 Chr.14: 21102182-21102182 on Build GRCh38
rs1016141151 Chr.14: 21101932-21101932 on Build GRCh38
rs762619558 Chr.14: 21102065-21102065 on Build GRCh38
rs757600846 Chr.14: 21102121-21102121 on Build GRCh38
rs571270162 Chr.14: 21101814-21101814 on Build GRCh38
rs746479973 Chr.14: 21102061-21102063 on Build GRCh38
rs914057166 Chr.14: 21101828-21101828 on Build GRCh38
rs1023936595 Chr.14: 21101811-21101811 on Build GRCh38
rs898276138 Chr.14: 21102211-21102211 on Build GRCh38
rs1025861656 Chr.14: 21102265-21102265 on Build GRCh38
rs955436929 Chr.14: 21101922-21101922 on Build GRCh38
rs569981936 Chr.14: 21102097-21102097 on Build GRCh38
rs772902600 Chr.14: 21101843-21101843 on Build GRCh38
rs942716993 Chr.14: 21101941-21101941 on Build GRCh38
rs532039799 Chr.14: 21101915-21101915 on Build GRCh38
rs372143791 Chr.14: 21102025-21102025 on Build GRCh38
rs765909179 Chr.14: 21101862-21101862 on Build GRCh38
rs763829745 Chr.14: 21101866-21101866 on Build GRCh38
rs779655564 Chr.14: 21101851-21101852 on Build GRCh38
rs1053903301 Chr.14: 21101809-21101809 on Build GRCh38
rs367771233 Chr.14: 21102137-21102137 on Build GRCh38
rs367883550 Chr.14: 21101981-21101981 on Build GRCh38
rs1041286570 Chr.14: 21101872-21101872 on Build GRCh38
rs796183645 Chr.14: 21102074-21102074 on Build GRCh38
rs1024899643 Chr.14: 21101969-21101969 on Build GRCh38
rs746366161 Chr.14: 21102125-21102125 on Build GRCh38
rs913616778 Chr.14: 21102035-21102035 on Build GRCh38
rs905544156 Chr.14: 21101824-21101824 on Build GRCh38
rs374427336 Chr.14: 21101935-21101935 on Build GRCh38
rs568737292 Chr.14: 21102164-21102164 on Build GRCh38
rs772743009 Chr.14: 21102009-21102009 on Build GRCh38
rs1005627159 Chr.14: 21101930-21101930 on Build GRCh38
rs370554010 Chr.14: 21101951-21101951 on Build GRCh38

View Full Product Details