Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21102997-21103525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM253
Entrez Gene ID
643382
Gene Name
transmembrane protein 253
Gene Aliases
C14orf176, C14orf95, NCRNA00220
UniGene
Hs.540107
Gene Chromosome Location
Chr.14: 21098937-21103724 on Build GRCh38
Transcripts
NM_001146683.1 XM_006720233.3 XM_011537078.2 XM_011537079.1 XM_011537081.2 XM_011537082.2 XM_011537083.2 XM_011537084.2 XM_011537085.2 NM_001102454.1 XM_006720164.3

Gene Symbol
ZNF219
Entrez Gene ID
51222
Gene Name
zinc finger protein 219
Gene Aliases
ZFP219
UniGene
Hs.250493
Gene Chromosome Location
Chr.14: 21090046-21104704 on Build GRCh38
Transcripts
NM_001146683.1 XM_006720233.3 XM_011537078.2 XM_011537079.1 XM_011537081.2 XM_011537082.2 XM_011537083.2 XM_011537084.2 XM_011537085.2 NM_001102454.1 XM_006720164.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3706177 c.556G>T p.V186F Chr.14: 21103160-21103160 on Build GRCh38
4793334 c.611T>C p.V204A Chr.14: 21103215-21103215 on Build GRCh38
6015439 c.640C>G p.L214V Chr.14: 21103244-21103244 on Build GRCh38
1629235 c.544G>A p.E182K Chr.14: 21103148-21103148 on Build GRCh38
7291302 c.610G>A p.V204M Chr.14: 21103214-21103214 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761781290 Chr.14: 21103217-21103217 on Build GRCh38
rs1017094160 Chr.14: 21103355-21103355 on Build GRCh38
rs1000708801 Chr.14: 21103235-21103235 on Build GRCh38
rs929629599 Chr.14: 21103469-21103469 on Build GRCh38
rs541450097 Chr.14: 21103380-21103380 on Build GRCh38
rs1057718 Chr.14: 21103464-21103464 on Build GRCh38
rs200993615 Chr.14: 21103226-21103226 on Build GRCh38
rs780649914 Chr.14: 21103404-21103404 on Build GRCh38
rs1029832344 Chr.14: 21103308-21103308 on Build GRCh38
rs974052580 Chr.14: 21103411-21103411 on Build GRCh38
rs1009607226 Chr.14: 21103304-21103304 on Build GRCh38
rs150024369 Chr.14: 21103397-21103397 on Build GRCh38
rs893748439 Chr.14: 21103239-21103239 on Build GRCh38
rs755001481 Chr.14: 21103302-21103302 on Build GRCh38
rs997824599 Chr.14: 21103303-21103303 on Build GRCh38
rs537619570 Chr.14: 21103346-21103346 on Build GRCh38
rs763288851 Chr.14: 21103229-21103229 on Build GRCh38
rs568302904 Chr.14: 21103428-21103428 on Build GRCh38
rs534666750 Chr.14: 21103208-21103208 on Build GRCh38
rs980856772 Chr.14: 21103375-21103376 on Build GRCh38
rs919557252 Chr.14: 21103050-21103050 on Build GRCh38
rs57471121 Chr.14: 21103064-21103064 on Build GRCh38
rs1007487128 Chr.14: 21103331-21103331 on Build GRCh38
rs905914886 Chr.14: 21103163-21103163 on Build GRCh38
rs140414254 Chr.14: 21103266-21103266 on Build GRCh38
rs953790888 Chr.14: 21103324-21103324 on Build GRCh38
rs1025470542 Chr.14: 21103311-21103311 on Build GRCh38
rs368217898 Chr.14: 21103098-21103098 on Build GRCh38
rs981438857 Chr.14: 21103484-21103484 on Build GRCh38
rs765124716 Chr.14: 21103218-21103218 on Build GRCh38
rs773747539 Chr.14: 21103224-21103224 on Build GRCh38
rs746338417 Chr.14: 21103206-21103206 on Build GRCh38
rs937424325 Chr.14: 21103493-21103493 on Build GRCh38
rs963553081 Chr.14: 21103407-21103407 on Build GRCh38
rs1039472796 Chr.14: 21103220-21103220 on Build GRCh38
rs890915704 Chr.14: 21103115-21103115 on Build GRCh38
rs1046228971 Chr.14: 21103152-21103152 on Build GRCh38
rs962984615 Chr.14: 21103371-21103371 on Build GRCh38
rs145365080 Chr.14: 21103490-21103490 on Build GRCh38
rs751730317 Chr.14: 21103288-21103288 on Build GRCh38
rs115088850 Chr.14: 21103422-21103422 on Build GRCh38
rs929688055 Chr.14: 21103054-21103054 on Build GRCh38
rs1010767588 Chr.14: 21103268-21103268 on Build GRCh38
rs972972081 Chr.14: 21103379-21103379 on Build GRCh38
rs12880628 Chr.14: 21103049-21103049 on Build GRCh38
rs943797682 Chr.14: 21103142-21103142 on Build GRCh38
rs1042073637 Chr.14: 21103286-21103286 on Build GRCh38
rs950195515 Chr.14: 21103110-21103110 on Build GRCh38

View Full Product Details