Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 22976561-22977071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AJUBA
Entrez Gene ID
84962
Gene Name
ajuba LIM protein
Gene Aliases
JUB
UniGene
Hs.655832
Gene Chromosome Location
Chr.14: 22971174-22982642 on Build GRCh38
Transcripts
NM_032876.5 NM_198086.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4522543 c.1168G>A p.D390N Chr.14: 22976653-22976653 on Build GRCh38
1493209 c.1128G>T p.K376N Chr.14: 22976693-22976693 on Build GRCh38
3369929 c.1145A>G p.N382S Chr.14: 22976676-22976676 on Build GRCh38
4848858 c.1111C>T p.R371* Chr.14: 22976710-22976710 on Build GRCh38
4817275 c.1168G>C p.D390H Chr.14: 22976653-22976653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568641435 Chr.14: 22976919-22976919 on Build GRCh38
rs200090663 Chr.14: 22976626-22976626 on Build GRCh38
rs1049982541 Chr.14: 22977014-22977014 on Build GRCh38
rs932654854 Chr.14: 22976995-22976995 on Build GRCh38
rs752480248 Chr.14: 22976764-22976768 on Build GRCh38
rs369756410 Chr.14: 22976792-22976792 on Build GRCh38
rs768070804 Chr.14: 22976843-22976843 on Build GRCh38
rs547935184 Chr.14: 22976899-22976899 on Build GRCh38
rs959269123 Chr.14: 22976785-22976785 on Build GRCh38
rs1034687016 Chr.14: 22976765-22976765 on Build GRCh38
rs368135450 Chr.14: 22976946-22976946 on Build GRCh38
rs188186488 Chr.14: 22976845-22976845 on Build GRCh38
rs994609639 Chr.14: 22976952-22976952 on Build GRCh38
rs958549144 Chr.14: 22977041-22977041 on Build GRCh38
rs776110106 Chr.14: 22977039-22977039 on Build GRCh38
rs997122397 Chr.14: 22976683-22976683 on Build GRCh38
rs981344955 Chr.14: 22977037-22977037 on Build GRCh38
rs1025642095 Chr.14: 22976988-22976988 on Build GRCh38
rs562314468 Chr.14: 22976808-22976808 on Build GRCh38
rs370292100 Chr.14: 22976663-22976663 on Build GRCh38
rs779708491 Chr.14: 22976794-22976794 on Build GRCh38
rs776576069 Chr.14: 22976715-22976715 on Build GRCh38
rs746812676 Chr.14: 22976707-22976707 on Build GRCh38
rs1051227687 Chr.14: 22976852-22976852 on Build GRCh38
rs902258551 Chr.14: 22977043-22977043 on Build GRCh38
rs747506674 Chr.14: 22976972-22976972 on Build GRCh38
rs1028480159 Chr.14: 22976698-22976698 on Build GRCh38
rs751930617 Chr.14: 22976635-22976635 on Build GRCh38
rs1011091608 Chr.14: 22976728-22976728 on Build GRCh38
rs373374150 Chr.14: 22976682-22976682 on Build GRCh38
rs770186251 Chr.14: 22976917-22976917 on Build GRCh38
rs756119022 Chr.14: 22976676-22976676 on Build GRCh38
rs1034240154 Chr.14: 22977038-22977038 on Build GRCh38
rs532177650 Chr.14: 22976786-22976786 on Build GRCh38
rs764998253 Chr.14: 22976600-22976600 on Build GRCh38
rs370473701 Chr.14: 22976733-22976733 on Build GRCh38
rs200978900 Chr.14: 22976601-22976601 on Build GRCh38
rs779750264 Chr.14: 22976706-22976706 on Build GRCh38
rs971744481 Chr.14: 22977015-22977015 on Build GRCh38
rs768530887 Chr.14: 22976710-22976710 on Build GRCh38
rs367620944 Chr.14: 22976754-22976754 on Build GRCh38
rs781720935 Chr.14: 22976648-22976648 on Build GRCh38
rs937103484 Chr.14: 22976712-22976712 on Build GRCh38
rs561161756 Chr.14: 22976615-22976615 on Build GRCh38
rs560465344 Chr.14: 22976780-22976780 on Build GRCh38
rs761416020 Chr.14: 22976732-22976732 on Build GRCh38
rs771115920 Chr.14: 22976700-22976700 on Build GRCh38
rs748768764 Chr.14: 22976821-22976821 on Build GRCh38
rs922628848 Chr.14: 22976939-22976939 on Build GRCh38
rs1041051111 Chr.14: 22976927-22976927 on Build GRCh38
rs930508049 Chr.14: 22976585-22976585 on Build GRCh38
rs749462895 Chr.14: 22976684-22976684 on Build GRCh38
rs201959157 Chr.14: 22976736-22976736 on Build GRCh38
rs762751270 Chr.14: 22976639-22976642 on Build GRCh38
rs889938160 Chr.14: 22976853-22976853 on Build GRCh38
rs940162081 Chr.14: 22976918-22976918 on Build GRCh38
rs369150221 Chr.14: 22976598-22976598 on Build GRCh38
rs751937613 Chr.14: 22976756-22976756 on Build GRCh38
rs755479715 Chr.14: 22976637-22976637 on Build GRCh38
rs945703750 Chr.14: 22976778-22976778 on Build GRCh38
rs902796479 Chr.14: 22976819-22976819 on Build GRCh38
rs927095175 Chr.14: 22976660-22976660 on Build GRCh38

View Full Product Details