Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23052381-23052876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH24
Entrez Gene ID
64403
Gene Name
cadherin 24
Gene Aliases
CDH11L
UniGene
Hs.155912
Gene Chromosome Location
Chr.14: 23047061-23057619 on Build GRCh38
Transcripts
NM_022478.3 NM_144985.3 XM_011537089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6625012 c.1254G>A p.P418P Chr.14: 23052582-23052582 on Build GRCh38
3955916 c.1325G>A p.R442H Chr.14: 23052511-23052511 on Build GRCh38
4670929 c.1342A>G p.T448A Chr.14: 23052494-23052494 on Build GRCh38
7157604 c.1318G>A p.E440K Chr.14: 23052518-23052518 on Build GRCh38
1477467 c.1301C>A p.A434E Chr.14: 23052535-23052535 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575134680 Chr.14: 23052714-23052714 on Build GRCh38
rs766379218 Chr.14: 23052613-23052613 on Build GRCh38
rs777165147 Chr.14: 23052459-23052459 on Build GRCh38
rs763508518 Chr.14: 23052500-23052500 on Build GRCh38
rs765796891 Chr.14: 23052404-23052404 on Build GRCh38
rs969824321 Chr.14: 23052618-23052618 on Build GRCh38
rs183768991 Chr.14: 23052406-23052406 on Build GRCh38
rs751771818 Chr.14: 23052429-23052429 on Build GRCh38
rs1024238244 Chr.14: 23052743-23052743 on Build GRCh38
rs936796225 Chr.14: 23052668-23052668 on Build GRCh38
rs766378415 Chr.14: 23052509-23052509 on Build GRCh38
rs770587246 Chr.14: 23052505-23052506 on Build GRCh38
rs1050111158 Chr.14: 23052795-23052795 on Build GRCh38
rs779549121 Chr.14: 23052451-23052451 on Build GRCh38
rs781037505 Chr.14: 23052562-23052562 on Build GRCh38
rs56251692 Chr.14: 23052793-23052793 on Build GRCh38
rs750183388 Chr.14: 23052597-23052597 on Build GRCh38
rs552602186 Chr.14: 23052447-23052447 on Build GRCh38
rs747784902 Chr.14: 23052566-23052566 on Build GRCh38
rs186696970 Chr.14: 23052418-23052418 on Build GRCh38
rs771171698 Chr.14: 23052643-23052643 on Build GRCh38
rs778025537 Chr.14: 23052641-23052641 on Build GRCh38
rs538293845 Chr.14: 23052673-23052673 on Build GRCh38
rs768305211 Chr.14: 23052458-23052458 on Build GRCh38
rs754590298 Chr.14: 23052623-23052623 on Build GRCh38
rs759702442 Chr.14: 23052583-23052583 on Build GRCh38
rs548615172 Chr.14: 23052558-23052558 on Build GRCh38
rs1014234818 Chr.14: 23052733-23052733 on Build GRCh38
rs775482024 Chr.14: 23052661-23052661 on Build GRCh38
rs759373642 Chr.14: 23052584-23052585 on Build GRCh38
rs752398946 Chr.14: 23052627-23052627 on Build GRCh38
rs773649311 Chr.14: 23052577-23052577 on Build GRCh38
rs1005973481 Chr.14: 23052816-23052816 on Build GRCh38
rs746654322 Chr.14: 23052455-23052455 on Build GRCh38
rs773910869 Chr.14: 23052474-23052474 on Build GRCh38
rs558119794 Chr.14: 23052702-23052702 on Build GRCh38
rs35692156 Chr.14: 23052823-23052823 on Build GRCh38
rs772207238 Chr.14: 23052655-23052655 on Build GRCh38
rs1045209021 Chr.14: 23052416-23052416 on Build GRCh38
rs780013499 Chr.14: 23052648-23052648 on Build GRCh38
rs763893296 Chr.14: 23052419-23052419 on Build GRCh38
rs942619685 Chr.14: 23052832-23052832 on Build GRCh38
rs879772102 Chr.14: 23052766-23052766 on Build GRCh38
rs147450903 Chr.14: 23052512-23052512 on Build GRCh38
rs772119411 Chr.14: 23052585-23052585 on Build GRCh38
rs538488794 Chr.14: 23052671-23052671 on Build GRCh38
rs778863234 Chr.14: 23052435-23052435 on Build GRCh38
rs1041727505 Chr.14: 23052762-23052762 on Build GRCh38
rs757904056 Chr.14: 23052449-23052449 on Build GRCh38
rs904624116 Chr.14: 23052409-23052409 on Build GRCh38
rs767768911 Chr.14: 23052510-23052510 on Build GRCh38
rs1013492339 Chr.14: 23052829-23052829 on Build GRCh38
rs766465202 Chr.14: 23052408-23052408 on Build GRCh38
rs767688832 Chr.14: 23052514-23052514 on Build GRCh38
rs775747588 Chr.14: 23052591-23052591 on Build GRCh38
rs111407353 Chr.14: 23052593-23052593 on Build GRCh38
rs749096320 Chr.14: 23052404-23052405 on Build GRCh38
rs779451314 Chr.14: 23052557-23052557 on Build GRCh38
rs750499050 Chr.14: 23052520-23052520 on Build GRCh38
rs57329219 Chr.14: 23052824-23052824 on Build GRCh38
rs755774421 Chr.14: 23052637-23052637 on Build GRCh38
rs980835614 Chr.14: 23052635-23052635 on Build GRCh38
rs751799984 Chr.14: 23052413-23052413 on Build GRCh38
rs577605236 Chr.14: 23052826-23052826 on Build GRCh38
rs944302375 Chr.14: 23052758-23052758 on Build GRCh38
rs199718070 Chr.14: 23052578-23052578 on Build GRCh38
rs751427420 Chr.14: 23052616-23052616 on Build GRCh38
rs374513962 Chr.14: 23052624-23052624 on Build GRCh38
rs148111355 Chr.14: 23052511-23052511 on Build GRCh38
rs754504269 Chr.14: 23052560-23052560 on Build GRCh38
rs2331830 Chr.14: 23052465-23052465 on Build GRCh38
rs774372531 Chr.14: 23052583-23052584 on Build GRCh38
rs770258783 Chr.14: 23052473-23052473 on Build GRCh38
rs887539314 Chr.14: 23052798-23052798 on Build GRCh38
rs373969242 Chr.14: 23052717-23052717 on Build GRCh38
rs369762156 Chr.14: 23052519-23052519 on Build GRCh38
rs375742256 Chr.14: 23052653-23052653 on Build GRCh38
rs757218480 Chr.14: 23052432-23052432 on Build GRCh38
rs752887104 Chr.14: 23052518-23052518 on Build GRCh38
rs758296999 Chr.14: 23052540-23052540 on Build GRCh38
rs554615851 Chr.14: 23052791-23052791 on Build GRCh38
rs750394497 Chr.14: 23052521-23052521 on Build GRCh38
rs762827550 Chr.14: 23052600-23052600 on Build GRCh38
rs756058912 Chr.14: 23052581-23052581 on Build GRCh38
rs192321646 Chr.14: 23052647-23052647 on Build GRCh38
rs972887019 Chr.14: 23052796-23052796 on Build GRCh38

View Full Product Details