Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23053370-23053865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH24
Entrez Gene ID
64403
Gene Name
cadherin 24
Gene Aliases
CDH11L
UniGene
Hs.155912
Gene Chromosome Location
Chr.14: 23047061-23057619 on Build GRCh38
Transcripts
NM_022478.3 NM_144985.3 XM_011537089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4670930 c.1193C>T p.A398V Chr.14: 23053529-23053529 on Build GRCh38
6838545 c.1129G>T p.A377S Chr.14: 23053593-23053593 on Build GRCh38
4670932 c.1095G>T p.V365V Chr.14: 23053627-23053627 on Build GRCh38
6838547 c.1102G>T p.A368S Chr.14: 23053620-23053620 on Build GRCh38
7139827 c.1048C>T p.L350L Chr.14: 23053674-23053674 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755865066 Chr.14: 23053500-23053500 on Build GRCh38
rs150135230 Chr.14: 23053767-23053767 on Build GRCh38
rs767525829 Chr.14: 23053636-23053636 on Build GRCh38
rs771973764 Chr.14: 23053685-23053685 on Build GRCh38
rs751576916 Chr.14: 23053462-23053462 on Build GRCh38
rs748311894 Chr.14: 23053587-23053587 on Build GRCh38
rs756794379 Chr.14: 23053650-23053650 on Build GRCh38
rs767098720 Chr.14: 23053556-23053556 on Build GRCh38
rs775628380 Chr.14: 23053690-23053690 on Build GRCh38
rs762853301 Chr.14: 23053461-23053461 on Build GRCh38
rs368415384 Chr.14: 23053666-23053666 on Build GRCh38
rs563161053 Chr.14: 23053637-23053637 on Build GRCh38
rs768594265 Chr.14: 23053559-23053559 on Build GRCh38
rs772518973 Chr.14: 23053530-23053530 on Build GRCh38
rs368280984 Chr.14: 23053670-23053670 on Build GRCh38
rs757700243 Chr.14: 23053519-23053519 on Build GRCh38
rs776685659 Chr.14: 23053454-23053454 on Build GRCh38
rs752210711 Chr.14: 23053471-23053471 on Build GRCh38
rs746980735 Chr.14: 23053782-23053782 on Build GRCh38
rs919892216 Chr.14: 23053707-23053707 on Build GRCh38
rs932905670 Chr.14: 23053420-23053420 on Build GRCh38
rs972150125 Chr.14: 23053833-23053833 on Build GRCh38
rs765084547 Chr.14: 23053595-23053595 on Build GRCh38
rs200684669 Chr.14: 23053699-23053699 on Build GRCh38
rs577468762 Chr.14: 23053433-23053433 on Build GRCh38
rs377064007 Chr.14: 23053548-23053548 on Build GRCh38
rs747498115 Chr.14: 23053667-23053667 on Build GRCh38
rs758086932 Chr.14: 23053683-23053683 on Build GRCh38
rs768824233 Chr.14: 23053445-23053445 on Build GRCh38
rs750053080 Chr.14: 23053718-23053718 on Build GRCh38
rs753753123 Chr.14: 23053556-23053556 on Build GRCh38
rs779931001 Chr.14: 23053781-23053781 on Build GRCh38
rs781350801 Chr.14: 23053752-23053752 on Build GRCh38
rs746379911 Chr.14: 23053529-23053529 on Build GRCh38
rs145622932 Chr.14: 23053705-23053705 on Build GRCh38
rs748140441 Chr.14: 23053756-23053756 on Build GRCh38
rs753594961 Chr.14: 23053649-23053649 on Build GRCh38
rs908873063 Chr.14: 23053437-23053437 on Build GRCh38
rs753411461 Chr.14: 23053514-23053514 on Build GRCh38
rs199897921 Chr.14: 23053565-23053565 on Build GRCh38
rs747501624 Chr.14: 23053390-23053390 on Build GRCh38
rs370996228 Chr.14: 23053582-23053582 on Build GRCh38
rs750745464 Chr.14: 23053660-23053660 on Build GRCh38
rs759476763 Chr.14: 23053465-23053465 on Build GRCh38
rs987977212 Chr.14: 23053550-23053550 on Build GRCh38
rs559917130 Chr.14: 23053758-23053758 on Build GRCh38
rs773956377 Chr.14: 23053619-23053619 on Build GRCh38
rs766251914 Chr.14: 23053528-23053528 on Build GRCh38
rs765648344 Chr.14: 23053655-23053655 on Build GRCh38
rs781062574 Chr.14: 23053669-23053669 on Build GRCh38
rs754773200 Chr.14: 23053577-23053577 on Build GRCh38
rs768430522 Chr.14: 23053795-23053795 on Build GRCh38
rs760771694 Chr.14: 23053402-23053402 on Build GRCh38
rs1018052102 Chr.14: 23053455-23053455 on Build GRCh38
rs941717129 Chr.14: 23053448-23053448 on Build GRCh38
rs956410444 Chr.14: 23053798-23053798 on Build GRCh38
rs189915240 Chr.14: 23053671-23053671 on Build GRCh38
rs749511407 Chr.14: 23053771-23053771 on Build GRCh38
rs917827943 Chr.14: 23053552-23053552 on Build GRCh38
rs185739956 Chr.14: 23053469-23053469 on Build GRCh38
rs763665789 Chr.14: 23053708-23053708 on Build GRCh38
rs769059058 Chr.14: 23053808-23053808 on Build GRCh38
rs575672582 Chr.14: 23053393-23053393 on Build GRCh38
rs1000554642 Chr.14: 23053516-23053516 on Build GRCh38
rs759572756 Chr.14: 23053621-23053621 on Build GRCh38
rs758730540 Chr.14: 23053662-23053662 on Build GRCh38
rs963049531 Chr.14: 23053809-23053809 on Build GRCh38
rs780138714 Chr.14: 23053531-23053531 on Build GRCh38
rs770761026 Chr.14: 23053604-23053604 on Build GRCh38
rs772061618 Chr.14: 23053776-23053776 on Build GRCh38
rs774978811 Chr.14: 23053795-23053795 on Build GRCh38
rs774323762 Chr.14: 23053672-23053672 on Build GRCh38
rs547444086 Chr.14: 23053731-23053731 on Build GRCh38
rs984322766 Chr.14: 23053800-23053800 on Build GRCh38
rs755377837 Chr.14: 23053749-23053749 on Build GRCh38
rs777524523 Chr.14: 23053512-23053512 on Build GRCh38
rs199667974 Chr.14: 23053668-23053668 on Build GRCh38
rs370840001 Chr.14: 23053712-23053712 on Build GRCh38
rs745701083 Chr.14: 23053676-23053676 on Build GRCh38
rs74763939 Chr.14: 23053728-23053728 on Build GRCh38
rs368110959 Chr.14: 23053759-23053759 on Build GRCh38
rs764837282 Chr.14: 23053713-23053713 on Build GRCh38
rs763500859 Chr.14: 23053638-23053638 on Build GRCh38
rs905067429 Chr.14: 23053555-23053555 on Build GRCh38
rs893792262 Chr.14: 23053646-23053646 on Build GRCh38

View Full Product Details