Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24075088-24075590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE6
Entrez Gene ID
9362
Gene Name
copine 6
Gene Aliases
-
UniGene
Hs.6132
Gene Chromosome Location
Chr.14: 24070837-24078100 on Build GRCh38
Transcripts
NM_001280558.1 NM_006032.3 XM_005268216.1 XM_005268217.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5027827 c.775G>C p.E259Q Chr.14: 24075274-24075274 on Build GRCh38
3936485 c.785G>A p.W262* Chr.14: 24075512-24075512 on Build GRCh38
5792572 c.798C>A p.N266K Chr.14: 24075525-24075525 on Build GRCh38
6521371 c.936G>A p.G312G Chr.14: 24075270-24075270 on Build GRCh38
3885928 c.755G>A p.G252E Chr.14: 24075254-24075254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569293799 Chr.14: 24075117-24075117 on Build GRCh38
rs888044762 Chr.14: 24075182-24075182 on Build GRCh38
rs770676238 Chr.14: 24075291-24075291 on Build GRCh38
rs758679803 Chr.14: 24075468-24075468 on Build GRCh38
rs945122681 Chr.14: 24075411-24075411 on Build GRCh38
rs766950533 Chr.14: 24075200-24075200 on Build GRCh38
rs369875632 Chr.14: 24075463-24075463 on Build GRCh38
rs775462282 Chr.14: 24075147-24075147 on Build GRCh38
rs777609525 Chr.14: 24075280-24075280 on Build GRCh38
rs745734343 Chr.14: 24075518-24075518 on Build GRCh38
rs769761804 Chr.14: 24075279-24075279 on Build GRCh38
rs754350770 Chr.14: 24075456-24075456 on Build GRCh38
rs776283249 Chr.14: 24075546-24075546 on Build GRCh38
rs142535870 Chr.14: 24075375-24075375 on Build GRCh38
rs75347172 Chr.14: 24075570-24075570 on Build GRCh38
rs766929108 Chr.14: 24075336-24075336 on Build GRCh38
rs771724164 Chr.14: 24075520-24075520 on Build GRCh38
rs748051353 Chr.14: 24075149-24075149 on Build GRCh38
rs750632519 Chr.14: 24075254-24075254 on Build GRCh38
rs762799361 Chr.14: 24075162-24075162 on Build GRCh38
rs749057723 Chr.14: 24075288-24075288 on Build GRCh38
rs554532366 Chr.14: 24075258-24075258 on Build GRCh38
rs527555523 Chr.14: 24075422-24075422 on Build GRCh38
rs376799418 Chr.14: 24075535-24075535 on Build GRCh38
rs370885987 Chr.14: 24075122-24075122 on Build GRCh38
rs1019122088 Chr.14: 24075362-24075362 on Build GRCh38
rs967098372 Chr.14: 24075425-24075425 on Build GRCh38
rs752166444 Chr.14: 24075201-24075201 on Build GRCh38
rs757747996 Chr.14: 24075232-24075232 on Build GRCh38
rs752049882 Chr.14: 24075376-24075376 on Build GRCh38
rs779320376 Chr.14: 24075132-24075132 on Build GRCh38
rs561752228 Chr.14: 24075406-24075406 on Build GRCh38
rs778144831 Chr.14: 24075126-24075126 on Build GRCh38
rs754200666 Chr.14: 24075220-24075220 on Build GRCh38
rs745921993 Chr.14: 24075135-24075135 on Build GRCh38
rs780203852 Chr.14: 24075471-24075471 on Build GRCh38
rs576889549 Chr.14: 24075301-24075301 on Build GRCh38
rs989759125 Chr.14: 24075407-24075407 on Build GRCh38
rs370690636 Chr.14: 24075560-24075560 on Build GRCh38
rs758698066 Chr.14: 24075257-24075257 on Build GRCh38
rs768324856 Chr.14: 24075161-24075161 on Build GRCh38
rs200954701 Chr.14: 24075236-24075236 on Build GRCh38
rs564075739 Chr.14: 24075499-24075499 on Build GRCh38
rs554873847 Chr.14: 24075152-24075152 on Build GRCh38
rs773704385 Chr.14: 24075569-24075569 on Build GRCh38
rs747112244 Chr.14: 24075264-24075264 on Build GRCh38
rs765902727 Chr.14: 24075567-24075567 on Build GRCh38
rs540905941 Chr.14: 24075481-24075481 on Build GRCh38
rs753209687 Chr.14: 24075124-24075124 on Build GRCh38
rs779444184 Chr.14: 24075253-24075253 on Build GRCh38
rs556218212 Chr.14: 24075354-24075354 on Build GRCh38
rs756586826 Chr.14: 24075125-24075125 on Build GRCh38
rs902048715 Chr.14: 24075140-24075140 on Build GRCh38
rs957295607 Chr.14: 24075384-24075384 on Build GRCh38
rs774037684 Chr.14: 24075187-24075187 on Build GRCh38
rs201683346 Chr.14: 24075127-24075127 on Build GRCh38
rs755058815 Chr.14: 24075482-24075482 on Build GRCh38
rs373558163 Chr.14: 24075514-24075514 on Build GRCh38
rs759134968 Chr.14: 24075294-24075294 on Build GRCh38
rs150373138 Chr.14: 24075159-24075159 on Build GRCh38
rs761234762 Chr.14: 24075207-24075207 on Build GRCh38
rs760398800 Chr.14: 24075536-24075536 on Build GRCh38
rs1014962876 Chr.14: 24075266-24075266 on Build GRCh38
rs1034244669 Chr.14: 24075381-24075381 on Build GRCh38
rs138155826 Chr.14: 24075366-24075366 on Build GRCh38
rs201205758 Chr.14: 24075313-24075313 on Build GRCh38
rs781352622 Chr.14: 24075494-24075494 on Build GRCh38
rs764641219 Chr.14: 24075320-24075320 on Build GRCh38
rs1003374166 Chr.14: 24075217-24075217 on Build GRCh38
rs772227566 Chr.14: 24075146-24075146 on Build GRCh38
rs932216844 Chr.14: 24075153-24075153 on Build GRCh38
rs759220922 Chr.14: 24075193-24075193 on Build GRCh38
rs534518114 Chr.14: 24075174-24075174 on Build GRCh38
rs373434263 Chr.14: 24075216-24075216 on Build GRCh38

View Full Product Details