Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24117977-24118479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF11
Entrez Gene ID
80344
Gene Name
DDB1 and CUL4 associated factor 11
Gene Aliases
GL014, PRO2389, WDR23
UniGene
Hs.525251
Gene Chromosome Location
Chr.14: 24114697-24125242 on Build GRCh38
Transcripts
NM_001163484.1 NM_025230.4 NM_181357.2 NR_028099.1 NR_028100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4148029 c.620G>A p.R207H Chr.14: 24118430-24118430 on Build GRCh38
954884 c.644G>A p.R215H Chr.14: 24118454-24118454 on Build GRCh38
1730995 c.477-1G>A p.? Chr.14: 24118054-24118054 on Build GRCh38
5422033 c.508A>G p.S170G Chr.14: 24118086-24118086 on Build GRCh38
3495332 c.590G>A p.R197Q Chr.14: 24118400-24118400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201961254 Chr.14: 24118190-24118190 on Build GRCh38
rs759327535 Chr.14: 24118140-24118140 on Build GRCh38
rs948674949 Chr.14: 24118235-24118235 on Build GRCh38
rs963947029 Chr.14: 24118327-24118327 on Build GRCh38
rs899507071 Chr.14: 24118166-24118166 on Build GRCh38
rs191768847 Chr.14: 24118160-24118160 on Build GRCh38
rs367858431 Chr.14: 24118420-24118420 on Build GRCh38
rs982978233 Chr.14: 24118363-24118363 on Build GRCh38
rs1019527939 Chr.14: 24118112-24118112 on Build GRCh38
rs370828167 Chr.14: 24118017-24118017 on Build GRCh38
rs778907712 Chr.14: 24118351-24118351 on Build GRCh38
rs559136856 Chr.14: 24118066-24118066 on Build GRCh38
rs998972054 Chr.14: 24118259-24118259 on Build GRCh38
rs763340668 Chr.14: 24118410-24118410 on Build GRCh38
rs745670580 Chr.14: 24118356-24118356 on Build GRCh38
rs2277481 Chr.14: 24118336-24118336 on Build GRCh38
rs566743075 Chr.14: 24118309-24118309 on Build GRCh38
rs183350518 Chr.14: 24118029-24118029 on Build GRCh38
rs186195111 Chr.14: 24118150-24118150 on Build GRCh38
rs775034648 Chr.14: 24118365-24118365 on Build GRCh38
rs759734838 Chr.14: 24118459-24118459 on Build GRCh38
rs760179824 Chr.14: 24118394-24118394 on Build GRCh38
rs151212223 Chr.14: 24118311-24118311 on Build GRCh38
rs377556570 Chr.14: 24118455-24118455 on Build GRCh38
rs368549286 Chr.14: 24118171-24118171 on Build GRCh38
rs373152781 Chr.14: 24118019-24118019 on Build GRCh38
rs377657161 Chr.14: 24118023-24118023 on Build GRCh38
rs931087213 Chr.14: 24118183-24118183 on Build GRCh38
rs766575438 Chr.14: 24118412-24118412 on Build GRCh38
rs774115158 Chr.14: 24118089-24118089 on Build GRCh38
rs2277480 Chr.14: 24118306-24118306 on Build GRCh38
rs963647265 Chr.14: 24118137-24118137 on Build GRCh38
rs973439664 Chr.14: 24118310-24118310 on Build GRCh38
rs755034469 Chr.14: 24118338-24118338 on Build GRCh38
rs770898555 Chr.14: 24118086-24118086 on Build GRCh38
rs142387074 Chr.14: 24118079-24118079 on Build GRCh38
rs531379729 Chr.14: 24118269-24118269 on Build GRCh38
rs17101367 Chr.14: 24118458-24118458 on Build GRCh38
rs780260343 Chr.14: 24118207-24118207 on Build GRCh38
rs912116879 Chr.14: 24118116-24118116 on Build GRCh38
rs774170158 Chr.14: 24118006-24118006 on Build GRCh38
rs773499857 Chr.14: 24118404-24118404 on Build GRCh38
rs768203244 Chr.14: 24118437-24118437 on Build GRCh38
rs768097303 Chr.14: 24118395-24118395 on Build GRCh38
rs149132073 Chr.14: 24118236-24118236 on Build GRCh38
rs575893059 Chr.14: 24118119-24118119 on Build GRCh38
rs754320005 Chr.14: 24118169-24118169 on Build GRCh38
rs904591758 Chr.14: 24118237-24118237 on Build GRCh38
rs151306695 Chr.14: 24118429-24118429 on Build GRCh38
rs145287754 Chr.14: 24118189-24118189 on Build GRCh38
rs764618430 Chr.14: 24118164-24118164 on Build GRCh38
rs770845553 Chr.14: 24118345-24118345 on Build GRCh38
rs547952788 Chr.14: 24118279-24118279 on Build GRCh38
rs750421668 Chr.14: 24118002-24118002 on Build GRCh38
rs751779478 Chr.14: 24118414-24118414 on Build GRCh38
rs759093739 Chr.14: 24118014-24118014 on Build GRCh38
rs775948849 Chr.14: 24118438-24118438 on Build GRCh38
rs3825584 Chr.14: 24118430-24118430 on Build GRCh38
rs183061328 Chr.14: 24118208-24118208 on Build GRCh38
rs780169242 Chr.14: 24118058-24118058 on Build GRCh38
rs371898654 Chr.14: 24118343-24118343 on Build GRCh38
rs143253976 Chr.14: 24118280-24118280 on Build GRCh38
rs140411068 Chr.14: 24118344-24118344 on Build GRCh38
rs749063211 Chr.14: 24118080-24118080 on Build GRCh38
rs894739945 Chr.14: 24118228-24118228 on Build GRCh38
rs747665449 Chr.14: 24118442-24118442 on Build GRCh38
rs750688032 Chr.14: 24118034-24118034 on Build GRCh38
rs757202262 Chr.14: 24118422-24118422 on Build GRCh38
rs975061446 Chr.14: 24118155-24118155 on Build GRCh38
rs192332230 Chr.14: 24118454-24118454 on Build GRCh38
rs938944497 Chr.14: 24118210-24118210 on Build GRCh38
rs755053674 Chr.14: 24118417-24118417 on Build GRCh38
rs975436748 Chr.14: 24118406-24118406 on Build GRCh38
rs772041063 Chr.14: 24118111-24118111 on Build GRCh38
rs1000679395 Chr.14: 24118027-24118027 on Build GRCh38
rs754161976 Chr.14: 24118025-24118025 on Build GRCh38
rs745615600 Chr.14: 24118428-24118428 on Build GRCh38
rs2277479 Chr.14: 24118295-24118295 on Build GRCh38
rs145411486 Chr.14: 24118436-24118436 on Build GRCh38
rs771781591 Chr.14: 24118360-24118360 on Build GRCh38
rs747073593 Chr.14: 24118065-24118065 on Build GRCh38
rs377458609 Chr.14: 24118053-24118053 on Build GRCh38
rs767176066 Chr.14: 24118015-24118015 on Build GRCh38
rs993026594 Chr.14: 24118400-24118400 on Build GRCh38
rs373070338 Chr.14: 24118424-24118424 on Build GRCh38
rs758561532 Chr.14: 24118206-24118206 on Build GRCh38
rs753081565 Chr.14: 24118418-24118418 on Build GRCh38
rs371242058 Chr.14: 24118071-24118071 on Build GRCh38
rs538753994 Chr.14: 24118041-24118041 on Build GRCh38
rs762369220 Chr.14: 24118399-24118399 on Build GRCh38
rs1057495012 Chr.14: 24118213-24118213 on Build GRCh38
rs1019920084 Chr.14: 24118323-24118323 on Build GRCh38
rs781372280 Chr.14: 24118396-24118396 on Build GRCh38
rs371332713 Chr.14: 24118421-24118421 on Build GRCh38
rs1012925355 Chr.14: 24118004-24118004 on Build GRCh38
rs562277616 Chr.14: 24118266-24118266 on Build GRCh38
rs576815916 Chr.14: 24118212-24118212 on Build GRCh38
rs751619807 Chr.14: 24118337-24118337 on Build GRCh38

View Full Product Details