Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24317846-24318361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY4
Entrez Gene ID
196883
Gene Name
adenylate cyclase 4
Gene Aliases
AC4
UniGene
Hs.443428
Gene Chromosome Location
Chr.14: 24318349-24335071 on Build GRCh38
Transcripts
NM_001198568.1 NM_001198592.1 NM_139247.3 NM_001143919.2 NM_181657.3

Gene Symbol
LTB4R
Entrez Gene ID
1241
Gene Name
leukotriene B4 receptor
Gene Aliases
BLT1, BLTR, CMKRL1, GPR16, LTB4R1, LTBR1, P2RY7, P2Y7
UniGene
Hs.567248
Gene Chromosome Location
Chr.14: 24311499-24318036 on Build GRCh38
Transcripts
NM_001198568.1 NM_001198592.1 NM_139247.3 NM_001143919.2 NM_181657.3

SNP Detail

SNP ID Chromosome Location
rs890024411 Chr.14: 24317899-24317899 on Build GRCh38
rs934021204 Chr.14: 24318099-24318099 on Build GRCh38
rs959721427 Chr.14: 24317926-24317926 on Build GRCh38
rs994156508 Chr.14: 24318268-24318268 on Build GRCh38
rs998336861 Chr.14: 24317874-24317874 on Build GRCh38
rs760368981 Chr.14: 24318225-24318225 on Build GRCh38
rs901200046 Chr.14: 24318045-24318045 on Build GRCh38
rs948433624 Chr.14: 24317954-24317954 on Build GRCh38
rs770959670 Chr.14: 24318294-24318294 on Build GRCh38
rs760181714 Chr.14: 24318020-24318020 on Build GRCh38
rs909711775 Chr.14: 24317989-24317989 on Build GRCh38
rs1024363399 Chr.14: 24317967-24317967 on Build GRCh38
rs528520641 Chr.14: 24317977-24317977 on Build GRCh38
rs945009168 Chr.14: 24318135-24318135 on Build GRCh38
rs1029011279 Chr.14: 24317889-24317889 on Build GRCh38
rs934176958 Chr.14: 24318200-24318200 on Build GRCh38
rs1036934289 Chr.14: 24318044-24318044 on Build GRCh38
rs897170283 Chr.14: 24318246-24318246 on Build GRCh38
rs567033351 Chr.14: 24318243-24318243 on Build GRCh38
rs992507636 Chr.14: 24318256-24318257 on Build GRCh38
rs534535394 Chr.14: 24317933-24317933 on Build GRCh38
rs751174261 Chr.14: 24318287-24318289 on Build GRCh38
rs530877490 Chr.14: 24318100-24318100 on Build GRCh38
rs970984482 Chr.14: 24317993-24317993 on Build GRCh38
rs753555021 Chr.14: 24318108-24318108 on Build GRCh38
rs972817208 Chr.14: 24317965-24317965 on Build GRCh38
rs150364578 Chr.14: 24318184-24318184 on Build GRCh38
rs961495460 Chr.14: 24317949-24317949 on Build GRCh38
rs999760518 Chr.14: 24318241-24318241 on Build GRCh38
rs72694355 Chr.14: 24317934-24317934 on Build GRCh38
rs561026171 Chr.14: 24317898-24317898 on Build GRCh38
rs1023017700 Chr.14: 24318266-24318266 on Build GRCh38
rs551463639 Chr.14: 24318027-24318027 on Build GRCh38
rs140536712 Chr.14: 24317887-24317887 on Build GRCh38
rs983870926 Chr.14: 24318271-24318271 on Build GRCh38
rs550769438 Chr.14: 24318140-24318140 on Build GRCh38
rs961221613 Chr.14: 24318245-24318245 on Build GRCh38
rs111415008 Chr.14: 24318206-24318206 on Build GRCh38
rs982388059 Chr.14: 24318298-24318298 on Build GRCh38
rs1033471704 Chr.14: 24318303-24318303 on Build GRCh38
rs766815522 Chr.14: 24317928-24317928 on Build GRCh38
rs752161207 Chr.14: 24318004-24318004 on Build GRCh38
rs529949593 Chr.14: 24317903-24317903 on Build GRCh38
rs1005807057 Chr.14: 24318222-24318222 on Build GRCh38
rs914911190 Chr.14: 24318111-24318111 on Build GRCh38
rs1011091880 Chr.14: 24317886-24317886 on Build GRCh38
rs753263856 Chr.14: 24318269-24318269 on Build GRCh38
rs567471723 Chr.14: 24318155-24318155 on Build GRCh38
rs763677022 Chr.14: 24318051-24318051 on Build GRCh38
rs540567448 Chr.14: 24317925-24317925 on Build GRCh38
rs780459896 Chr.14: 24318219-24318222 on Build GRCh38
rs890078453 Chr.14: 24318212-24318212 on Build GRCh38
rs538653543 Chr.14: 24318277-24318277 on Build GRCh38
rs372352454 Chr.14: 24318095-24318095 on Build GRCh38
rs983960720 Chr.14: 24317957-24317957 on Build GRCh38

View Full Product Details