Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24427562-24428091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLN3
Entrez Gene ID
643866
Gene Name
cerebellin 3 precursor
Gene Aliases
PRO1486
UniGene
Hs.207603
Gene Chromosome Location
Chr.14: 24426532-24429525 on Build GRCh38
Transcripts
NM_001039771.2 XM_011536590.2

Gene Symbol
KHNYN
Entrez Gene ID
23351
Gene Name
KH and NYN domain containing
Gene Aliases
KIAA0323
UniGene
Hs.713590
Gene Chromosome Location
Chr.14: 24423186-24441342 on Build GRCh38
Transcripts
NM_001039771.2 XM_011536590.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5929521 c.529G>A p.D177N Chr.14: 24427878-24427878 on Build GRCh38
3495553 c.464C>T p.A155V Chr.14: 24427943-24427943 on Build GRCh38
5825261 c.486G>A p.R162R Chr.14: 24427921-24427921 on Build GRCh38
3369952 c.523C>T p.P175S Chr.14: 24427884-24427884 on Build GRCh38
4401036 c.497C>T p.T166I Chr.14: 24427910-24427910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761611188 Chr.14: 24427927-24427927 on Build GRCh38
rs753295530 Chr.14: 24427978-24427978 on Build GRCh38
rs748180106 Chr.14: 24428008-24428008 on Build GRCh38
rs189882134 Chr.14: 24427685-24427685 on Build GRCh38
rs773073895 Chr.14: 24428012-24428012 on Build GRCh38
rs753104573 Chr.14: 24427875-24427875 on Build GRCh38
rs201500795 Chr.14: 24427853-24427853 on Build GRCh38
rs139294047 Chr.14: 24427936-24427936 on Build GRCh38
rs757768855 Chr.14: 24427967-24427968 on Build GRCh38
rs779962718 Chr.14: 24428002-24428002 on Build GRCh38
rs761676366 Chr.14: 24427856-24427856 on Build GRCh38
rs965221257 Chr.14: 24428037-24428037 on Build GRCh38
rs1056554586 Chr.14: 24427588-24427588 on Build GRCh38
rs753665053 Chr.14: 24427743-24427743 on Build GRCh38
rs754244519 Chr.14: 24427988-24427988 on Build GRCh38
rs778349840 Chr.14: 24427984-24427984 on Build GRCh38
rs988288490 Chr.14: 24427834-24427834 on Build GRCh38
rs747551102 Chr.14: 24427770-24427770 on Build GRCh38
rs773513524 Chr.14: 24427794-24427794 on Build GRCh38
rs370461144 Chr.14: 24427806-24427806 on Build GRCh38
rs755502123 Chr.14: 24427772-24427772 on Build GRCh38
rs773017319 Chr.14: 24427788-24427788 on Build GRCh38
rs184915473 Chr.14: 24427930-24427930 on Build GRCh38
rs758112861 Chr.14: 24427723-24427723 on Build GRCh38
rs908701081 Chr.14: 24427592-24427592 on Build GRCh38
rs772214814 Chr.14: 24428037-24428037 on Build GRCh38
rs756921301 Chr.14: 24427748-24427748 on Build GRCh38
rs758011354 Chr.14: 24427858-24427858 on Build GRCh38
rs757640049 Chr.14: 24427992-24427992 on Build GRCh38
rs958295611 Chr.14: 24427825-24427826 on Build GRCh38
rs980706749 Chr.14: 24427680-24427680 on Build GRCh38
rs573834366 Chr.14: 24427635-24427635 on Build GRCh38
rs562291434 Chr.14: 24427857-24427857 on Build GRCh38
rs779475393 Chr.14: 24427986-24427986 on Build GRCh38
rs756547814 Chr.14: 24427981-24427981 on Build GRCh38
rs776665719 Chr.14: 24427925-24427925 on Build GRCh38
rs913752619 Chr.14: 24427688-24427688 on Build GRCh38
rs774014147 Chr.14: 24428038-24428038 on Build GRCh38
rs976460992 Chr.14: 24428044-24428044 on Build GRCh38
rs762739904 Chr.14: 24428024-24428024 on Build GRCh38
rs184552844 Chr.14: 24427683-24427683 on Build GRCh38
rs10140312 Chr.14: 24428000-24428000 on Build GRCh38
rs778261582 Chr.14: 24427884-24427884 on Build GRCh38
rs202139109 Chr.14: 24427967-24427967 on Build GRCh38
rs542692779 Chr.14: 24427649-24427649 on Build GRCh38
rs912958391 Chr.14: 24427636-24427636 on Build GRCh38
rs746298684 Chr.14: 24428008-24428009 on Build GRCh38
rs749636641 Chr.14: 24427964-24427965 on Build GRCh38
rs1045371825 Chr.14: 24427662-24427662 on Build GRCh38
rs764958474 Chr.14: 24427751-24427751 on Build GRCh38
rs7154785 Chr.14: 24427624-24427624 on Build GRCh38
rs1041137362 Chr.14: 24428039-24428039 on Build GRCh38
rs757939582 Chr.14: 24427758-24427758 on Build GRCh38
rs759404862 Chr.14: 24427959-24427959 on Build GRCh38
rs577973641 Chr.14: 24427610-24427610 on Build GRCh38
rs36102398 Chr.14: 24427598-24427599 on Build GRCh38
rs771274975 Chr.14: 24427903-24427903 on Build GRCh38
rs769985392 Chr.14: 24427793-24427793 on Build GRCh38
rs781464415 Chr.14: 24427787-24427787 on Build GRCh38
rs1032567101 Chr.14: 24427823-24427823 on Build GRCh38
rs779285675 Chr.14: 24427993-24427993 on Build GRCh38
rs146286325 Chr.14: 24427862-24427862 on Build GRCh38
rs745976316 Chr.14: 24427921-24427921 on Build GRCh38
rs759115311 Chr.14: 24427966-24427966 on Build GRCh38
rs920924970 Chr.14: 24427733-24427733 on Build GRCh38
rs557444186 Chr.14: 24427631-24427631 on Build GRCh38
rs774542820 Chr.14: 24427810-24427810 on Build GRCh38
rs144101051 Chr.14: 24427951-24427951 on Build GRCh38
rs945790145 Chr.14: 24427645-24427645 on Build GRCh38
rs192944889 Chr.14: 24427789-24427789 on Build GRCh38
rs905850443 Chr.14: 24427922-24427922 on Build GRCh38
rs769666640 Chr.14: 24427929-24427929 on Build GRCh38
rs765937823 Chr.14: 24427861-24427861 on Build GRCh38
rs766031762 Chr.14: 24427940-24427940 on Build GRCh38
rs760763185 Chr.14: 24427816-24427816 on Build GRCh38
rs1038617059 Chr.14: 24427606-24427606 on Build GRCh38
rs988359090 Chr.14: 24427686-24427686 on Build GRCh38
rs751144349 Chr.14: 24427944-24427944 on Build GRCh38
rs746041135 Chr.14: 24427999-24427999 on Build GRCh38
rs754387052 Chr.14: 24427870-24427870 on Build GRCh38
rs199940555 Chr.14: 24427909-24427909 on Build GRCh38
rs147712930 Chr.14: 24427587-24427587 on Build GRCh38
rs770670571 Chr.14: 24428034-24428034 on Build GRCh38
rs781723932 Chr.14: 24427874-24427874 on Build GRCh38
rs559568517 Chr.14: 24427660-24427660 on Build GRCh38
rs779513102 Chr.14: 24427760-24427760 on Build GRCh38
rs887163875 Chr.14: 24427757-24427757 on Build GRCh38
rs771981227 Chr.14: 24427923-24427923 on Build GRCh38
rs750014293 Chr.14: 24427752-24427752 on Build GRCh38
rs943758386 Chr.14: 24427702-24427702 on Build GRCh38
rs940235633 Chr.14: 24427596-24427596 on Build GRCh38
rs771185018 Chr.14: 24427807-24427807 on Build GRCh38
rs921039404 Chr.14: 24428052-24428052 on Build GRCh38
rs12898182 Chr.14: 24427976-24427976 on Build GRCh38
rs769703763 Chr.14: 24428009-24428009 on Build GRCh38
rs749562457 Chr.14: 24427885-24427885 on Build GRCh38

View Full Product Details