Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24428677-24429176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLN3
Entrez Gene ID
643866
Gene Name
cerebellin 3 precursor
Gene Aliases
PRO1486
UniGene
Hs.207603
Gene Chromosome Location
Chr.14: 24426532-24429525 on Build GRCh38
Transcripts
NM_001039771.2 XM_011536590.2

Gene Symbol
KHNYN
Entrez Gene ID
23351
Gene Name
KH and NYN domain containing
Gene Aliases
KIAA0323
UniGene
Hs.713590
Gene Chromosome Location
Chr.14: 24423186-24441342 on Build GRCh38
Transcripts
NM_001039771.2 XM_011536590.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4519829 c.102G>A p.E34E Chr.14: 24428953-24428953 on Build GRCh38
1180806 c.182delG p.G61fs*43 Chr.14: 24428873-24428873 on Build GRCh38
4507211 c.73C>T p.L25L Chr.14: 24428982-24428982 on Build GRCh38
7429720 c.235A>T p.S79C Chr.14: 24428820-24428820 on Build GRCh38
698061 c.173G>T p.G58V Chr.14: 24428882-24428882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758329838 Chr.14: 24428777-24428777 on Build GRCh38
rs780458725 Chr.14: 24428940-24428940 on Build GRCh38
rs932013420 Chr.14: 24429112-24429112 on Build GRCh38
rs762804814 Chr.14: 24428709-24428710 on Build GRCh38
rs915070331 Chr.14: 24429148-24429148 on Build GRCh38
rs201024767 Chr.14: 24428760-24428760 on Build GRCh38
rs764625082 Chr.14: 24428986-24428986 on Build GRCh38
rs772372835 Chr.14: 24428822-24428822 on Build GRCh38
rs777316818 Chr.14: 24428793-24428793 on Build GRCh38
rs766436191 Chr.14: 24428839-24428839 on Build GRCh38
rs994941384 Chr.14: 24428968-24428968 on Build GRCh38
rs768914584 Chr.14: 24428878-24428878 on Build GRCh38
rs58379089 Chr.14: 24429096-24429096 on Build GRCh38
rs555986746 Chr.14: 24428828-24428828 on Build GRCh38
rs889687447 Chr.14: 24429137-24429137 on Build GRCh38
rs993928821 Chr.14: 24428951-24428951 on Build GRCh38
rs936216031 Chr.14: 24428818-24428818 on Build GRCh38
rs762502274 Chr.14: 24429014-24429014 on Build GRCh38
rs755779556 Chr.14: 24428855-24428855 on Build GRCh38
rs752533087 Chr.14: 24428897-24428897 on Build GRCh38
rs965025431 Chr.14: 24429038-24429038 on Build GRCh38
rs371680375 Chr.14: 24428941-24428941 on Build GRCh38
rs774362547 Chr.14: 24428917-24428918 on Build GRCh38
rs965452361 Chr.14: 24428848-24428848 on Build GRCh38
rs768857696 Chr.14: 24428950-24428950 on Build GRCh38
rs748289588 Chr.14: 24428970-24428970 on Build GRCh38
rs764191353 Chr.14: 24428770-24428770 on Build GRCh38
rs748834442 Chr.14: 24428865-24428865 on Build GRCh38
rs376939942 Chr.14: 24428844-24428844 on Build GRCh38
rs752437042 Chr.14: 24429114-24429114 on Build GRCh38
rs750363924 Chr.14: 24428772-24428772 on Build GRCh38
rs776663495 Chr.14: 24428879-24428879 on Build GRCh38
rs767729762 Chr.14: 24428761-24428761 on Build GRCh38
rs779766705 Chr.14: 24428781-24428781 on Build GRCh38
rs775233007 Chr.14: 24429019-24429019 on Build GRCh38
rs1049700957 Chr.14: 24428896-24428896 on Build GRCh38
rs771464590 Chr.14: 24428801-24428801 on Build GRCh38
rs751409484 Chr.14: 24428782-24428782 on Build GRCh38
rs73593269 Chr.14: 24428742-24428742 on Build GRCh38
rs773414234 Chr.14: 24428985-24428985 on Build GRCh38
rs998018200 Chr.14: 24428890-24428890 on Build GRCh38
rs548072166 Chr.14: 24428926-24428926 on Build GRCh38
rs375714567 Chr.14: 24429006-24429006 on Build GRCh38
rs989682311 Chr.14: 24429134-24429134 on Build GRCh38
rs780929171 Chr.14: 24428715-24428715 on Build GRCh38
rs759907388 Chr.14: 24428807-24428807 on Build GRCh38
rs756790878 Chr.14: 24428924-24428924 on Build GRCh38
rs10143703 Chr.14: 24429149-24429149 on Build GRCh38
rs971092498 Chr.14: 24428966-24428966 on Build GRCh38
rs117443441 Chr.14: 24428735-24428735 on Build GRCh38
rs1023939619 Chr.14: 24428960-24428960 on Build GRCh38
rs749818641 Chr.14: 24428795-24428795 on Build GRCh38
rs755798787 Chr.14: 24428898-24428898 on Build GRCh38
rs1042898422 Chr.14: 24428927-24428927 on Build GRCh38
rs1050090560 Chr.14: 24429129-24429129 on Build GRCh38
rs1003740892 Chr.14: 24428729-24428729 on Build GRCh38
rs145699313 Chr.14: 24428843-24428843 on Build GRCh38
rs112261621 Chr.14: 24429111-24429111 on Build GRCh38
rs766022566 Chr.14: 24428739-24428739 on Build GRCh38
rs764124635 Chr.14: 24428824-24428824 on Build GRCh38
rs746332705 Chr.14: 24428874-24428874 on Build GRCh38
rs368998125 Chr.14: 24429007-24429007 on Build GRCh38
rs189368065 Chr.14: 24428732-24428732 on Build GRCh38
rs758885623 Chr.14: 24428935-24428935 on Build GRCh38
rs775718996 Chr.14: 24428823-24428823 on Build GRCh38
rs181787327 Chr.14: 24429138-24429138 on Build GRCh38
rs866254952 Chr.14: 24428812-24428812 on Build GRCh38
rs952513048 Chr.14: 24428827-24428827 on Build GRCh38
rs1015800215 Chr.14: 24429036-24429036 on Build GRCh38
rs754765912 Chr.14: 24428786-24428786 on Build GRCh38
rs763755426 Chr.14: 24429048-24429048 on Build GRCh38
rs761978741 Chr.14: 24428881-24428881 on Build GRCh38
rs760539880 Chr.14: 24429034-24429034 on Build GRCh38
rs772464184 Chr.14: 24428876-24428876 on Build GRCh38
rs767480433 Chr.14: 24428885-24428885 on Build GRCh38
rs781017023 Chr.14: 24428788-24428788 on Build GRCh38
rs200467816 Chr.14: 24428877-24428877 on Build GRCh38
rs776946776 Chr.14: 24428967-24428967 on Build GRCh38
rs779417158 Chr.14: 24428873-24428873 on Build GRCh38
rs199897938 Chr.14: 24428790-24428790 on Build GRCh38
rs753382708 Chr.14: 24428919-24428919 on Build GRCh38
rs372087794 Chr.14: 24428838-24428838 on Build GRCh38
rs375332763 Chr.14: 24428882-24428882 on Build GRCh38
rs774773920 Chr.14: 24428802-24428802 on Build GRCh38
rs946100594 Chr.14: 24429095-24429095 on Build GRCh38
rs553070221 Chr.14: 24428976-24428976 on Build GRCh38
rs200916795 Chr.14: 24428767-24428767 on Build GRCh38
rs763820466 Chr.14: 24428899-24428899 on Build GRCh38
rs768423432 Chr.14: 24428714-24428714 on Build GRCh38
rs760690740 Chr.14: 24428768-24428768 on Build GRCh38
rs777215649 Chr.14: 24428856-24428856 on Build GRCh38
rs776754412 Chr.14: 24428830-24428830 on Build GRCh38
rs747678229 Chr.14: 24428724-24428724 on Build GRCh38
rs759346972 Chr.14: 24428970-24428970 on Build GRCh38
rs756663678 Chr.14: 24428870-24428870 on Build GRCh38
rs769229753 Chr.14: 24428791-24428791 on Build GRCh38
rs771251138 Chr.14: 24428744-24428744 on Build GRCh38
rs57646536 Chr.14: 24428929-24428929 on Build GRCh38

View Full Product Details