Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 44505198-44505712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSCB
Entrez Gene ID
84075
Gene Name
fibrous sheath CABYR binding protein
Gene Aliases
C14orf155
UniGene
Hs.307086
Gene Chromosome Location
Chr.14: 44504149-44507296 on Build GRCh38
Transcripts
NM_032135.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
955729 c.1680G>C p.Q560H Chr.14: 44505308-44505308 on Build GRCh38
4878663 c.1771G>A p.A591T Chr.14: 44505217-44505217 on Build GRCh38
955731 c.1511C>A p.A504D Chr.14: 44505477-44505477 on Build GRCh38
955733 c.1405G>T p.E469* Chr.14: 44505583-44505583 on Build GRCh38
6040633 c.1657G>A p.E553K Chr.14: 44505331-44505331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867459431 Chr.14: 44505396-44505396 on Build GRCh38
rs765495778 Chr.14: 44505364-44505364 on Build GRCh38
rs781774468 Chr.14: 44505506-44505506 on Build GRCh38
rs17115207 Chr.14: 44505548-44505548 on Build GRCh38
rs553066717 Chr.14: 44505601-44505601 on Build GRCh38
rs1022302036 Chr.14: 44505478-44505478 on Build GRCh38
rs748570146 Chr.14: 44505403-44505403 on Build GRCh38
rs779908422 Chr.14: 44505604-44505604 on Build GRCh38
rs199888788 Chr.14: 44505428-44505428 on Build GRCh38
rs746758057 Chr.14: 44505543-44505543 on Build GRCh38
rs756477383 Chr.14: 44505579-44505579 on Build GRCh38
rs370240065 Chr.14: 44505649-44505649 on Build GRCh38
rs758919849 Chr.14: 44505283-44505283 on Build GRCh38
rs987644431 Chr.14: 44505344-44505344 on Build GRCh38
rs757675485 Chr.14: 44505651-44505651 on Build GRCh38
rs758340039 Chr.14: 44505675-44505676 on Build GRCh38
rs758440278 Chr.14: 44505370-44505370 on Build GRCh38
rs766374546 Chr.14: 44505494-44505494 on Build GRCh38
rs775775877 Chr.14: 44505645-44505645 on Build GRCh38
rs374042910 Chr.14: 44505427-44505427 on Build GRCh38
rs749821330 Chr.14: 44505522-44505522 on Build GRCh38
rs750632970 Chr.14: 44505492-44505492 on Build GRCh38
rs754757464 Chr.14: 44505252-44505252 on Build GRCh38
rs779535850 Chr.14: 44505670-44505670 on Build GRCh38
rs780562129 Chr.14: 44505375-44505375 on Build GRCh38
rs371199022 Chr.14: 44505514-44505514 on Build GRCh38
rs1050277793 Chr.14: 44505587-44505587 on Build GRCh38
rs768648392 Chr.14: 44505613-44505613 on Build GRCh38
rs765403690 Chr.14: 44505475-44505475 on Build GRCh38
rs777596508 Chr.14: 44505408-44505408 on Build GRCh38
rs374248988 Chr.14: 44505545-44505545 on Build GRCh38
rs370149746 Chr.14: 44505361-44505361 on Build GRCh38
rs780467660 Chr.14: 44505289-44505289 on Build GRCh38
rs748012817 Chr.14: 44505622-44505622 on Build GRCh38
rs764242311 Chr.14: 44505494-44505496 on Build GRCh38
rs753067531 Chr.14: 44505510-44505510 on Build GRCh38
rs946071591 Chr.14: 44505681-44505681 on Build GRCh38
rs759313834 Chr.14: 44505352-44505352 on Build GRCh38
rs569048873 Chr.14: 44505322-44505322 on Build GRCh38
rs142350780 Chr.14: 44505591-44505591 on Build GRCh38
rs759719884 Chr.14: 44505563-44505563 on Build GRCh38
rs776266363 Chr.14: 44505620-44505620 on Build GRCh38
rs750472129 Chr.14: 44505368-44505368 on Build GRCh38
rs961681691 Chr.14: 44505588-44505588 on Build GRCh38
rs764749139 Chr.14: 44505655-44505655 on Build GRCh38
rs747847580 Chr.14: 44505677-44505677 on Build GRCh38
rs754069503 Chr.14: 44505263-44505263 on Build GRCh38
rs147426737 Chr.14: 44505433-44505433 on Build GRCh38
rs751717732 Chr.14: 44505501-44505501 on Build GRCh38
rs768084268 Chr.14: 44505576-44505576 on Build GRCh38
rs201205110 Chr.14: 44505654-44505654 on Build GRCh38
rs752021418 Chr.14: 44505391-44505391 on Build GRCh38
rs748781019 Chr.14: 44505426-44505426 on Build GRCh38
rs199726278 Chr.14: 44505660-44505660 on Build GRCh38
rs766274427 Chr.14: 44505243-44505243 on Build GRCh38
rs767224148 Chr.14: 44505360-44505360 on Build GRCh38
rs376194119 Chr.14: 44505632-44505632 on Build GRCh38
rs770859334 Chr.14: 44505334-44505334 on Build GRCh38
rs766671624 Chr.14: 44505552-44505552 on Build GRCh38
rs778963490 Chr.14: 44505273-44505273 on Build GRCh38
rs755574099 Chr.14: 44505395-44505395 on Build GRCh38
rs764546354 Chr.14: 44505580-44505580 on Build GRCh38
rs931818088 Chr.14: 44505251-44505251 on Build GRCh38
rs920547359 Chr.14: 44505402-44505402 on Build GRCh38
rs369651441 Chr.14: 44505526-44505526 on Build GRCh38
rs776365925 Chr.14: 44505313-44505313 on Build GRCh38
rs550825439 Chr.14: 44505266-44505266 on Build GRCh38
rs866751776 Chr.14: 44505513-44505513 on Build GRCh38
rs867972087 Chr.14: 44505354-44505354 on Build GRCh38
rs139795665 Chr.14: 44505455-44505455 on Build GRCh38
rs879007271 Chr.14: 44505474-44505474 on Build GRCh38
rs973274203 Chr.14: 44505392-44505392 on Build GRCh38
rs775438824 Chr.14: 44505542-44505542 on Build GRCh38
rs753265665 Chr.14: 44505577-44505577 on Build GRCh38
rs747361133 Chr.14: 44505308-44505308 on Build GRCh38
rs535022618 Chr.14: 44505596-44505596 on Build GRCh38
rs751335150 Chr.14: 44505246-44505246 on Build GRCh38
rs768925200 Chr.14: 44505310-44505310 on Build GRCh38
rs748894618 Chr.14: 44505521-44505521 on Build GRCh38
rs201456569 Chr.14: 44505547-44505547 on Build GRCh38
rs376664572 Chr.14: 44505520-44505520 on Build GRCh38
rs762406599 Chr.14: 44505328-44505328 on Build GRCh38
rs891809017 Chr.14: 44505435-44505435 on Build GRCh38
rs781679494 Chr.14: 44505400-44505400 on Build GRCh38
rs775246299 Chr.14: 44505460-44505460 on Build GRCh38
rs149992888 Chr.14: 44505682-44505682 on Build GRCh38
rs750273533 Chr.14: 44505514-44505514 on Build GRCh38
rs35809445 Chr.14: 44505676-44505676 on Build GRCh38
rs201606526 Chr.14: 44505665-44505665 on Build GRCh38
rs771018416 Chr.14: 44505625-44505625 on Build GRCh38
rs763071301 Chr.14: 44505549-44505549 on Build GRCh38
rs968905222 Chr.14: 44505486-44505486 on Build GRCh38
rs560432740 Chr.14: 44505675-44505675 on Build GRCh38
rs746007576 Chr.14: 44505282-44505282 on Build GRCh38
rs747797098 Chr.14: 44505321-44505321 on Build GRCh38
rs759627915 Chr.14: 44505643-44505643 on Build GRCh38
rs868307119 Chr.14: 44505532-44505532 on Build GRCh38
rs145232784 Chr.14: 44505471-44505471 on Build GRCh38
rs774263359 Chr.14: 44505553-44505553 on Build GRCh38
rs151240998 Chr.14: 44505644-44505644 on Build GRCh38
rs774173349 Chr.14: 44505350-44505350 on Build GRCh38
rs771970244 Chr.14: 44505529-44505529 on Build GRCh38
rs755416277 Chr.14: 44505505-44505505 on Build GRCh38
rs866560729 Chr.14: 44505453-44505453 on Build GRCh38

View Full Product Details