Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 49625091-49625597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAAF2
Entrez Gene ID
55172
Gene Name
dynein axonemal assembly factor 2
Gene Aliases
C14orf104, CILD10, KTU, PF13
UniGene
Hs.231761
Gene Chromosome Location
Chr.14: 49625174-49635230 on Build GRCh38
Transcripts
NM_001083908.1 NM_018139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7146145 c.972A>T p.L324L Chr.14: 49625557-49625557 on Build GRCh38
7146143 c.2499A>T p.L833L Chr.14: 49625557-49625557 on Build GRCh38
4414859 c.2478C>A p.F826L Chr.14: 49625578-49625578 on Build GRCh38
7273147 c.2483T>G p.F828C Chr.14: 49625573-49625573 on Build GRCh38
4414860 c.951C>A p.F317L Chr.14: 49625578-49625578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561374548 Chr.14: 49625453-49625453 on Build GRCh38
rs1022298479 Chr.14: 49625157-49625157 on Build GRCh38
rs372666364 Chr.14: 49625415-49625415 on Build GRCh38
rs772368193 Chr.14: 49625501-49625501 on Build GRCh38
rs927687588 Chr.14: 49625405-49625405 on Build GRCh38
rs371013573 Chr.14: 49625542-49625542 on Build GRCh38
rs962199981 Chr.14: 49625252-49625252 on Build GRCh38
rs79975294 Chr.14: 49625415-49625416 on Build GRCh38
rs571401882 Chr.14: 49625417-49625417 on Build GRCh38
rs749123028 Chr.14: 49625499-49625499 on Build GRCh38
rs751218274 Chr.14: 49625569-49625569 on Build GRCh38
rs965211982 Chr.14: 49625223-49625223 on Build GRCh38
rs936675864 Chr.14: 49625337-49625337 on Build GRCh38
rs886050524 Chr.14: 49625248-49625252 on Build GRCh38
rs541064529 Chr.14: 49625215-49625215 on Build GRCh38
rs939054850 Chr.14: 49625406-49625406 on Build GRCh38
rs921071231 Chr.14: 49625266-49625266 on Build GRCh38
rs1033235488 Chr.14: 49625254-49625269 on Build GRCh38
rs765290990 Chr.14: 49625511-49625517 on Build GRCh38
rs368195964 Chr.14: 49625553-49625553 on Build GRCh38
rs951735674 Chr.14: 49625461-49625461 on Build GRCh38
rs971195879 Chr.14: 49625151-49625151 on Build GRCh38
rs777482756 Chr.14: 49625497-49625497 on Build GRCh38
rs542798692 Chr.14: 49625166-49625166 on Build GRCh38
rs939666787 Chr.14: 49625342-49625342 on Build GRCh38
rs8952 Chr.14: 49625416-49625416 on Build GRCh38
rs1055814090 Chr.14: 49625123-49625123 on Build GRCh38
rs932055061 Chr.14: 49625268-49625268 on Build GRCh38
rs777335572 Chr.14: 49625245-49625245 on Build GRCh38
rs886050525 Chr.14: 49625351-49625351 on Build GRCh38
rs967105356 Chr.14: 49625201-49625201 on Build GRCh38
rs187830950 Chr.14: 49625458-49625458 on Build GRCh38
rs909585414 Chr.14: 49625320-49625320 on Build GRCh38
rs12160 Chr.14: 49625214-49625214 on Build GRCh38
rs999603420 Chr.14: 49625203-49625203 on Build GRCh38
rs765604726 Chr.14: 49625547-49625547 on Build GRCh38
rs201753356 Chr.14: 49625409-49625410 on Build GRCh38
rs546757721 Chr.14: 49625463-49625463 on Build GRCh38
rs1044364957 Chr.14: 49625443-49625443 on Build GRCh38
rs528698497 Chr.14: 49625537-49625537 on Build GRCh38
rs770435695 Chr.14: 49625521-49625521 on Build GRCh38
rs765960374 Chr.14: 49625568-49625568 on Build GRCh38
rs564891226 Chr.14: 49625330-49625330 on Build GRCh38
rs572958055 Chr.14: 49625256-49625256 on Build GRCh38
rs895413084 Chr.14: 49625139-49625139 on Build GRCh38
rs1001597178 Chr.14: 49625165-49625165 on Build GRCh38
rs539721977 Chr.14: 49625152-49625152 on Build GRCh38
rs545151034 Chr.14: 49625297-49625297 on Build GRCh38
rs953988451 Chr.14: 49625269-49625269 on Build GRCh38
rs530460999 Chr.14: 49625373-49625373 on Build GRCh38
rs774072576 Chr.14: 49625566-49625566 on Build GRCh38
rs984408431 Chr.14: 49625272-49625272 on Build GRCh38
rs1036808221 Chr.14: 49625395-49625395 on Build GRCh38
rs925268737 Chr.14: 49625311-49625311 on Build GRCh38
rs192899037 Chr.14: 49625534-49625534 on Build GRCh38
rs1003697494 Chr.14: 49625118-49625118 on Build GRCh38
rs879160187 Chr.14: 49625360-49625360 on Build GRCh38
rs879075266 Chr.14: 49625359-49625360 on Build GRCh38
rs757361238 Chr.14: 49625181-49625181 on Build GRCh38
rs566758458 Chr.14: 49625525-49625525 on Build GRCh38
rs1010007979 Chr.14: 49625385-49625385 on Build GRCh38

View Full Product Details