Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54427258-54427771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNIH1
Entrez Gene ID
10175
Gene Name
cornichon family AMPA receptor auxiliary protein 1
Gene Aliases
CNIH, CNIH-1, CNIL, TGAM77
UniGene
Hs.294603
Gene Chromosome Location
Chr.14: 54426929-54441430 on Build GRCh38
Transcripts
NM_005776.2

SNP Detail

SNP ID Chromosome Location
rs887117272 Chr.14: 54427311-54427311 on Build GRCh38
rs10220437 Chr.14: 54427578-54427578 on Build GRCh38
rs551639240 Chr.14: 54427547-54427547 on Build GRCh38
rs896268740 Chr.14: 54427450-54427450 on Build GRCh38
rs1004189783 Chr.14: 54427332-54427332 on Build GRCh38
rs746790873 Chr.14: 54427438-54427438 on Build GRCh38
rs191170776 Chr.14: 54427487-54427487 on Build GRCh38
rs1055584463 Chr.14: 54427683-54427683 on Build GRCh38
rs898492184 Chr.14: 54427628-54427628 on Build GRCh38
rs1002687013 Chr.14: 54427419-54427419 on Build GRCh38
rs934301915 Chr.14: 54427289-54427289 on Build GRCh38
rs111490953 Chr.14: 54427558-54427558 on Build GRCh38
rs779887686 Chr.14: 54427412-54427412 on Build GRCh38
rs904309257 Chr.14: 54427326-54427326 on Build GRCh38
rs1026231578 Chr.14: 54427453-54427453 on Build GRCh38
rs560451364 Chr.14: 54427552-54427552 on Build GRCh38
rs1013510371 Chr.14: 54427458-54427458 on Build GRCh38
rs562162583 Chr.14: 54427401-54427401 on Build GRCh38
rs79890403 Chr.14: 54427531-54427531 on Build GRCh38
rs776229022 Chr.14: 54427608-54427608 on Build GRCh38
rs970893250 Chr.14: 54427429-54427430 on Build GRCh38
rs540474402 Chr.14: 54427394-54427394 on Build GRCh38
rs74707690 Chr.14: 54427339-54427339 on Build GRCh38
rs372903257 Chr.14: 54427354-54427354 on Build GRCh38
rs1034046423 Chr.14: 54427336-54427336 on Build GRCh38
rs984599831 Chr.14: 54427473-54427473 on Build GRCh38
rs749273065 Chr.14: 54427296-54427296 on Build GRCh38
rs868680965 Chr.14: 54427590-54427590 on Build GRCh38
rs1021262363 Chr.14: 54427465-54427465 on Build GRCh38
rs1000343156 Chr.14: 54427329-54427329 on Build GRCh38
rs574055625 Chr.14: 54427407-54427407 on Build GRCh38
rs184763336 Chr.14: 54427505-54427505 on Build GRCh38
rs8016671 Chr.14: 54427663-54427663 on Build GRCh38
rs1051746498 Chr.14: 54427291-54427291 on Build GRCh38
rs550521723 Chr.14: 54427726-54427726 on Build GRCh38
rs865970633 Chr.14: 54427623-54427623 on Build GRCh38
rs942682404 Chr.14: 54427493-54427496 on Build GRCh38
rs987086732 Chr.14: 54427599-54427599 on Build GRCh38
rs966366517 Chr.14: 54427497-54427497 on Build GRCh38
rs539372402 Chr.14: 54427304-54427304 on Build GRCh38
rs192912423 Chr.14: 54427619-54427619 on Build GRCh38
rs563962848 Chr.14: 54427598-54427598 on Build GRCh38
rs952861522 Chr.14: 54427468-54427468 on Build GRCh38
rs1012520645 Chr.14: 54427347-54427347 on Build GRCh38
rs898606038 Chr.14: 54427373-54427373 on Build GRCh38
rs1013893120 Chr.14: 54427739-54427739 on Build GRCh38
rs745760205 Chr.14: 54427284-54427284 on Build GRCh38
rs186764033 Chr.14: 54427386-54427386 on Build GRCh38
rs533651432 Chr.14: 54427546-54427546 on Build GRCh38

View Full Product Details