Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54427635-54428136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNIH1
Entrez Gene ID
10175
Gene Name
cornichon family AMPA receptor auxiliary protein 1
Gene Aliases
CNIH, CNIH-1, CNIL, TGAM77
UniGene
Hs.294603
Gene Chromosome Location
Chr.14: 54426929-54441430 on Build GRCh38
Transcripts
NM_005776.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6140880 c.424G>T p.V142L Chr.14: 54427825-54427825 on Build GRCh38
1629342 c.408C>G p.G136G Chr.14: 54427841-54427841 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766844962 Chr.14: 54427849-54427849 on Build GRCh38
rs763588082 Chr.14: 54427833-54427833 on Build GRCh38
rs764526541 Chr.14: 54427886-54427887 on Build GRCh38
rs773600831 Chr.14: 54427992-54427992 on Build GRCh38
rs185470039 Chr.14: 54427891-54427891 on Build GRCh38
rs1030467536 Chr.14: 54427999-54427999 on Build GRCh38
rs752011533 Chr.14: 54427858-54427858 on Build GRCh38
rs1055584463 Chr.14: 54427683-54427683 on Build GRCh38
rs946011381 Chr.14: 54427914-54427914 on Build GRCh38
rs79100399 Chr.14: 54428064-54428064 on Build GRCh38
rs769856364 Chr.14: 54427779-54427779 on Build GRCh38
rs974320019 Chr.14: 54428065-54428065 on Build GRCh38
rs11552807 Chr.14: 54427768-54427768 on Build GRCh38
rs375233739 Chr.14: 54427862-54427862 on Build GRCh38
rs768130926 Chr.14: 54427847-54427849 on Build GRCh38
rs759443436 Chr.14: 54427812-54427812 on Build GRCh38
rs372789436 Chr.14: 54427840-54427840 on Build GRCh38
rs111790713 Chr.14: 54427878-54427878 on Build GRCh38
rs902465607 Chr.14: 54427888-54427888 on Build GRCh38
rs566820073 Chr.14: 54427927-54427927 on Build GRCh38
rs144305125 Chr.14: 54428070-54428070 on Build GRCh38
rs755517979 Chr.14: 54427859-54427859 on Build GRCh38
rs533928658 Chr.14: 54427996-54427996 on Build GRCh38
rs752675694 Chr.14: 54427861-54427861 on Build GRCh38
rs189946266 Chr.14: 54428045-54428045 on Build GRCh38
rs949778630 Chr.14: 54428097-54428097 on Build GRCh38
rs573223293 Chr.14: 54427887-54427887 on Build GRCh38
rs201278380 Chr.14: 54427796-54427796 on Build GRCh38
rs774202383 Chr.14: 54427808-54427808 on Build GRCh38
rs375313617 Chr.14: 54427766-54427766 on Build GRCh38
rs531991338 Chr.14: 54428012-54428012 on Build GRCh38
rs370541376 Chr.14: 54427993-54427994 on Build GRCh38
rs560365619 Chr.14: 54428077-54428077 on Build GRCh38
rs555610505 Chr.14: 54428003-54428003 on Build GRCh38
rs8016671 Chr.14: 54427663-54427663 on Build GRCh38
rs371629087 Chr.14: 54427887-54427887 on Build GRCh38
rs919932072 Chr.14: 54428069-54428069 on Build GRCh38
rs78211993 Chr.14: 54427896-54427896 on Build GRCh38
rs952974225 Chr.14: 54427855-54427855 on Build GRCh38
rs550521723 Chr.14: 54427726-54427726 on Build GRCh38
rs774906612 Chr.14: 54428105-54428105 on Build GRCh38
rs777795751 Chr.14: 54427870-54427870 on Build GRCh38
rs776193445 Chr.14: 54427852-54427853 on Build GRCh38
rs774935586 Chr.14: 54427821-54427821 on Build GRCh38
rs370026434 Chr.14: 54427820-54427820 on Build GRCh38
rs3783635 Chr.14: 54428074-54428074 on Build GRCh38
rs78339426 Chr.14: 54427895-54427895 on Build GRCh38
rs558074460 Chr.14: 54427912-54427912 on Build GRCh38
rs375738110 Chr.14: 54427803-54427803 on Build GRCh38
rs181382920 Chr.14: 54428075-54428075 on Build GRCh38
rs987398847 Chr.14: 54428018-54428018 on Build GRCh38
rs1013893120 Chr.14: 54427739-54427739 on Build GRCh38
rs367907346 Chr.14: 54427801-54427801 on Build GRCh38
rs760264050 Chr.14: 54427826-54427826 on Build GRCh38

View Full Product Details