Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54436203-54436700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNIH1
Entrez Gene ID
10175
Gene Name
cornichon family AMPA receptor auxiliary protein 1
Gene Aliases
CNIH, CNIH-1, CNIL, TGAM77
UniGene
Hs.294603
Gene Chromosome Location
Chr.14: 54426929-54441430 on Build GRCh38
Transcripts
NM_005776.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
459152 c.97G>A p.E33K Chr.14: 54436422-54436422 on Build GRCh38
956297 c.117G>T p.K39N Chr.14: 54436402-54436402 on Build GRCh38
7361703 c.124A>G p.I42V Chr.14: 54436395-54436395 on Build GRCh38
5414490 c.94G>A p.D32N Chr.14: 54436425-54436425 on Build GRCh38
6900678 c.82-1G>A p.? Chr.14: 54436438-54436438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971734200 Chr.14: 54436440-54436440 on Build GRCh38
rs568342357 Chr.14: 54436276-54436276 on Build GRCh38
rs192182056 Chr.14: 54436637-54436637 on Build GRCh38
rs916744867 Chr.14: 54436543-54436543 on Build GRCh38
rs769439015 Chr.14: 54436395-54436395 on Build GRCh38
rs984257195 Chr.14: 54436500-54436500 on Build GRCh38
rs373068766 Chr.14: 54436619-54436619 on Build GRCh38
rs779866415 Chr.14: 54436359-54436359 on Build GRCh38
rs372698284 Chr.14: 54436453-54436453 on Build GRCh38
rs948675878 Chr.14: 54436240-54436240 on Build GRCh38
rs754893254 Chr.14: 54436369-54436369 on Build GRCh38
rs775879066 Chr.14: 54436456-54436456 on Build GRCh38
rs893887997 Chr.14: 54436674-54436674 on Build GRCh38
rs777479761 Chr.14: 54436411-54436411 on Build GRCh38
rs760979618 Chr.14: 54436467-54436467 on Build GRCh38
rs758687312 Chr.14: 54436329-54436329 on Build GRCh38
rs772190977 Chr.14: 54436548-54436548 on Build GRCh38
rs914544204 Chr.14: 54436234-54436235 on Build GRCh38
rs987380088 Chr.14: 54436354-54436354 on Build GRCh38
rs60565495 Chr.14: 54436432-54436432 on Build GRCh38
rs111637216 Chr.14: 54436498-54436498 on Build GRCh38
rs938483479 Chr.14: 54436362-54436362 on Build GRCh38
rs771219905 Chr.14: 54436368-54436369 on Build GRCh38
rs750471849 Chr.14: 54436454-54436455 on Build GRCh38
rs1031824529 Chr.14: 54436634-54436634 on Build GRCh38
rs187739848 Chr.14: 54436607-54436607 on Build GRCh38
rs375971919 Chr.14: 54436387-54436387 on Build GRCh38
rs184129419 Chr.14: 54436501-54436501 on Build GRCh38
rs900143632 Chr.14: 54436584-54436584 on Build GRCh38
rs746326789 Chr.14: 54436456-54436459 on Build GRCh38
rs537586002 Chr.14: 54436601-54436601 on Build GRCh38
rs1008439271 Chr.14: 54436631-54436631 on Build GRCh38
rs538048620 Chr.14: 54436327-54436328 on Build GRCh38
rs762781690 Chr.14: 54436252-54436252 on Build GRCh38
rs369101586 Chr.14: 54436444-54436444 on Build GRCh38
rs561558253 Chr.14: 54436287-54436287 on Build GRCh38
rs1047071953 Chr.14: 54436612-54436612 on Build GRCh38
rs116172065 Chr.14: 54436606-54436606 on Build GRCh38
rs527615412 Chr.14: 54436641-54436641 on Build GRCh38
rs576584533 Chr.14: 54436382-54436382 on Build GRCh38
rs1052416391 Chr.14: 54436669-54436669 on Build GRCh38
rs751360007 Chr.14: 54436361-54436361 on Build GRCh38
rs953317220 Chr.14: 54436342-54436342 on Build GRCh38

View Full Product Details