Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 56618461-56618999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM260
Entrez Gene ID
54916
Gene Name
transmembrane protein 260
Gene Aliases
C14orf101
UniGene
Hs.497253
Gene Chromosome Location
Chr.14: 56579791-56660629 on Build GRCh38
Transcripts
NM_017799.3 XM_005267771.1 XM_006720176.1 XM_006720178.1 XM_011536851.1 XM_011536852.1 XM_011536853.1 XM_017021379.1 XM_017021380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7417722 c.1068C>A p.F356L Chr.14: 56618605-56618605 on Build GRCh38
5568162 c.1080C>T p.S360S Chr.14: 56618617-56618617 on Build GRCh38
1607763 c.1120G>A p.A374T Chr.14: 56618657-56618657 on Build GRCh38
254579 c.1123G>A p.V375I Chr.14: 56618660-56618660 on Build GRCh38
7394286 c.1207C>G p.Q403E Chr.14: 56618744-56618744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761458932 Chr.14: 56618638-56618638 on Build GRCh38
rs748540718 Chr.14: 56618579-56618579 on Build GRCh38
rs753367404 Chr.14: 56618635-56618635 on Build GRCh38
rs564143023 Chr.14: 56618601-56618601 on Build GRCh38
rs769045869 Chr.14: 56618499-56618499 on Build GRCh38
rs758076398 Chr.14: 56618678-56618678 on Build GRCh38
rs764775633 Chr.14: 56618770-56618770 on Build GRCh38
rs1044969724 Chr.14: 56618813-56618813 on Build GRCh38
rs1002625142 Chr.14: 56618820-56618820 on Build GRCh38
rs957001555 Chr.14: 56618769-56618769 on Build GRCh38
rs770350609 Chr.14: 56618583-56618583 on Build GRCh38
rs12589660 Chr.14: 56618775-56618775 on Build GRCh38
rs1028068637 Chr.14: 56618856-56618856 on Build GRCh38
rs563621600 Chr.14: 56618511-56618511 on Build GRCh38
rs781683046 Chr.14: 56618547-56618547 on Build GRCh38
rs749814417 Chr.14: 56618720-56618720 on Build GRCh38
rs760516068 Chr.14: 56618632-56618632 on Build GRCh38
rs760558334 Chr.14: 56618754-56618754 on Build GRCh38
rs764674565 Chr.14: 56618949-56618949 on Build GRCh38
rs207474868 Chr.14: 56618679-56618679 on Build GRCh38
rs113535823 Chr.14: 56618797-56618797 on Build GRCh38
rs766125709 Chr.14: 56618801-56618801 on Build GRCh38
rs149868833 Chr.14: 56618761-56618761 on Build GRCh38
rs974999753 Chr.14: 56618570-56618570 on Build GRCh38
rs187777722 Chr.14: 56618945-56618945 on Build GRCh38
rs376276411 Chr.14: 56618762-56618762 on Build GRCh38
rs778297038 Chr.14: 56618717-56618717 on Build GRCh38
rs912422414 Chr.14: 56618925-56618925 on Build GRCh38
rs373343466 Chr.14: 56618747-56618747 on Build GRCh38
rs373475204 Chr.14: 56618870-56618870 on Build GRCh38
rs772394630 Chr.14: 56618746-56618746 on Build GRCh38
rs747635397 Chr.14: 56618585-56618585 on Build GRCh38
rs756613606 Chr.14: 56618681-56618681 on Build GRCh38
rs764193089 Chr.14: 56618633-56618633 on Build GRCh38
rs771695037 Chr.14: 56618616-56618616 on Build GRCh38
rs746018532 Chr.14: 56618730-56618730 on Build GRCh38
rs747617902 Chr.14: 56618492-56618492 on Build GRCh38
rs535125009 Chr.14: 56618940-56618940 on Build GRCh38
rs779052468 Chr.14: 56618729-56618729 on Build GRCh38
rs369130953 Chr.14: 56618548-56618548 on Build GRCh38
rs767176382 Chr.14: 56618658-56618658 on Build GRCh38
rs752977869 Chr.14: 56618802-56618802 on Build GRCh38
rs201148915 Chr.14: 56618634-56618634 on Build GRCh38
rs986636409 Chr.14: 56618918-56618918 on Build GRCh38
rs550161813 Chr.14: 56618944-56618944 on Build GRCh38
rs762502644 Chr.14: 56618787-56618787 on Build GRCh38
rs767245835 Chr.14: 56618510-56618510 on Build GRCh38
rs192620006 Chr.14: 56618948-56618948 on Build GRCh38
rs374793497 Chr.14: 56618599-56618599 on Build GRCh38
rs770954602 Chr.14: 56618725-56618725 on Build GRCh38
rs756417619 Chr.14: 56618815-56618815 on Build GRCh38
rs774504453 Chr.14: 56618600-56618600 on Build GRCh38
rs930646088 Chr.14: 56618642-56618642 on Build GRCh38
rs184872567 Chr.14: 56618951-56618951 on Build GRCh38
rs953863458 Chr.14: 56618530-56618530 on Build GRCh38
rs775367896 Chr.14: 56618620-56618620 on Build GRCh38
rs921020661 Chr.14: 56618503-56618503 on Build GRCh38
rs117677460 Chr.14: 56618542-56618542 on Build GRCh38
rs201584812 Chr.14: 56618575-56618575 on Build GRCh38
rs761875479 Chr.14: 56618767-56618767 on Build GRCh38
rs1053331388 Chr.14: 56618694-56618694 on Build GRCh38
rs7148317 Chr.14: 56618939-56618939 on Build GRCh38
rs749405692 Chr.14: 56618596-56618596 on Build GRCh38
rs755250986 Chr.14: 56618561-56618561 on Build GRCh38
rs1011661179 Chr.14: 56618855-56618855 on Build GRCh38
rs370134577 Chr.14: 56618654-56618654 on Build GRCh38
rs142847668 Chr.14: 56618771-56618771 on Build GRCh38

View Full Product Details