Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64543155-64543666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA2
Entrez Gene ID
3306
Gene Name
heat shock protein family A (Hsp70) member 2
Gene Aliases
HSP70-2, HSP70-3
UniGene
Hs.432648
Gene Chromosome Location
Chr.14: 64540468-64543237 on Build GRCh38
Transcripts
NM_021979.3

SNP Detail

SNP ID Chromosome Location
rs568582166 Chr.14: 64543385-64543385 on Build GRCh38
rs928394968 Chr.14: 64543552-64543552 on Build GRCh38
rs929532577 Chr.14: 64543499-64543499 on Build GRCh38
rs965423144 Chr.14: 64543343-64543344 on Build GRCh38
rs980753684 Chr.14: 64543404-64543404 on Build GRCh38
rs115466074 Chr.14: 64543326-64543326 on Build GRCh38
rs998003139 Chr.14: 64543309-64543309 on Build GRCh38
rs938393647 Chr.14: 64543558-64543558 on Build GRCh38
rs926705323 Chr.14: 64543440-64543440 on Build GRCh38
rs563437169 Chr.14: 64543503-64543503 on Build GRCh38
rs1044180903 Chr.14: 64543514-64543514 on Build GRCh38
rs988009962 Chr.14: 64543353-64543353 on Build GRCh38
rs968001736 Chr.14: 64543382-64543382 on Build GRCh38
rs747410798 Chr.14: 64543529-64543529 on Build GRCh38
rs747853946 Chr.14: 64543208-64543208 on Build GRCh38
rs964145681 Chr.14: 64543296-64543296 on Build GRCh38
rs45470505 Chr.14: 64543572-64543572 on Build GRCh38
rs191209915 Chr.14: 64543312-64543312 on Build GRCh38
rs953111255 Chr.14: 64543439-64543439 on Build GRCh38
rs1049375766 Chr.14: 64543205-64543205 on Build GRCh38
rs919413832 Chr.14: 64543486-64543486 on Build GRCh38
rs11848114 Chr.14: 64543642-64543642 on Build GRCh38
rs537412913 Chr.14: 64543527-64543527 on Build GRCh38
rs552028636 Chr.14: 64543285-64543285 on Build GRCh38
rs1056602420 Chr.14: 64543464-64543464 on Build GRCh38
rs984356317 Chr.14: 64543445-64543445 on Build GRCh38
rs571820113 Chr.14: 64543303-64543303 on Build GRCh38
rs1005246529 Chr.14: 64543227-64543227 on Build GRCh38
rs771486780 Chr.14: 64543555-64543555 on Build GRCh38
rs778316698 Chr.14: 64543449-64543449 on Build GRCh38
rs777066668 Chr.14: 64543629-64543629 on Build GRCh38
rs918585646 Chr.14: 64543393-64543393 on Build GRCh38
rs915197243 Chr.14: 64543354-64543354 on Build GRCh38
rs948221297 Chr.14: 64543500-64543500 on Build GRCh38
rs574720904 Chr.14: 64543290-64543290 on Build GRCh38

View Full Product Details