Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 65411775-65412262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT8
Entrez Gene ID
2530
Gene Name
fucosyltransferase 8
Gene Aliases
-
UniGene
Hs.597649
Gene Chromosome Location
Chr.14: 65410592-65744122 on Build GRCh38
Transcripts
NM_004480.4 NM_178155.2 NM_178156.2 NR_038167.1 XM_017021137.1 XM_017021138.1 NR_024334.1

Gene Symbol
FUT8-AS1
Entrez Gene ID
645431
Gene Name
FUT8 antisense RNA 1
Gene Aliases
-
UniGene
Hs.713717
Gene Chromosome Location
Chr.14: 65410592-65412617 on Build GRCh38
Transcripts
NM_004480.4 NM_178155.2 NM_178156.2 NR_038167.1 XM_017021137.1 XM_017021138.1 NR_024334.1

SNP Detail

SNP ID Chromosome Location
rs920761570 Chr.14: 65412145-65412145 on Build GRCh38
rs763775935 Chr.14: 65411988-65411988 on Build GRCh38
rs766689461 Chr.14: 65411973-65411973 on Build GRCh38
rs899145223 Chr.14: 65411886-65411886 on Build GRCh38
rs1008985904 Chr.14: 65411899-65411899 on Build GRCh38
rs761452449 Chr.14: 65412033-65412033 on Build GRCh38
rs930877745 Chr.14: 65412152-65412152 on Build GRCh38
rs969056730 Chr.14: 65412132-65412132 on Build GRCh38
rs371413321 Chr.14: 65412173-65412173 on Build GRCh38
rs921946620 Chr.14: 65412239-65412239 on Build GRCh38
rs1953416 Chr.14: 65412089-65412089 on Build GRCh38
rs978255966 Chr.14: 65412203-65412203 on Build GRCh38
rs957964704 Chr.14: 65411939-65411939 on Build GRCh38
rs1050697818 Chr.14: 65412154-65412154 on Build GRCh38
rs928111241 Chr.14: 65411980-65411980 on Build GRCh38
rs578017401 Chr.14: 65412024-65412024 on Build GRCh38
rs1010765941 Chr.14: 65412232-65412232 on Build GRCh38
rs940448829 Chr.14: 65412135-65412135 on Build GRCh38
rs899385401 Chr.14: 65412009-65412009 on Build GRCh38
rs775545817 Chr.14: 65412195-65412195 on Build GRCh38
rs756538977 Chr.14: 65412097-65412097 on Build GRCh38
rs991031512 Chr.14: 65412025-65412025 on Build GRCh38
rs753568175 Chr.14: 65412005-65412005 on Build GRCh38
rs1005538118 Chr.14: 65411872-65411872 on Build GRCh38
rs749881151 Chr.14: 65412078-65412078 on Build GRCh38
rs995572939 Chr.14: 65411974-65411974 on Build GRCh38
rs936365586 Chr.14: 65411845-65411845 on Build GRCh38
rs996716186 Chr.14: 65411922-65411922 on Build GRCh38
rs1052217520 Chr.14: 65412222-65412222 on Build GRCh38
rs553999657 Chr.14: 65412091-65412091 on Build GRCh38
rs1023532687 Chr.14: 65412234-65412234 on Build GRCh38
rs1007949469 Chr.14: 65411950-65411950 on Build GRCh38
rs191030169 Chr.14: 65411847-65411847 on Build GRCh38
rs542843643 Chr.14: 65412146-65412146 on Build GRCh38
rs150063048 Chr.14: 65412167-65412167 on Build GRCh38
rs924045735 Chr.14: 65412213-65412213 on Build GRCh38
rs989395628 Chr.14: 65412121-65412121 on Build GRCh38
rs1953417 Chr.14: 65412191-65412191 on Build GRCh38
rs890930666 Chr.14: 65411943-65411943 on Build GRCh38
rs566772515 Chr.14: 65411884-65411884 on Build GRCh38
rs916251232 Chr.14: 65411958-65411958 on Build GRCh38
rs944702370 Chr.14: 65412176-65412176 on Build GRCh38
rs991390780 Chr.14: 65411875-65411875 on Build GRCh38
rs1019054559 Chr.14: 65412070-65412070 on Build GRCh38
rs746889636 Chr.14: 65412225-65412225 on Build GRCh38
rs989073905 Chr.14: 65411956-65411956 on Build GRCh38
rs760311034 Chr.14: 65411915-65411915 on Build GRCh38
rs544822790 Chr.14: 65411896-65411896 on Build GRCh38
rs936697821 Chr.14: 65412111-65412111 on Build GRCh38
rs558149359 Chr.14: 65411951-65411951 on Build GRCh38
rs779605479 Chr.14: 65412134-65412134 on Build GRCh38
rs571217449 Chr.14: 65411826-65411826 on Build GRCh38
rs777992252 Chr.14: 65412105-65412105 on Build GRCh38
rs745901488 Chr.14: 65412183-65412183 on Build GRCh38
rs1050013668 Chr.14: 65411929-65411929 on Build GRCh38
rs978397935 Chr.14: 65411920-65411920 on Build GRCh38
rs1023251253 Chr.14: 65412130-65412130 on Build GRCh38
rs972223480 Chr.14: 65412139-65412139 on Build GRCh38
rs750060169 Chr.14: 65411898-65411898 on Build GRCh38
rs1012917850 Chr.14: 65412188-65412188 on Build GRCh38
rs758249967 Chr.14: 65412029-65412029 on Build GRCh38
rs918090839 Chr.14: 65412032-65412032 on Build GRCh38
rs1046962162 Chr.14: 65411859-65411859 on Build GRCh38
rs764787191 Chr.14: 65412047-65412047 on Build GRCh38
rs549241472 Chr.14: 65412133-65412133 on Build GRCh38
rs923591501 Chr.14: 65411837-65411837 on Build GRCh38
rs1030801991 Chr.14: 65412117-65412117 on Build GRCh38
rs961036738 Chr.14: 65411970-65411970 on Build GRCh38
rs947805021 Chr.14: 65411959-65411959 on Build GRCh38
rs1036955378 Chr.14: 65411993-65411993 on Build GRCh38
rs896904482 Chr.14: 65412169-65412169 on Build GRCh38
rs1009387450 Chr.14: 65412065-65412065 on Build GRCh38
rs906742317 Chr.14: 65411864-65411864 on Build GRCh38
rs779335082 Chr.14: 65411894-65411894 on Build GRCh38
rs546771953 Chr.14: 65411860-65411860 on Build GRCh38
rs749576845 Chr.14: 65412123-65412123 on Build GRCh38
rs1030896412 Chr.14: 65411930-65411930 on Build GRCh38
rs1036644157 Chr.14: 65411883-65411883 on Build GRCh38
rs958096753 Chr.14: 65412221-65412221 on Build GRCh38
rs34007703 Chr.14: 65412129-65412130 on Build GRCh38
rs947815107 Chr.14: 65411857-65411857 on Build GRCh38
rs569537470 Chr.14: 65411904-65411904 on Build GRCh38
rs537824436 Chr.14: 65411924-65411924 on Build GRCh38
rs957910781 Chr.14: 65412119-65412119 on Build GRCh38
rs774237224 Chr.14: 65411849-65411849 on Build GRCh38
rs755483282 Chr.14: 65412087-65412087 on Build GRCh38
rs555600599 Chr.14: 65411897-65411897 on Build GRCh38
rs577816927 Chr.14: 65411851-65411852 on Build GRCh38

View Full Product Details