Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69053662-69054159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF5
Entrez Gene ID
8816
Gene Name
DDB1 and CUL4 associated factor 5
Gene Aliases
BCRG2, BCRP2, D14S1461E, WDR22
UniGene
Hs.509780
Gene Chromosome Location
Chr.14: 69050920-69153393 on Build GRCh38
Transcripts
NM_001284206.1 NM_001284207.1 NM_003861.2 XM_006720297.2 XM_006720298.2 XM_006720299.3 XM_006720300.2 XM_011537278.1 XM_011537279.2 XM_011537280.2 XM_017021733.1 XM_017021734.1 XM_017021735.1 XM_017021736.1 XM_017021737.1 XM_017021738.1 XM_017021739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4052003 c.2653G>A p.G885R Chr.14: 69054033-69054033 on Build GRCh38
5693406 c.2569G>A p.V857M Chr.14: 69054117-69054117 on Build GRCh38
6076202 c.2787A>T p.T929T Chr.14: 69053899-69053899 on Build GRCh38
4909948 c.2568G>A p.V856V Chr.14: 69054118-69054118 on Build GRCh38
6640529 c.2605C>T p.R869C Chr.14: 69054081-69054081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746044894 Chr.14: 69053959-69053959 on Build GRCh38
rs758494282 Chr.14: 69053802-69053803 on Build GRCh38
rs936506320 Chr.14: 69053758-69053758 on Build GRCh38
rs760985782 Chr.14: 69053885-69053885 on Build GRCh38
rs772390014 Chr.14: 69054006-69054006 on Build GRCh38
rs779124260 Chr.14: 69053989-69053989 on Build GRCh38
rs76981261 Chr.14: 69053813-69053813 on Build GRCh38
rs959878156 Chr.14: 69053695-69053695 on Build GRCh38
rs200601552 Chr.14: 69054092-69054092 on Build GRCh38
rs562028591 Chr.14: 69053945-69053945 on Build GRCh38
rs767020168 Chr.14: 69054122-69054122 on Build GRCh38
rs377495549 Chr.14: 69053850-69053850 on Build GRCh38
rs754319970 Chr.14: 69053891-69053891 on Build GRCh38
rs765788180 Chr.14: 69054018-69054018 on Build GRCh38
rs539412134 Chr.14: 69053829-69053829 on Build GRCh38
rs565501310 Chr.14: 69053814-69053814 on Build GRCh38
rs748304211 Chr.14: 69054005-69054005 on Build GRCh38
rs756901508 Chr.14: 69053940-69053940 on Build GRCh38
rs925079168 Chr.14: 69053757-69053757 on Build GRCh38
rs878988165 Chr.14: 69053804-69053804 on Build GRCh38
rs373148713 Chr.14: 69054070-69054070 on Build GRCh38
rs147437843 Chr.14: 69054034-69054034 on Build GRCh38
rs547435274 Chr.14: 69053807-69053807 on Build GRCh38
rs777707629 Chr.14: 69054128-69054129 on Build GRCh38
rs1034611504 Chr.14: 69053694-69053694 on Build GRCh38
rs57470426 Chr.14: 69053802-69053802 on Build GRCh38
rs771227013 Chr.14: 69054014-69054014 on Build GRCh38
rs771266367 Chr.14: 69054082-69054082 on Build GRCh38
rs773706463 Chr.14: 69053852-69053852 on Build GRCh38
rs770265811 Chr.14: 69054110-69054110 on Build GRCh38
rs763596947 Chr.14: 69053917-69053917 on Build GRCh38
rs954936891 Chr.14: 69054055-69054055 on Build GRCh38
rs767112073 Chr.14: 69054024-69054024 on Build GRCh38
rs143850623 Chr.14: 69053890-69053890 on Build GRCh38
rs552468850 Chr.14: 69053843-69053843 on Build GRCh38
rs369898066 Chr.14: 69054081-69054081 on Build GRCh38
rs756412543 Chr.14: 69053815-69053815 on Build GRCh38
rs773616880 Chr.14: 69054012-69054012 on Build GRCh38
rs779032272 Chr.14: 69054047-69054047 on Build GRCh38
rs745732516 Chr.14: 69053810-69053810 on Build GRCh38
rs761276992 Chr.14: 69054115-69054115 on Build GRCh38
rs755872623 Chr.14: 69054031-69054031 on Build GRCh38
rs987589042 Chr.14: 69054073-69054073 on Build GRCh38
rs933522733 Chr.14: 69054098-69054098 on Build GRCh38
rs142890884 Chr.14: 69054023-69054023 on Build GRCh38
rs773822684 Chr.14: 69054021-69054021 on Build GRCh38
rs773101970 Chr.14: 69053739-69053739 on Build GRCh38
rs780003053 Chr.14: 69053956-69053956 on Build GRCh38
rs766102697 Chr.14: 69054033-69054033 on Build GRCh38
rs376381362 Chr.14: 69054080-69054080 on Build GRCh38
rs780890835 Chr.14: 69053803-69053803 on Build GRCh38
rs750214119 Chr.14: 69053943-69053943 on Build GRCh38
rs977988446 Chr.14: 69053756-69053756 on Build GRCh38
rs978771870 Chr.14: 69054094-69054094 on Build GRCh38
rs200458341 Chr.14: 69054026-69054026 on Build GRCh38
rs148204424 Chr.14: 69053942-69053942 on Build GRCh38
rs751152443 Chr.14: 69053918-69053918 on Build GRCh38
rs933469419 Chr.14: 69053796-69053796 on Build GRCh38
rs762481039 Chr.14: 69053912-69053912 on Build GRCh38
rs189780129 Chr.14: 69053787-69053787 on Build GRCh38
rs776003275 Chr.14: 69054114-69054114 on Build GRCh38
rs796282764 Chr.14: 69053814-69053814 on Build GRCh38
rs768800933 Chr.14: 69053976-69053976 on Build GRCh38
rs1052478322 Chr.14: 69053799-69053799 on Build GRCh38
rs140850060 Chr.14: 69053693-69053693 on Build GRCh38
rs374913170 Chr.14: 69053830-69053830 on Build GRCh38
rs945267972 Chr.14: 69053750-69053750 on Build GRCh38
rs755886819 Chr.14: 69053949-69053949 on Build GRCh38
rs770160949 Chr.14: 69054017-69054017 on Build GRCh38
rs199823757 Chr.14: 69053815-69053815 on Build GRCh38
rs895649017 Chr.14: 69053952-69053952 on Build GRCh38
rs913116929 Chr.14: 69053717-69053717 on Build GRCh38
rs974829934 Chr.14: 69053772-69053772 on Build GRCh38
rs774650930 Chr.14: 69053832-69053832 on Build GRCh38
rs754961000 Chr.14: 69054036-69054036 on Build GRCh38
rs1001710354 Chr.14: 69053870-69053870 on Build GRCh38
rs993158568 Chr.14: 69053705-69053705 on Build GRCh38
rs746287363 Chr.14: 69054109-69054109 on Build GRCh38

View Full Product Details