Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73491980-73492473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf169
Entrez Gene ID
79697
Gene Name
chromosome 14 open reading frame 169
Gene Aliases
MAPJD, NO66, ROX, URLC2, hsNO66
UniGene
Hs.509916
Gene Chromosome Location
Chr.14: 73490934-73493401 on Build GRCh38
Transcripts
NM_024644.4 NM_001220484.1 NM_203309.2 XM_006720143.2 XM_011536760.2 XM_017021289.1

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
-
UniGene
Hs.660774
Gene Chromosome Location
Chr.14: 73478484-73634418 on Build GRCh38
Transcripts
NM_024644.4 NM_001220484.1 NM_203309.2 XM_006720143.2 XM_011536760.2 XM_017021289.1

SNP Detail

SNP ID Chromosome Location
rs755394026 Chr.14: 73492016-73492016 on Build GRCh38
rs1032136206 Chr.14: 73492403-73492403 on Build GRCh38
rs774321251 Chr.14: 73492039-73492039 on Build GRCh38
rs749141342 Chr.14: 73492118-73492118 on Build GRCh38
rs766754627 Chr.14: 73492069-73492069 on Build GRCh38
rs973283876 Chr.14: 73492189-73492189 on Build GRCh38
rs920517839 Chr.14: 73492196-73492196 on Build GRCh38
rs369146034 Chr.14: 73492283-73492283 on Build GRCh38
rs748047436 Chr.14: 73492147-73492147 on Build GRCh38
rs761072269 Chr.14: 73492447-73492447 on Build GRCh38
rs756770892 Chr.14: 73492024-73492024 on Build GRCh38
rs755339913 Chr.14: 73492091-73492091 on Build GRCh38
rs900665392 Chr.14: 73492410-73492410 on Build GRCh38
rs760833500 Chr.14: 73492062-73492062 on Build GRCh38
rs753234159 Chr.14: 73492099-73492099 on Build GRCh38
rs745446376 Chr.14: 73492030-73492030 on Build GRCh38
rs929268742 Chr.14: 73492201-73492202 on Build GRCh38
rs778715177 Chr.14: 73492279-73492279 on Build GRCh38
rs979254434 Chr.14: 73492354-73492354 on Build GRCh38
rs779820725 Chr.14: 73492113-73492113 on Build GRCh38
rs756533980 Chr.14: 73492007-73492007 on Build GRCh38
rs142849889 Chr.14: 73492163-73492163 on Build GRCh38
rs780433095 Chr.14: 73492079-73492080 on Build GRCh38
rs773430467 Chr.14: 73492052-73492052 on Build GRCh38
rs3813563 Chr.14: 73492108-73492108 on Build GRCh38
rs957448503 Chr.14: 73492440-73492440 on Build GRCh38
rs761852553 Chr.14: 73492047-73492047 on Build GRCh38
rs957659122 Chr.14: 73492021-73492021 on Build GRCh38
rs201606461 Chr.14: 73492019-73492019 on Build GRCh38
rs760921243 Chr.14: 73492161-73492161 on Build GRCh38
rs773231441 Chr.14: 73492150-73492150 on Build GRCh38
rs752096493 Chr.14: 73492407-73492407 on Build GRCh38
rs763467884 Chr.14: 73492203-73492203 on Build GRCh38
rs780766062 Chr.14: 73492025-73492025 on Build GRCh38
rs777660153 Chr.14: 73492318-73492318 on Build GRCh38
rs747344738 Chr.14: 73492083-73492084 on Build GRCh38
rs764746561 Chr.14: 73492401-73492401 on Build GRCh38
rs374728003 Chr.14: 73492022-73492022 on Build GRCh38
rs553099723 Chr.14: 73492361-73492361 on Build GRCh38
rs755786721 Chr.14: 73492110-73492110 on Build GRCh38
rs767626494 Chr.14: 73492048-73492048 on Build GRCh38
rs769724580 Chr.14: 73492031-73492031 on Build GRCh38
rs890515773 Chr.14: 73492289-73492289 on Build GRCh38
rs759738436 Chr.14: 73492353-73492353 on Build GRCh38
rs775884852 Chr.14: 73492395-73492395 on Build GRCh38
rs768481418 Chr.14: 73492134-73492134 on Build GRCh38
rs766668527 Chr.14: 73492010-73492010 on Build GRCh38
rs765714311 Chr.14: 73492098-73492098 on Build GRCh38
rs896116434 Chr.14: 73492238-73492238 on Build GRCh38
rs749992591 Chr.14: 73492218-73492218 on Build GRCh38
rs767165747 Chr.14: 73492210-73492210 on Build GRCh38
rs754165408 Chr.14: 73492080-73492080 on Build GRCh38
rs1048831716 Chr.14: 73492122-73492122 on Build GRCh38
rs776906419 Chr.14: 73492347-73492347 on Build GRCh38
rs200603832 Chr.14: 73492148-73492148 on Build GRCh38
rs368447512 Chr.14: 73492144-73492144 on Build GRCh38
rs1014601695 Chr.14: 73492191-73492191 on Build GRCh38
rs1013526695 Chr.14: 73492280-73492280 on Build GRCh38
rs759878315 Chr.14: 73492171-73492171 on Build GRCh38
rs754359850 Chr.14: 73492012-73492012 on Build GRCh38
rs187143550 Chr.14: 73492253-73492253 on Build GRCh38
rs756784555 Chr.14: 73492102-73492102 on Build GRCh38
rs755660287 Chr.14: 73492247-73492247 on Build GRCh38
rs17850736 Chr.14: 73492109-73492109 on Build GRCh38
rs768512571 Chr.14: 73492036-73492036 on Build GRCh38
rs747141988 Chr.14: 73492325-73492325 on Build GRCh38
rs763194090 Chr.14: 73492400-73492400 on Build GRCh38
rs199545926 Chr.14: 73492035-73492035 on Build GRCh38
rs776812249 Chr.14: 73492166-73492166 on Build GRCh38
rs534735274 Chr.14: 73492274-73492274 on Build GRCh38
rs753157707 Chr.14: 73492177-73492177 on Build GRCh38
rs770987206 Chr.14: 73492340-73492340 on Build GRCh38
rs765631665 Chr.14: 73492173-73492173 on Build GRCh38
rs377119311 Chr.14: 73492132-73492132 on Build GRCh38
rs770076164 Chr.14: 73492363-73492363 on Build GRCh38
rs866113327 Chr.14: 73492011-73492011 on Build GRCh38
rs760325765 Chr.14: 73492450-73492450 on Build GRCh38
rs753520163 Chr.14: 73492259-73492259 on Build GRCh38
rs750627357 Chr.14: 73492001-73492001 on Build GRCh38

View Full Product Details