Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74888803-74889341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLST
Entrez Gene ID
1743
Gene Name
dihydrolipoamide S-succinyltransferase
Gene Aliases
DLTS
UniGene
Hs.525459
Gene Chromosome Location
Chr.14: 74881891-74903747 on Build GRCh38
Transcripts
NM_001244883.1 NM_001933.4 NR_033814.1 NR_045209.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
260443 c.201G>T p.K67N Chr.14: 74889276-74889276 on Build GRCh38
136978 c.242C>T p.S81F Chr.14: 74889317-74889317 on Build GRCh38
6370871 c.149T>C p.I50T Chr.14: 74889097-74889097 on Build GRCh38
957871 c.216A>G p.T72T Chr.14: 74889291-74889291 on Build GRCh38
1238685 c.170T>C p.V57A Chr.14: 74889118-74889118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199662378 Chr.14: 74889065-74889065 on Build GRCh38
rs755919109 Chr.14: 74889167-74889167 on Build GRCh38
rs962359490 Chr.14: 74888947-74888947 on Build GRCh38
rs781280966 Chr.14: 74889225-74889227 on Build GRCh38
rs747212553 Chr.14: 74889194-74889197 on Build GRCh38
rs376908827 Chr.14: 74889304-74889304 on Build GRCh38
rs749175326 Chr.14: 74889250-74889251 on Build GRCh38
rs772702319 Chr.14: 74888945-74888945 on Build GRCh38
rs778326717 Chr.14: 74889255-74889255 on Build GRCh38
rs148493046 Chr.14: 74889228-74889228 on Build GRCh38
rs775260019 Chr.14: 74889126-74889126 on Build GRCh38
rs766708231 Chr.14: 74889121-74889121 on Build GRCh38
rs57898381 Chr.14: 74889065-74889065 on Build GRCh38
rs780384489 Chr.14: 74889133-74889133 on Build GRCh38
rs944550348 Chr.14: 74889189-74889189 on Build GRCh38
rs775844914 Chr.14: 74888974-74888974 on Build GRCh38
rs751511772 Chr.14: 74889047-74889047 on Build GRCh38
rs141349679 Chr.14: 74889289-74889289 on Build GRCh38
rs761093767 Chr.14: 74889116-74889116 on Build GRCh38
rs768693345 Chr.14: 74889068-74889068 on Build GRCh38
rs763870039 Chr.14: 74889128-74889128 on Build GRCh38
rs564729500 Chr.14: 74888972-74888972 on Build GRCh38
rs955919358 Chr.14: 74889114-74889114 on Build GRCh38
rs975022349 Chr.14: 74889190-74889190 on Build GRCh38
rs559187037 Chr.14: 74888865-74888865 on Build GRCh38
rs747517093 Chr.14: 74889262-74889262 on Build GRCh38
rs1012862759 Chr.14: 74889011-74889011 on Build GRCh38
rs369555129 Chr.14: 74889248-74889248 on Build GRCh38
rs201737883 Chr.14: 74889275-74889275 on Build GRCh38
rs750346842 Chr.14: 74889316-74889316 on Build GRCh38
rs995218963 Chr.14: 74888951-74888951 on Build GRCh38
rs1055546433 Chr.14: 74889220-74889224 on Build GRCh38
rs780243673 Chr.14: 74889066-74889066 on Build GRCh38
rs530073855 Chr.14: 74889225-74889225 on Build GRCh38
rs544757896 Chr.14: 74888971-74888971 on Build GRCh38
rs1034125376 Chr.14: 74888838-74888838 on Build GRCh38
rs199918787 Chr.14: 74889063-74889063 on Build GRCh38
rs750386371 Chr.14: 74889155-74889155 on Build GRCh38
rs767485421 Chr.14: 74889141-74889141 on Build GRCh38
rs975453714 Chr.14: 74889030-74889030 on Build GRCh38
rs964201769 Chr.14: 74889028-74889028 on Build GRCh38
rs1049054515 Chr.14: 74888884-74888884 on Build GRCh38
rs911829574 Chr.14: 74889171-74889171 on Build GRCh38
rs912705019 Chr.14: 74889014-74889014 on Build GRCh38
rs377574623 Chr.14: 74889197-74889197 on Build GRCh38
rs930738098 Chr.14: 74889223-74889223 on Build GRCh38
rs548822051 Chr.14: 74889296-74889296 on Build GRCh38
rs561148939 Chr.14: 74889194-74889194 on Build GRCh38
rs907017643 Chr.14: 74889195-74889198 on Build GRCh38
rs749564778 Chr.14: 74889067-74889067 on Build GRCh38
rs202061804 Chr.14: 74889112-74889112 on Build GRCh38
rs373501223 Chr.14: 74889137-74889137 on Build GRCh38
rs762733268 Chr.14: 74889127-74889127 on Build GRCh38
rs1026316635 Chr.14: 74888963-74888963 on Build GRCh38
rs35027509 Chr.14: 74888946-74888947 on Build GRCh38
rs755023041 Chr.14: 74889196-74889196 on Build GRCh38
rs539340428 Chr.14: 74889305-74889305 on Build GRCh38
rs769156375 Chr.14: 74889143-74889143 on Build GRCh38
rs371402604 Chr.14: 74889306-74889306 on Build GRCh38
rs542578531 Chr.14: 74888848-74888848 on Build GRCh38
rs987469277 Chr.14: 74888989-74888989 on Build GRCh38
rs398077873 Chr.14: 74889067-74889067 on Build GRCh38
rs191062417 Chr.14: 74888973-74888973 on Build GRCh38
rs376444659 Chr.14: 74889057-74889057 on Build GRCh38
rs200375753 Chr.14: 74889278-74889278 on Build GRCh38
rs147816400 Chr.14: 74889096-74889096 on Build GRCh38
rs772107822 Chr.14: 74889249-74889249 on Build GRCh38
rs572709560 Chr.14: 74888966-74888966 on Build GRCh38
rs532606848 Chr.14: 74889181-74889181 on Build GRCh38
rs781007119 Chr.14: 74889054-74889054 on Build GRCh38
rs907862524 Chr.14: 74889240-74889240 on Build GRCh38
rs202151716 Chr.14: 74889069-74889069 on Build GRCh38
rs201278080 Chr.14: 74889078-74889078 on Build GRCh38
rs796387204 Chr.14: 74889068-74889068 on Build GRCh38
rs751064187 Chr.14: 74889136-74889136 on Build GRCh38
rs773592672 Chr.14: 74889107-74889107 on Build GRCh38
rs201088603 Chr.14: 74889060-74889060 on Build GRCh38
rs200136157 Chr.14: 74889111-74889111 on Build GRCh38

View Full Product Details