Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 76828187-76828691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC74A
Entrez Gene ID
145497
Gene Name
leucine rich repeat containing 74A
Gene Aliases
C14orf166B, LRRC74
UniGene
Hs.147276
Gene Chromosome Location
Chr.14: 76826370-76870302 on Build GRCh38
Transcripts
NM_194287.2 XM_011536479.1 XM_011536483.1 XM_011536487.1 XM_017021032.1 XM_017021033.1 XM_017021034.1 XM_017021035.1 XM_017021036.1 XM_017021037.1 XM_017021038.1 XM_017021039.1 XM_017021040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5664069 c.26C>A p.P9Q Chr.14: 76828357-76828357 on Build GRCh38
5563803 c.156G>A p.Q52Q Chr.14: 76828487-76828487 on Build GRCh38
5362570 c.232C>G p.R78G Chr.14: 76828563-76828563 on Build GRCh38
5718723 c.97G>A p.E33K Chr.14: 76828299-76828299 on Build GRCh38
4394329 c.196G>A p.E66K Chr.14: 76828398-76828398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117254981 Chr.14: 76828386-76828386 on Build GRCh38
rs756104384 Chr.14: 76828442-76828449 on Build GRCh38
rs375929129 Chr.14: 76828664-76828664 on Build GRCh38
rs769766997 Chr.14: 76828419-76828419 on Build GRCh38
rs1004762428 Chr.14: 76828322-76828322 on Build GRCh38
rs934913224 Chr.14: 76828234-76828234 on Build GRCh38
rs776556612 Chr.14: 76828328-76828328 on Build GRCh38
rs1038480397 Chr.14: 76828416-76828416 on Build GRCh38
rs368136760 Chr.14: 76828297-76828297 on Build GRCh38
rs911897936 Chr.14: 76828268-76828268 on Build GRCh38
rs193060849 Chr.14: 76828421-76828421 on Build GRCh38
rs770840640 Chr.14: 76828247-76828247 on Build GRCh38
rs754340654 Chr.14: 76828522-76828522 on Build GRCh38
rs773473884 Chr.14: 76828468-76828468 on Build GRCh38
rs372636254 Chr.14: 76828412-76828412 on Build GRCh38
rs944674444 Chr.14: 76828425-76828425 on Build GRCh38
rs768365328 Chr.14: 76828561-76828561 on Build GRCh38
rs752408385 Chr.14: 76828524-76828524 on Build GRCh38
rs754322171 Chr.14: 76828361-76828361 on Build GRCh38
rs755623685 Chr.14: 76828614-76828614 on Build GRCh38
rs61745625 Chr.14: 76828431-76828431 on Build GRCh38
rs558611602 Chr.14: 76828450-76828450 on Build GRCh38
rs75720627 Chr.14: 76828563-76828563 on Build GRCh38
rs752982441 Chr.14: 76828360-76828361 on Build GRCh38
rs559616252 Chr.14: 76828217-76828217 on Build GRCh38
rs751196561 Chr.14: 76828526-76828526 on Build GRCh38
rs767051479 Chr.14: 76828496-76828496 on Build GRCh38
rs17104907 Chr.14: 76828533-76828533 on Build GRCh38
rs1000623394 Chr.14: 76828596-76828596 on Build GRCh38
rs751459185 Chr.14: 76828460-76828460 on Build GRCh38
rs960356227 Chr.14: 76828335-76828335 on Build GRCh38
rs886273472 Chr.14: 76828527-76828527 on Build GRCh38
rs61740619 Chr.14: 76828587-76828587 on Build GRCh38
rs975117591 Chr.14: 76828661-76828661 on Build GRCh38
rs113978608 Chr.14: 76828597-76828597 on Build GRCh38
rs148747129 Chr.14: 76828360-76828360 on Build GRCh38
rs750350010 Chr.14: 76828511-76828512 on Build GRCh38
rs754990471 Chr.14: 76828629-76828629 on Build GRCh38
rs764948397 Chr.14: 76828256-76828256 on Build GRCh38
rs200189415 Chr.14: 76828314-76828314 on Build GRCh38
rs538117698 Chr.14: 76828354-76828354 on Build GRCh38
rs766502380 Chr.14: 76828584-76828584 on Build GRCh38
rs568264390 Chr.14: 76828358-76828358 on Build GRCh38
rs770419685 Chr.14: 76828326-76828326 on Build GRCh38
rs760832719 Chr.14: 76828480-76828480 on Build GRCh38
rs575624682 Chr.14: 76828531-76828531 on Build GRCh38
rs114437600 Chr.14: 76828311-76828311 on Build GRCh38
rs772463355 Chr.14: 76828564-76828564 on Build GRCh38
rs758690710 Chr.14: 76828605-76828605 on Build GRCh38
rs779977598 Chr.14: 76828540-76828540 on Build GRCh38
rs760852743 Chr.14: 76828575-76828575 on Build GRCh38
rs749180244 Chr.14: 76828560-76828560 on Build GRCh38
rs746560301 Chr.14: 76828325-76828325 on Build GRCh38
rs1018648336 Chr.14: 76828223-76828223 on Build GRCh38
rs778683960 Chr.14: 76828641-76828641 on Build GRCh38
rs552624929 Chr.14: 76828312-76828312 on Build GRCh38
rs561001907 Chr.14: 76828627-76828627 on Build GRCh38
rs765598737 Chr.14: 76828525-76828525 on Build GRCh38
rs753150054 Chr.14: 76828601-76828601 on Build GRCh38
rs751626615 Chr.14: 76828275-76828275 on Build GRCh38
rs764285903 Chr.14: 76828351-76828351 on Build GRCh38
rs745363240 Chr.14: 76828387-76828387 on Build GRCh38
rs376082185 Chr.14: 76828384-76828384 on Build GRCh38
rs201170754 Chr.14: 76828257-76828257 on Build GRCh38
rs776618874 Chr.14: 76828248-76828248 on Build GRCh38
rs561030028 Chr.14: 76828218-76828218 on Build GRCh38
rs34018192 Chr.14: 76828503-76828504 on Build GRCh38
rs371545140 Chr.14: 76828259-76828259 on Build GRCh38
rs777037895 Chr.14: 76828586-76828586 on Build GRCh38
rs1039056140 Chr.14: 76828427-76828427 on Build GRCh38
rs760202231 Chr.14: 76828241-76828241 on Build GRCh38
rs780795797 Chr.14: 76828532-76828532 on Build GRCh38
rs769603711 Chr.14: 76828341-76828341 on Build GRCh38
rs778145098 Chr.14: 76828662-76828662 on Build GRCh38
rs77362293 Chr.14: 76828656-76828656 on Build GRCh38
rs759433222 Chr.14: 76828337-76828337 on Build GRCh38
rs763117012 Chr.14: 76828264-76828264 on Build GRCh38
rs1023788303 Chr.14: 76828357-76828357 on Build GRCh38
rs773818563 Chr.14: 76828349-76828349 on Build GRCh38
rs750997291 Chr.14: 76828359-76828359 on Build GRCh38
rs561073167 Chr.14: 76828534-76828534 on Build GRCh38
rs762701243 Chr.14: 76828350-76828350 on Build GRCh38
rs766351506 Chr.14: 76828346-76828346 on Build GRCh38
rs777305722 Chr.14: 76828364-76828364 on Build GRCh38
rs769632033 Chr.14: 76828224-76828224 on Build GRCh38
rs764271131 Chr.14: 76828451-76828452 on Build GRCh38
rs759341485 Chr.14: 76828250-76828250 on Build GRCh38
rs746608206 Chr.14: 76828385-76828385 on Build GRCh38
rs749091853 Chr.14: 76828422-76828422 on Build GRCh38
rs753739900 Chr.14: 76828621-76828621 on Build GRCh38
rs762054237 Chr.14: 76828440-76828440 on Build GRCh38
rs373979790 Chr.14: 76828272-76828272 on Build GRCh38
rs2142189 Chr.14: 76828607-76828607 on Build GRCh38
rs781605165 Chr.14: 76828283-76828283 on Build GRCh38
rs61742043 Chr.14: 76828613-76828613 on Build GRCh38
rs896432162 Chr.14: 76828551-76828551 on Build GRCh38
rs771934942 Chr.14: 76828657-76828657 on Build GRCh38
rs531888715 Chr.14: 76828620-76828620 on Build GRCh38
rs554766770 Chr.14: 76828355-76828355 on Build GRCh38
rs777075183 Chr.14: 76828251-76828251 on Build GRCh38
rs773555405 Chr.14: 76828570-76828570 on Build GRCh38
rs768438118 Chr.14: 76828424-76828424 on Build GRCh38

View Full Product Details