Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77457388-77457883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHSA1
Entrez Gene ID
10598
Gene Name
activator of Hsp90 ATPase activity 1
Gene Aliases
AHA1, C14orf3, hAha1, p38
UniGene
Hs.204041
Gene Chromosome Location
Chr.14: 77457877-77469472 on Build GRCh38
Transcripts
NM_001321441.1 NM_001193314.1 NM_001193315.1 NM_001193316.1 NM_001193317.1 NM_022067.3 XM_011537066.1 XM_017021580.1

Gene Symbol
VIPAS39
Entrez Gene ID
63894
Gene Name
VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog
Gene Aliases
C14orf133, SPE-39, SPE39, VIPAR, VPS16B, hSPE-39
UniGene
Hs.16157
Gene Chromosome Location
Chr.14: 77426675-77457640 on Build GRCh38
Transcripts
NM_001321441.1 NM_001193314.1 NM_001193315.1 NM_001193316.1 NM_001193317.1 NM_022067.3 XM_011537066.1 XM_017021580.1

SNP Detail

SNP ID Chromosome Location
rs1042685542 Chr.14: 77457660-77457660 on Build GRCh38
rs1012552334 Chr.14: 77457735-77457735 on Build GRCh38
rs140162641 Chr.14: 77457522-77457522 on Build GRCh38
rs983330227 Chr.14: 77457442-77457442 on Build GRCh38
rs11845345 Chr.14: 77457440-77457440 on Build GRCh38
rs1004906979 Chr.14: 77457700-77457700 on Build GRCh38
rs936500018 Chr.14: 77457695-77457695 on Build GRCh38
rs970919482 Chr.14: 77457476-77457476 on Build GRCh38
rs1053910489 Chr.14: 77457744-77457744 on Build GRCh38
rs181945939 Chr.14: 77457464-77457464 on Build GRCh38
rs923730639 Chr.14: 77457790-77457790 on Build GRCh38
rs758824700 Chr.14: 77457414-77457414 on Build GRCh38
rs562086652 Chr.14: 77457616-77457616 on Build GRCh38
rs755735741 Chr.14: 77457437-77457437 on Build GRCh38
rs56029059 Chr.14: 77457745-77457745 on Build GRCh38
rs944980620 Chr.14: 77457856-77457856 on Build GRCh38
rs781060108 Chr.14: 77457420-77457420 on Build GRCh38
rs886521546 Chr.14: 77457696-77457696 on Build GRCh38
rs943921031 Chr.14: 77457531-77457531 on Build GRCh38
rs765544292 Chr.14: 77457673-77457673 on Build GRCh38
rs1018177016 Chr.14: 77457562-77457562 on Build GRCh38
rs750070273 Chr.14: 77457427-77457427 on Build GRCh38
rs141151001 Chr.14: 77457780-77457780 on Build GRCh38
rs1021489299 Chr.14: 77457441-77457441 on Build GRCh38
rs912176080 Chr.14: 77457637-77457637 on Build GRCh38
rs1032823383 Chr.14: 77457579-77457579 on Build GRCh38
rs944924710 Chr.14: 77457645-77457645 on Build GRCh38
rs548570233 Chr.14: 77457756-77457756 on Build GRCh38
rs529513710 Chr.14: 77457751-77457751 on Build GRCh38
rs916383822 Chr.14: 77457618-77457618 on Build GRCh38
rs1009092141 Chr.14: 77457538-77457538 on Build GRCh38
rs923847321 Chr.14: 77457825-77457825 on Build GRCh38
rs367721441 Chr.14: 77457532-77457532 on Build GRCh38
rs748375921 Chr.14: 77457614-77457614 on Build GRCh38
rs986497371 Chr.14: 77457802-77457802 on Build GRCh38
rs1024946030 Chr.14: 77457605-77457605 on Build GRCh38
rs895189603 Chr.14: 77457580-77457580 on Build GRCh38
rs912261326 Chr.14: 77457794-77457794 on Build GRCh38
rs140081211 Chr.14: 77457412-77457412 on Build GRCh38
rs774740359 Chr.14: 77457613-77457614 on Build GRCh38
rs570028474 Chr.14: 77457747-77457747 on Build GRCh38
rs745600059 Chr.14: 77457847-77457847 on Build GRCh38
rs964998138 Chr.14: 77457797-77457797 on Build GRCh38
rs1046451586 Chr.14: 77457746-77457746 on Build GRCh38
rs78883841 Chr.14: 77457615-77457615 on Build GRCh38
rs932299756 Chr.14: 77457516-77457516 on Build GRCh38
rs764730503 Chr.14: 77457587-77457587 on Build GRCh38
rs890693601 Chr.14: 77457528-77457528 on Build GRCh38
rs565326401 Chr.14: 77457602-77457602 on Build GRCh38
rs994900122 Chr.14: 77457586-77457586 on Build GRCh38
rs868487738 Chr.14: 77457633-77457633 on Build GRCh38
rs543637739 Chr.14: 77457581-77457581 on Build GRCh38
rs1035417398 Chr.14: 77457711-77457711 on Build GRCh38
rs541478418 Chr.14: 77457671-77457671 on Build GRCh38
rs974199655 Chr.14: 77457501-77457501 on Build GRCh38
rs934915740 Chr.14: 77457511-77457511 on Build GRCh38
rs532432455 Chr.14: 77457632-77457632 on Build GRCh38
rs960667836 Chr.14: 77457722-77457722 on Build GRCh38
rs533396978 Chr.14: 77457702-77457702 on Build GRCh38
rs919630313 Chr.14: 77457514-77457514 on Build GRCh38
rs151172490 Chr.14: 77457474-77457474 on Build GRCh38
rs1049617614 Chr.14: 77457423-77457423 on Build GRCh38
rs1042244816 Chr.14: 77457535-77457535 on Build GRCh38
rs117264193 Chr.14: 77457482-77457482 on Build GRCh38
rs967789717 Chr.14: 77457622-77457622 on Build GRCh38
rs1043383716 Chr.14: 77457485-77457485 on Build GRCh38
rs891032694 Chr.14: 77457530-77457530 on Build GRCh38
rs979226227 Chr.14: 77457761-77457761 on Build GRCh38
rs906503647 Chr.14: 77457750-77457750 on Build GRCh38
rs923616169 Chr.14: 77457626-77457626 on Build GRCh38
rs902164018 Chr.14: 77457507-77457507 on Build GRCh38
rs145506107 Chr.14: 77457457-77457457 on Build GRCh38
rs144874673 Chr.14: 77457803-77457803 on Build GRCh38
rs995404719 Chr.14: 77457768-77457768 on Build GRCh38
rs1015572756 Chr.14: 77457757-77457757 on Build GRCh38
rs956647413 Chr.14: 77457792-77457792 on Build GRCh38
rs527786452 Chr.14: 77457623-77457623 on Build GRCh38
rs149884162 Chr.14: 77457675-77457675 on Build GRCh38
rs953797703 Chr.14: 77457788-77457788 on Build GRCh38
rs114784880 Chr.14: 77457454-77457454 on Build GRCh38
rs1005162430 Chr.14: 77457752-77457752 on Build GRCh38
rs907996734 Chr.14: 77457486-77457486 on Build GRCh38
rs1051185282 Chr.14: 77457635-77457635 on Build GRCh38
rs761217427 Chr.14: 77457489-77457489 on Build GRCh38
rs1049211128 Chr.14: 77457689-77457689 on Build GRCh38
rs912208931 Chr.14: 77457618-77457619 on Build GRCh38
rs573969004 Chr.14: 77457612-77457612 on Build GRCh38
rs936569077 Chr.14: 77457857-77457857 on Build GRCh38
rs950664037 Chr.14: 77457600-77457600 on Build GRCh38
rs577808219 Chr.14: 77457540-77457540 on Build GRCh38

View Full Product Details