Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 88014187-88014727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR65
Entrez Gene ID
8477
Gene Name
G protein-coupled receptor 65
Gene Aliases
TDAG8, hTDAG8
UniGene
Hs.443243
Gene Chromosome Location
Chr.14: 88005124-88014811 on Build GRCh38
Transcripts
NM_003608.3

SNP Detail

SNP ID Chromosome Location
rs534370394 Chr.14: 88014633-88014633 on Build GRCh38
rs963405832 Chr.14: 88014402-88014402 on Build GRCh38
rs769629146 Chr.14: 88014271-88014271 on Build GRCh38
rs899403343 Chr.14: 88014681-88014681 on Build GRCh38
rs975391716 Chr.14: 88014507-88014507 on Build GRCh38
rs938421080 Chr.14: 88014624-88014624 on Build GRCh38
rs1013128754 Chr.14: 88014375-88014375 on Build GRCh38
rs1044526400 Chr.14: 88014331-88014331 on Build GRCh38
rs114611580 Chr.14: 88014627-88014627 on Build GRCh38
rs377453338 Chr.14: 88014371-88014371 on Build GRCh38
rs780550981 Chr.14: 88014295-88014295 on Build GRCh38
rs554246336 Chr.14: 88014212-88014212 on Build GRCh38
rs912360151 Chr.14: 88014464-88014464 on Build GRCh38
rs954030595 Chr.14: 88014516-88014516 on Build GRCh38
rs370804092 Chr.14: 88014704-88014704 on Build GRCh38
rs947076002 Chr.14: 88014529-88014529 on Build GRCh38
rs1026430164 Chr.14: 88014423-88014423 on Build GRCh38
rs935675334 Chr.14: 88014684-88014684 on Build GRCh38
rs949855065 Chr.14: 88014313-88014313 on Build GRCh38
rs367995572 Chr.14: 88014254-88014254 on Build GRCh38
rs114064763 Chr.14: 88014275-88014275 on Build GRCh38
rs560485000 Chr.14: 88014225-88014225 on Build GRCh38
rs1018817461 Chr.14: 88014455-88014455 on Build GRCh38
rs758037361 Chr.14: 88014356-88014356 on Build GRCh38
rs964315623 Chr.14: 88014500-88014500 on Build GRCh38
rs1057268820 Chr.14: 88014343-88014343 on Build GRCh38
rs5014611 Chr.14: 88014222-88014222 on Build GRCh38
rs901654771 Chr.14: 88014491-88014491 on Build GRCh38
rs754320506 Chr.14: 88014325-88014325 on Build GRCh38
rs564032046 Chr.14: 88014382-88014382 on Build GRCh38
rs1041349925 Chr.14: 88014598-88014598 on Build GRCh38
rs1003479568 Chr.14: 88014311-88014311 on Build GRCh38
rs7156702 Chr.14: 88014673-88014673 on Build GRCh38
rs984088109 Chr.14: 88014541-88014541 on Build GRCh38
rs77865971 Chr.14: 88014615-88014615 on Build GRCh38
rs574065189 Chr.14: 88014217-88014217 on Build GRCh38
rs914485924 Chr.14: 88014509-88014509 on Build GRCh38
rs35347074 Chr.14: 88014230-88014231 on Build GRCh38
rs566167351 Chr.14: 88014461-88014461 on Build GRCh38
rs141334693 Chr.14: 88014682-88014682 on Build GRCh38
rs1016940378 Chr.14: 88014550-88014550 on Build GRCh38
rs1013493058 Chr.14: 88014654-88014654 on Build GRCh38
rs993479916 Chr.14: 88014411-88014411 on Build GRCh38
rs532909272 Chr.14: 88014448-88014448 on Build GRCh38
rs546280011 Chr.14: 88014451-88014451 on Build GRCh38

View Full Product Details