Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91232440-91232940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR68
Entrez Gene ID
8111
Gene Name
G protein-coupled receptor 68
Gene Aliases
GPR12A, OGR1
UniGene
Hs.8882
Gene Chromosome Location
Chr.14: 91232532-91264581 on Build GRCh38
Transcripts
NM_001177676.1 NM_003485.3 XM_005268110.4 XM_005268111.3 XM_005268112.3 XM_006720262.3 XM_011537196.2 XM_011537197.2 XM_011537198.2 XM_011537199.2

SNP Detail

SNP ID Chromosome Location
rs565189614 Chr.14: 91232894-91232894 on Build GRCh38
rs906306178 Chr.14: 91232563-91232563 on Build GRCh38
rs1052897326 Chr.14: 91232708-91232708 on Build GRCh38
rs1039357894 Chr.14: 91232895-91232895 on Build GRCh38
rs960349009 Chr.14: 91232841-91232841 on Build GRCh38
rs867025462 Chr.14: 91232879-91232879 on Build GRCh38
rs973423636 Chr.14: 91232790-91232790 on Build GRCh38
rs185818230 Chr.14: 91232671-91232671 on Build GRCh38
rs370308874 Chr.14: 91232799-91232801 on Build GRCh38
rs570485819 Chr.14: 91232510-91232510 on Build GRCh38
rs899783024 Chr.14: 91232904-91232904 on Build GRCh38
rs542589181 Chr.14: 91232756-91232756 on Build GRCh38
rs1050143700 Chr.14: 91232891-91232891 on Build GRCh38
rs114312192 Chr.14: 91232500-91232500 on Build GRCh38
rs146370096 Chr.14: 91232469-91232469 on Build GRCh38
rs544930097 Chr.14: 91232881-91232881 on Build GRCh38
rs951189850 Chr.14: 91232743-91232743 on Build GRCh38
rs943689396 Chr.14: 91232911-91232911 on Build GRCh38
rs748459063 Chr.14: 91232884-91232884 on Build GRCh38
rs1010846619 Chr.14: 91232580-91232580 on Build GRCh38
rs980138467 Chr.14: 91232472-91232472 on Build GRCh38
rs935085207 Chr.14: 91232566-91232566 on Build GRCh38
rs369050573 Chr.14: 91232793-91232793 on Build GRCh38
rs970351485 Chr.14: 91232789-91232789 on Build GRCh38
rs576285910 Chr.14: 91232868-91232868 on Build GRCh38
rs932490383 Chr.14: 91232885-91232885 on Build GRCh38
rs577774361 Chr.14: 91232591-91232591 on Build GRCh38
rs542803472 Chr.14: 91232590-91232590 on Build GRCh38
rs891513592 Chr.14: 91232748-91232748 on Build GRCh38
rs939896207 Chr.14: 91232515-91232515 on Build GRCh38
rs973896268 Chr.14: 91232882-91232882 on Build GRCh38
rs139376040 Chr.14: 91232636-91232637 on Build GRCh38
rs928603026 Chr.14: 91232486-91232486 on Build GRCh38
rs1007888436 Chr.14: 91232707-91232707 on Build GRCh38
rs1052153158 Chr.14: 91232571-91232571 on Build GRCh38
rs890817651 Chr.14: 91232688-91232688 on Build GRCh38
rs1002215499 Chr.14: 91232806-91232806 on Build GRCh38
rs983413232 Chr.14: 91232854-91232854 on Build GRCh38
rs143064270 Chr.14: 91232546-91232546 on Build GRCh38
rs115978184 Chr.14: 91232823-91232823 on Build GRCh38
rs73334637 Chr.14: 91232610-91232610 on Build GRCh38
rs1021771523 Chr.14: 91232763-91232763 on Build GRCh38
rs774061186 Chr.14: 91232503-91232503 on Build GRCh38
rs573427519 Chr.14: 91232732-91232732 on Build GRCh38
rs963748370 Chr.14: 91232769-91232769 on Build GRCh38
rs373581126 Chr.14: 91232653-91232653 on Build GRCh38
rs144110292 Chr.14: 91232795-91232797 on Build GRCh38

View Full Product Details