Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 100567446-100567973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEGAIN
Entrez Gene ID
57596
Gene Name
brain enriched guanylate kinase associated
Gene Aliases
-
UniGene
Hs.211751
Gene Chromosome Location
Chr.14: 100537147-100587404 on Build GRCh38
Transcripts
NM_001159531.1 NM_020836.3 XM_005267919.3 XM_005267920.4 XM_005267921.3 XM_011537031.1 XM_017021521.1 XM_017021522.1

SNP Detail

SNP ID Chromosome Location
rs1011948628 Chr.14: 100567546-100567546 on Build GRCh38
rs955202409 Chr.14: 100567787-100567787 on Build GRCh38
rs961458996 Chr.14: 100567485-100567485 on Build GRCh38
rs926045522 Chr.14: 100567514-100567514 on Build GRCh38
rs61993005 Chr.14: 100567853-100567853 on Build GRCh38
rs985369082 Chr.14: 100567545-100567545 on Build GRCh38
rs537836455 Chr.14: 100567635-100567635 on Build GRCh38
rs891813791 Chr.14: 100567837-100567837 on Build GRCh38
rs909235420 Chr.14: 100567675-100567675 on Build GRCh38
rs1044210445 Chr.14: 100567504-100567504 on Build GRCh38
rs957246778 Chr.14: 100567850-100567850 on Build GRCh38
rs976655319 Chr.14: 100567637-100567637 on Build GRCh38
rs986120801 Chr.14: 100567670-100567670 on Build GRCh38
rs912880908 Chr.14: 100567856-100567856 on Build GRCh38
rs566974773 Chr.14: 100567593-100567593 on Build GRCh38
rs985254325 Chr.14: 100567810-100567810 on Build GRCh38
rs945824169 Chr.14: 100567867-100567867 on Build GRCh38
rs773115411 Chr.14: 100567648-100567648 on Build GRCh38
rs912987765 Chr.14: 100567594-100567594 on Build GRCh38
rs539095251 Chr.14: 100567599-100567599 on Build GRCh38
rs956900656 Chr.14: 100567636-100567636 on Build GRCh38
rs981206154 Chr.14: 100567874-100567874 on Build GRCh38
rs929766898 Chr.14: 100567516-100567516 on Build GRCh38
rs111246626 Chr.14: 100567875-100567875 on Build GRCh38
rs576148148 Chr.14: 100567734-100567734 on Build GRCh38
rs534404254 Chr.14: 100567891-100567891 on Build GRCh38
rs552625374 Chr.14: 100567627-100567627 on Build GRCh38
rs569301200 Chr.14: 100567628-100567628 on Build GRCh38
rs185236742 Chr.14: 100567573-100567573 on Build GRCh38
rs911160162 Chr.14: 100567813-100567813 on Build GRCh38
rs546815379 Chr.14: 100567592-100567592 on Build GRCh38
rs1005982806 Chr.14: 100567700-100567700 on Build GRCh38
rs1056217781 Chr.14: 100567530-100567530 on Build GRCh38
rs939523906 Chr.14: 100567885-100567885 on Build GRCh38
rs774792133 Chr.14: 100567931-100567931 on Build GRCh38
rs895412897 Chr.14: 100567892-100567892 on Build GRCh38
rs953576237 Chr.14: 100567655-100567655 on Build GRCh38
rs775702994 Chr.14: 100567878-100567878 on Build GRCh38
rs1016905431 Chr.14: 100567705-100567705 on Build GRCh38
rs1030451530 Chr.14: 100567651-100567651 on Build GRCh38
rs1029945105 Chr.14: 100567780-100567780 on Build GRCh38
rs374438957 Chr.14: 100567898-100567898 on Build GRCh38
rs1055194415 Chr.14: 100567887-100567887 on Build GRCh38
rs896221268 Chr.14: 100567537-100567537 on Build GRCh38
rs994262328 Chr.14: 100567760-100567760 on Build GRCh38
rs769727342 Chr.14: 100567876-100567876 on Build GRCh38
rs769017696 Chr.14: 100567899-100567899 on Build GRCh38
rs922444323 Chr.14: 100567531-100567531 on Build GRCh38

View Full Product Details