Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102348174-102348673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CINP
Entrez Gene ID
51550
Gene Name
cyclin dependent kinase 2 interacting protein
Gene Aliases
-
UniGene
Hs.129634
Gene Chromosome Location
Chr.14: 102341102-102362916 on Build GRCh38
Transcripts
NM_001320046.1 NM_032630.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6238599 c.527C>A p.P176H Chr.14: 102348669-102348669 on Build GRCh38
2025062 c.559C>T p.L187L Chr.14: 102348637-102348637 on Build GRCh38
7160263 c.540G>C p.L180L Chr.14: 102348656-102348656 on Build GRCh38
5754541 c.571T>C p.Y191H Chr.14: 102348625-102348625 on Build GRCh38
4975305 c.616G>C p.E206Q Chr.14: 102348580-102348580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145787596 Chr.14: 102348401-102348401 on Build GRCh38
rs911581863 Chr.14: 102348545-102348545 on Build GRCh38
rs949567559 Chr.14: 102348383-102348383 on Build GRCh38
rs148982679 Chr.14: 102348415-102348415 on Build GRCh38
rs777147776 Chr.14: 102348533-102348533 on Build GRCh38
rs771267047 Chr.14: 102348528-102348528 on Build GRCh38
rs7012 Chr.14: 102348608-102348608 on Build GRCh38
rs558109503 Chr.14: 102348482-102348482 on Build GRCh38
rs1042687416 Chr.14: 102348384-102348384 on Build GRCh38
rs778021096 Chr.14: 102348614-102348614 on Build GRCh38
rs377018111 Chr.14: 102348567-102348567 on Build GRCh38
rs761746819 Chr.14: 102348568-102348568 on Build GRCh38
rs3210470 Chr.14: 102348444-102348444 on Build GRCh38
rs1054010821 Chr.14: 102348407-102348407 on Build GRCh38
rs758143734 Chr.14: 102348579-102348580 on Build GRCh38
rs867960947 Chr.14: 102348218-102348218 on Build GRCh38
rs969501140 Chr.14: 102348417-102348417 on Build GRCh38
rs1047409 Chr.14: 102348402-102348402 on Build GRCh38
rs57672078 Chr.14: 102348452-102348453 on Build GRCh38
rs983388865 Chr.14: 102348488-102348488 on Build GRCh38
rs918061068 Chr.14: 102348381-102348381 on Build GRCh38
rs535384057 Chr.14: 102348322-102348322 on Build GRCh38
rs879534943 Chr.14: 102348267-102348267 on Build GRCh38
rs3177582 Chr.14: 102348291-102348291 on Build GRCh38
rs1051480900 Chr.14: 102348379-102348379 on Build GRCh38
rs755948267 Chr.14: 102348588-102348588 on Build GRCh38
rs776164958 Chr.14: 102348563-102348563 on Build GRCh38
rs369642134 Chr.14: 102348539-102348539 on Build GRCh38
rs1052619501 Chr.14: 102348388-102348388 on Build GRCh38
rs7141141 Chr.14: 102348361-102348361 on Build GRCh38
rs961247847 Chr.14: 102348499-102348499 on Build GRCh38
rs567041580 Chr.14: 102348449-102348449 on Build GRCh38
rs1063204 Chr.14: 102348350-102348350 on Build GRCh38
rs747187772 Chr.14: 102348617-102348617 on Build GRCh38
rs139659344 Chr.14: 102348556-102348556 on Build GRCh38
rs150019678 Chr.14: 102348557-102348557 on Build GRCh38
rs1032758966 Chr.14: 102348298-102348298 on Build GRCh38
rs907809797 Chr.14: 102348498-102348498 on Build GRCh38
rs550809996 Chr.14: 102348378-102348378 on Build GRCh38
rs370724342 Chr.14: 102348363-102348363 on Build GRCh38
rs758475418 Chr.14: 102348613-102348613 on Build GRCh38
rs1028134120 Chr.14: 102348312-102348312 on Build GRCh38
rs767370292 Chr.14: 102348572-102348572 on Build GRCh38
rs574917230 Chr.14: 102348494-102348494 on Build GRCh38
rs3179846 Chr.14: 102348286-102348286 on Build GRCh38
rs969865087 Chr.14: 102348251-102348251 on Build GRCh38
rs1008475083 Chr.14: 102348462-102348462 on Build GRCh38
rs115491292 Chr.14: 102348307-102348307 on Build GRCh38
rs567803918 Chr.14: 102348400-102348400 on Build GRCh38
rs560193804 Chr.14: 102348316-102348316 on Build GRCh38
rs974060516 Chr.14: 102348341-102348341 on Build GRCh38
rs1063180 Chr.14: 102348441-102348441 on Build GRCh38
rs892907457 Chr.14: 102348410-102348410 on Build GRCh38
rs771320240 Chr.14: 102348626-102348626 on Build GRCh38
rs201362791 Chr.14: 102348448-102348448 on Build GRCh38
rs887164788 Chr.14: 102348627-102348627 on Build GRCh38
rs905350385 Chr.14: 102348618-102348618 on Build GRCh38
rs71470203 Chr.14: 102348451-102348452 on Build GRCh38
rs747415743 Chr.14: 102348521-102348521 on Build GRCh38
rs34125972 Chr.14: 102348449-102348450 on Build GRCh38
rs781473767 Chr.14: 102348644-102348644 on Build GRCh38
rs369989722 Chr.14: 102348450-102348451 on Build GRCh38
rs79109177 Chr.14: 102348540-102348540 on Build GRCh38
rs750196275 Chr.14: 102348558-102348559 on Build GRCh38
rs748940572 Chr.14: 102348546-102348546 on Build GRCh38
rs11544425 Chr.14: 102348354-102348354 on Build GRCh38
rs573841242 Chr.14: 102348448-102348449 on Build GRCh38
rs774801743 Chr.14: 102348555-102348555 on Build GRCh38
rs951345256 Chr.14: 102348483-102348483 on Build GRCh38
rs3187466 Chr.14: 102348348-102348348 on Build GRCh38
rs961408068 Chr.14: 102348323-102348323 on Build GRCh38
rs986763429 Chr.14: 102348319-102348319 on Build GRCh38
rs902882022 Chr.14: 102348275-102348275 on Build GRCh38
rs540481441 Chr.14: 102348308-102348308 on Build GRCh38
rs772240790 Chr.14: 102348518-102348518 on Build GRCh38
rs746395794 Chr.14: 102348653-102348653 on Build GRCh38
rs999762152 Chr.14: 102348382-102348382 on Build GRCh38
rs752606154 Chr.14: 102348360-102348360 on Build GRCh38
rs147053271 Chr.14: 102348480-102348480 on Build GRCh38
rs1003658466 Chr.14: 102348623-102348623 on Build GRCh38
rs750415975 Chr.14: 102348576-102348576 on Build GRCh38
rs371010127 Chr.14: 102348592-102348592 on Build GRCh38
rs777969369 Chr.14: 102348520-102348520 on Build GRCh38
rs973266483 Chr.14: 102348504-102348504 on Build GRCh38

View Full Product Details