Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102355691-102356131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CINP
Entrez Gene ID
51550
Gene Name
cyclin dependent kinase 2 interacting protein
Gene Aliases
-
UniGene
Hs.129634
Gene Chromosome Location
Chr.14: 102341102-102362916 on Build GRCh38
Transcripts
NM_001320046.1 NM_032630.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3793484 c.256G>A p.E86K Chr.14: 102355818-102355818 on Build GRCh38
7276009 c.282A>G p.E94E Chr.14: 102355792-102355792 on Build GRCh38
6445897 c.265G>C p.E89Q Chr.14: 102355809-102355809 on Build GRCh38
340993 c.297G>T p.L99F Chr.14: 102355777-102355777 on Build GRCh38
5381886 c.259G>A p.E87K Chr.14: 102355815-102355815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990234400 Chr.14: 102355878-102355878 on Build GRCh38
rs764705669 Chr.14: 102355819-102355819 on Build GRCh38
rs1025012227 Chr.14: 102355815-102355815 on Build GRCh38
rs546896902 Chr.14: 102355779-102355779 on Build GRCh38
rs192274526 Chr.14: 102355802-102355802 on Build GRCh38
rs985246277 Chr.14: 102355968-102355968 on Build GRCh38
rs780264140 Chr.14: 102356037-102356037 on Build GRCh38
rs918513125 Chr.14: 102356000-102356000 on Build GRCh38
rs543453819 Chr.14: 102356046-102356046 on Build GRCh38
rs778931599 Chr.14: 102355908-102355908 on Build GRCh38
rs926696683 Chr.14: 102355716-102355716 on Build GRCh38
rs768697783 Chr.14: 102355760-102355760 on Build GRCh38
rs755707633 Chr.14: 102355860-102355860 on Build GRCh38
rs759763886 Chr.14: 102355804-102355804 on Build GRCh38
rs780869152 Chr.14: 102355727-102355727 on Build GRCh38
rs183319851 Chr.14: 102355995-102355995 on Build GRCh38
rs756686481 Chr.14: 102355722-102355722 on Build GRCh38
rs776051752 Chr.14: 102355809-102355809 on Build GRCh38
rs750082264 Chr.14: 102355858-102355858 on Build GRCh38
rs953673382 Chr.14: 102355910-102355910 on Build GRCh38
rs779978572 Chr.14: 102355989-102355989 on Build GRCh38
rs948051003 Chr.14: 102356056-102356056 on Build GRCh38
rs887098476 Chr.14: 102355775-102355775 on Build GRCh38
rs372164643 Chr.14: 102355752-102355752 on Build GRCh38
rs113341322 Chr.14: 102355733-102355733 on Build GRCh38
rs1021327653 Chr.14: 102355897-102355897 on Build GRCh38
rs200254040 Chr.14: 102355914-102355914 on Build GRCh38
rs1028783097 Chr.14: 102355950-102355951 on Build GRCh38
rs972552946 Chr.14: 102355996-102355996 on Build GRCh38
rs137866895 Chr.14: 102355753-102355753 on Build GRCh38
rs371261020 Chr.14: 102355776-102355776 on Build GRCh38
rs1048770980 Chr.14: 102356027-102356027 on Build GRCh38
rs150324463 Chr.14: 102355770-102355770 on Build GRCh38
rs779759116 Chr.14: 102355754-102355754 on Build GRCh38
rs770866838 Chr.14: 102355799-102355799 on Build GRCh38
rs377133158 Chr.14: 102355856-102355856 on Build GRCh38
rs577560074 Chr.14: 102355818-102355818 on Build GRCh38
rs749238447 Chr.14: 102355756-102355756 on Build GRCh38
rs749174150 Chr.14: 102355873-102355873 on Build GRCh38
rs188058720 Chr.14: 102356008-102356008 on Build GRCh38
rs377039070 Chr.14: 102355855-102355855 on Build GRCh38
rs773713647 Chr.14: 102355758-102355759 on Build GRCh38
rs912306813 Chr.14: 102355972-102355972 on Build GRCh38
rs779668971 Chr.14: 102355865-102355865 on Build GRCh38
rs749619911 Chr.14: 102355948-102355950 on Build GRCh38
rs188263927 Chr.14: 102355720-102355720 on Build GRCh38
rs370092481 Chr.14: 102355811-102355811 on Build GRCh38
rs374077748 Chr.14: 102355840-102355840 on Build GRCh38
rs745926196 Chr.14: 102355932-102355932 on Build GRCh38
rs765707110 Chr.14: 102355807-102355807 on Build GRCh38
rs754706369 Chr.14: 102355903-102355903 on Build GRCh38
rs771137713 Chr.14: 102355917-102355917 on Build GRCh38
rs999668006 Chr.14: 102355839-102355839 on Build GRCh38
rs545768150 Chr.14: 102356106-102356106 on Build GRCh38
rs531066267 Chr.14: 102355714-102355714 on Build GRCh38
rs149463338 Chr.14: 102356081-102356081 on Build GRCh38

View Full Product Details